Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43576610-43577105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO10
Entrez Gene ID
55129
Gene Name
anoctamin 10
Gene Aliases
SCAR10, TMEM16K
UniGene
Hs.656657
Gene Chromosome Location
Chr.3: 43366324-43691594 on Build GRCh38
Transcripts
NM_001204831.1 NM_001204832.1 NM_001204833.1 NM_001204834.1 NM_018075.3 XM_011533885.2 XM_011533887.2 XM_011533889.2 XM_011533890.2 XM_017006716.1 XM_017006717.1 XM_017006718.1 XM_017006719.1 XM_017006720.1 XM_017006721.1 XM_017006722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6468909 c.980A>G p.Y327C Chr.3: 43576874-43576874 on Build GRCh38
3066688 c.954C>T p.S318S Chr.3: 43576900-43576900 on Build GRCh38
3365183 c.1090A>G p.I364V Chr.3: 43576764-43576764 on Build GRCh38
4149909 c.1050C>T p.S350S Chr.3: 43576804-43576804 on Build GRCh38
4456944 c.1028C>T p.A343V Chr.3: 43576826-43576826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745434303 Chr.3: 43576677-43576677 on Build GRCh38
rs372215383 Chr.3: 43577052-43577052 on Build GRCh38
rs758961704 Chr.3: 43576878-43576878 on Build GRCh38
rs369488032 Chr.3: 43577067-43577067 on Build GRCh38
rs768594040 Chr.3: 43576954-43576954 on Build GRCh38
rs769678648 Chr.3: 43577000-43577000 on Build GRCh38
rs794726680 Chr.3: 43576703-43576704 on Build GRCh38
rs760973973 Chr.3: 43576755-43576755 on Build GRCh38
rs200840765 Chr.3: 43577076-43577076 on Build GRCh38
rs890815374 Chr.3: 43576708-43576708 on Build GRCh38
rs756881754 Chr.3: 43576999-43576999 on Build GRCh38
rs373827579 Chr.3: 43576843-43576843 on Build GRCh38
rs777030076 Chr.3: 43576965-43576965 on Build GRCh38
rs78839506 Chr.3: 43576811-43576811 on Build GRCh38
rs1024962328 Chr.3: 43576865-43576865 on Build GRCh38
rs967906623 Chr.3: 43576978-43576978 on Build GRCh38
rs779618110 Chr.3: 43576787-43576787 on Build GRCh38
rs187964933 Chr.3: 43576666-43576666 on Build GRCh38
rs773720901 Chr.3: 43576973-43576973 on Build GRCh38
rs768484828 Chr.3: 43576955-43576955 on Build GRCh38
rs767953054 Chr.3: 43577082-43577082 on Build GRCh38
rs754095732 Chr.3: 43576871-43576871 on Build GRCh38
rs780706796 Chr.3: 43576826-43576826 on Build GRCh38
rs187425614 Chr.3: 43576842-43576842 on Build GRCh38
rs149196477 Chr.3: 43576891-43576891 on Build GRCh38
rs139077997 Chr.3: 43576890-43576890 on Build GRCh38
rs372441963 Chr.3: 43576656-43576656 on Build GRCh38
rs201479117 Chr.3: 43576917-43576917 on Build GRCh38
rs868729445 Chr.3: 43577026-43577026 on Build GRCh38
rs193181511 Chr.3: 43576667-43576667 on Build GRCh38
rs143141504 Chr.3: 43576804-43576804 on Build GRCh38
rs776868518 Chr.3: 43577045-43577045 on Build GRCh38
rs764440861 Chr.3: 43576868-43576868 on Build GRCh38
rs376195257 Chr.3: 43576930-43576930 on Build GRCh38
rs1009027652 Chr.3: 43576953-43576953 on Build GRCh38
rs746449970 Chr.3: 43576805-43576805 on Build GRCh38
rs746710432 Chr.3: 43576688-43576688 on Build GRCh38
rs146569520 Chr.3: 43576874-43576874 on Build GRCh38
rs184756780 Chr.3: 43577012-43577012 on Build GRCh38
rs373948580 Chr.3: 43576773-43576773 on Build GRCh38
rs773557659 Chr.3: 43577062-43577062 on Build GRCh38
rs767044966 Chr.3: 43577001-43577001 on Build GRCh38
rs775651909 Chr.3: 43576864-43576864 on Build GRCh38
rs144272231 Chr.3: 43576710-43576710 on Build GRCh38
rs375470443 Chr.3: 43577064-43577064 on Build GRCh38
rs371063079 Chr.3: 43576988-43576988 on Build GRCh38
rs41289586 Chr.3: 43577066-43577066 on Build GRCh38
rs760603889 Chr.3: 43576867-43576867 on Build GRCh38
rs768148483 Chr.3: 43576691-43576691 on Build GRCh38
rs926487903 Chr.3: 43576757-43576757 on Build GRCh38
rs761532493 Chr.3: 43576719-43576719 on Build GRCh38
rs757689043 Chr.3: 43576775-43576775 on Build GRCh38
rs571306863 Chr.3: 43576994-43576994 on Build GRCh38
rs377487986 Chr.3: 43576876-43576876 on Build GRCh38
rs144349000 Chr.3: 43576970-43576970 on Build GRCh38
rs763801421 Chr.3: 43576638-43576638 on Build GRCh38
rs762313080 Chr.3: 43576872-43576872 on Build GRCh38
rs774148896 Chr.3: 43576941-43576941 on Build GRCh38
rs768362659 Chr.3: 43576807-43576807 on Build GRCh38
rs775049285 Chr.3: 43576685-43576685 on Build GRCh38
rs201848805 Chr.3: 43576999-43576999 on Build GRCh38
rs143322256 Chr.3: 43577055-43577055 on Build GRCh38
rs370331216 Chr.3: 43576841-43576841 on Build GRCh38
rs374795191 Chr.3: 43577017-43577017 on Build GRCh38
rs754348871 Chr.3: 43576874-43576875 on Build GRCh38
rs772092273 Chr.3: 43577039-43577039 on Build GRCh38
rs767603679 Chr.3: 43576743-43576743 on Build GRCh38
rs61732728 Chr.3: 43576721-43576721 on Build GRCh38
rs551675434 Chr.3: 43576680-43576680 on Build GRCh38
rs778230480 Chr.3: 43576660-43576660 on Build GRCh38
rs752941925 Chr.3: 43576653-43576653 on Build GRCh38
rs192531429 Chr.3: 43577004-43577004 on Build GRCh38
rs757883422 Chr.3: 43576803-43576803 on Build GRCh38
rs553236835 Chr.3: 43576836-43576836 on Build GRCh38
rs747996814 Chr.3: 43576827-43576827 on Build GRCh38
rs780361433 Chr.3: 43576887-43576887 on Build GRCh38
rs537457363 Chr.3: 43576731-43576731 on Build GRCh38
rs370018426 Chr.3: 43576770-43576770 on Build GRCh38
rs370984977 Chr.3: 43576903-43576903 on Build GRCh38
rs748978132 Chr.3: 43576933-43576933 on Build GRCh38
rs756986703 Chr.3: 43577019-43577019 on Build GRCh38
rs755048076 Chr.3: 43576645-43576645 on Build GRCh38
rs1055034872 Chr.3: 43576733-43576733 on Build GRCh38
rs770904407 Chr.3: 43576934-43576934 on Build GRCh38
rs375157457 Chr.3: 43576997-43576997 on Build GRCh38
rs773121159 Chr.3: 43576722-43576722 on Build GRCh38
rs368194194 Chr.3: 43576961-43576961 on Build GRCh38
rs959496061 Chr.3: 43576816-43576816 on Build GRCh38
rs1046226208 Chr.3: 43576821-43576821 on Build GRCh38
rs763225867 Chr.3: 43576977-43576977 on Build GRCh38
rs367711525 Chr.3: 43577021-43577021 on Build GRCh38
rs371487223 Chr.3: 43576668-43576668 on Build GRCh38
rs760863716 Chr.3: 43576943-43576943 on Build GRCh38
rs777566392 Chr.3: 43576923-43576923 on Build GRCh38
rs752679400 Chr.3: 43576751-43576751 on Build GRCh38
rs970747480 Chr.3: 43576869-43576869 on Build GRCh38
rs56389778 Chr.3: 43576788-43576788 on Build GRCh38
rs747769148 Chr.3: 43576916-43576916 on Build GRCh38

View Full Product Details