Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43580274-43580772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO10
Entrez Gene ID
55129
Gene Name
anoctamin 10
Gene Aliases
SCAR10, TMEM16K
UniGene
Hs.656657
Gene Chromosome Location
Chr.3: 43366324-43691594 on Build GRCh38
Transcripts
NM_001204831.1 NM_001204832.1 NM_001204833.1 NM_001204834.1 NM_018075.3 XM_011533885.2 XM_011533887.2 XM_011533889.2 XM_011533890.2 XM_017006716.1 XM_017006717.1 XM_017006718.1 XM_017006719.1 XM_017006720.1 XM_017006721.1 XM_017006722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1245101 c.578A>C p.K193T Chr.3: 43580367-43580367 on Build GRCh38
730819 c.489G>A p.T163T Chr.3: 43580456-43580456 on Build GRCh38
4917212 c.578A>G p.K193R Chr.3: 43580367-43580367 on Build GRCh38
7206105 c.558G>T p.W186C Chr.3: 43580387-43580387 on Build GRCh38
5059737 c.486C>T p.L162L Chr.3: 43580459-43580459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753932398 Chr.3: 43580316-43580316 on Build GRCh38
rs768283501 Chr.3: 43580345-43580346 on Build GRCh38
rs34829628 Chr.3: 43580459-43580459 on Build GRCh38
rs558073768 Chr.3: 43580608-43580609 on Build GRCh38
rs376573390 Chr.3: 43580613-43580613 on Build GRCh38
rs769319255 Chr.3: 43580374-43580374 on Build GRCh38
rs759370845 Chr.3: 43580413-43580413 on Build GRCh38
rs565735406 Chr.3: 43580729-43580729 on Build GRCh38
rs775510692 Chr.3: 43580350-43580350 on Build GRCh38
rs750449971 Chr.3: 43580422-43580422 on Build GRCh38
rs750502916 Chr.3: 43580522-43580522 on Build GRCh38
rs758630215 Chr.3: 43580432-43580432 on Build GRCh38
rs775106408 Chr.3: 43580508-43580508 on Build GRCh38
rs929698404 Chr.3: 43580654-43580654 on Build GRCh38
rs527484493 Chr.3: 43580296-43580296 on Build GRCh38
rs1020440038 Chr.3: 43580730-43580730 on Build GRCh38
rs150160564 Chr.3: 43580356-43580356 on Build GRCh38
rs758463386 Chr.3: 43580524-43580524 on Build GRCh38
rs549123500 Chr.3: 43580720-43580720 on Build GRCh38
rs770151711 Chr.3: 43580456-43580456 on Build GRCh38
rs878968729 Chr.3: 43580612-43580612 on Build GRCh38
rs759590184 Chr.3: 43580608-43580608 on Build GRCh38
rs1035915918 Chr.3: 43580651-43580654 on Build GRCh38
rs927159908 Chr.3: 43580582-43580582 on Build GRCh38
rs766071472 Chr.3: 43580391-43580391 on Build GRCh38
rs779051667 Chr.3: 43580345-43580345 on Build GRCh38
rs200844849 Chr.3: 43580442-43580442 on Build GRCh38
rs901778139 Chr.3: 43580498-43580498 on Build GRCh38
rs113290332 Chr.3: 43580567-43580567 on Build GRCh38
rs960297779 Chr.3: 43580657-43580657 on Build GRCh38
rs980006503 Chr.3: 43580572-43580572 on Build GRCh38
rs746849526 Chr.3: 43580343-43580343 on Build GRCh38
rs777186155 Chr.3: 43580357-43580357 on Build GRCh38
rs888723786 Chr.3: 43580302-43580302 on Build GRCh38
rs1048090013 Chr.3: 43580661-43580661 on Build GRCh38
rs774131560 Chr.3: 43580470-43580470 on Build GRCh38
rs184416837 Chr.3: 43580478-43580478 on Build GRCh38
rs113133966 Chr.3: 43580689-43580689 on Build GRCh38
rs757346231 Chr.3: 43580341-43580341 on Build GRCh38
rs1047536742 Chr.3: 43580530-43580530 on Build GRCh38
rs780415367 Chr.3: 43580352-43580352 on Build GRCh38
rs781118336 Chr.3: 43580311-43580311 on Build GRCh38
rs752657568 Chr.3: 43580420-43580420 on Build GRCh38
rs901350192 Chr.3: 43580392-43580392 on Build GRCh38
rs1039935664 Chr.3: 43580741-43580741 on Build GRCh38
rs767412205 Chr.3: 43580417-43580417 on Build GRCh38
rs1048039125 Chr.3: 43580297-43580297 on Build GRCh38
rs774293364 Chr.3: 43580411-43580411 on Build GRCh38
rs759296175 Chr.3: 43580476-43580476 on Build GRCh38
rs747313286 Chr.3: 43580355-43580355 on Build GRCh38
rs534405750 Chr.3: 43580740-43580740 on Build GRCh38
rs189337526 Chr.3: 43580673-43580673 on Build GRCh38
rs201340219 Chr.3: 43580457-43580457 on Build GRCh38
rs993045333 Chr.3: 43580622-43580622 on Build GRCh38
rs776942971 Chr.3: 43580573-43580573 on Build GRCh38
rs781235297 Chr.3: 43580410-43580411 on Build GRCh38
rs762719135 Chr.3: 43580384-43580384 on Build GRCh38
rs746058345 Chr.3: 43580347-43580347 on Build GRCh38
rs781594739 Chr.3: 43580447-43580447 on Build GRCh38
rs373835376 Chr.3: 43580515-43580515 on Build GRCh38
rs148873732 Chr.3: 43580379-43580379 on Build GRCh38
rs199834409 Chr.3: 43580520-43580520 on Build GRCh38
rs757320803 Chr.3: 43580421-43580421 on Build GRCh38
rs755350106 Chr.3: 43580446-43580446 on Build GRCh38
rs747434303 Chr.3: 43580441-43580441 on Build GRCh38
rs367866407 Chr.3: 43580637-43580637 on Build GRCh38
rs938528412 Chr.3: 43580589-43580589 on Build GRCh38
rs373386030 Chr.3: 43580433-43580433 on Build GRCh38
rs748723990 Chr.3: 43580451-43580451 on Build GRCh38
rs747663849 Chr.3: 43580366-43580366 on Build GRCh38

View Full Product Details