Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44244678-44245183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOPAZ1
Entrez Gene ID
375337
Gene Name
testis and ovary specific PAZ domain containing 1
Gene Aliases
C3orf77
UniGene
Hs.132125
Gene Chromosome Location
Chr.3: 44241886-44337071 on Build GRCh38
Transcripts
NM_001145030.1 XM_011533694.2 XM_017006361.1 XM_017006362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4482418 c.2602C>T p.Q868* Chr.3: 44245108-44245108 on Build GRCh38
5675337 c.2452A>G p.S818G Chr.3: 44244958-44244958 on Build GRCh38
6019041 c.2456C>T p.P819L Chr.3: 44244962-44244962 on Build GRCh38
6528873 c.2514T>C p.G838G Chr.3: 44245020-44245020 on Build GRCh38
7272873 c.2301T>C p.Y767Y Chr.3: 44244807-44244807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773393055 Chr.3: 44244786-44244786 on Build GRCh38
rs767936264 Chr.3: 44244849-44244849 on Build GRCh38
rs866510884 Chr.3: 44245087-44245087 on Build GRCh38
rs888100240 Chr.3: 44244854-44244854 on Build GRCh38
rs114357122 Chr.3: 44244715-44244715 on Build GRCh38
rs1046733569 Chr.3: 44244836-44244836 on Build GRCh38
rs753838971 Chr.3: 44245002-44245002 on Build GRCh38
rs895817604 Chr.3: 44244745-44244745 on Build GRCh38
rs771851044 Chr.3: 44245088-44245088 on Build GRCh38
rs188815711 Chr.3: 44244826-44244826 on Build GRCh38
rs72868354 Chr.3: 44244718-44244718 on Build GRCh38
rs913586833 Chr.3: 44244846-44244846 on Build GRCh38
rs545097523 Chr.3: 44244939-44244939 on Build GRCh38
rs956955843 Chr.3: 44245017-44245017 on Build GRCh38
rs753046514 Chr.3: 44244881-44244881 on Build GRCh38
rs755135973 Chr.3: 44244831-44244831 on Build GRCh38
rs1003678849 Chr.3: 44244993-44244993 on Build GRCh38
rs375236858 Chr.3: 44244707-44244707 on Build GRCh38
rs766444748 Chr.3: 44244808-44244808 on Build GRCh38
rs555235098 Chr.3: 44244721-44244721 on Build GRCh38
rs975560923 Chr.3: 44244793-44244793 on Build GRCh38
rs199685520 Chr.3: 44244981-44244981 on Build GRCh38
rs775374791 Chr.3: 44244813-44244813 on Build GRCh38
rs937897218 Chr.3: 44244890-44244890 on Build GRCh38
rs778855845 Chr.3: 44245028-44245031 on Build GRCh38
rs867949259 Chr.3: 44245123-44245123 on Build GRCh38
rs192078606 Chr.3: 44245139-44245139 on Build GRCh38
rs895871731 Chr.3: 44244928-44244928 on Build GRCh38
rs745523571 Chr.3: 44245086-44245086 on Build GRCh38
rs372746432 Chr.3: 44244876-44244876 on Build GRCh38
rs1017503988 Chr.3: 44244782-44244782 on Build GRCh38
rs201546531 Chr.3: 44244764-44244764 on Build GRCh38
rs1029847805 Chr.3: 44245023-44245023 on Build GRCh38
rs184690990 Chr.3: 44244749-44244749 on Build GRCh38
rs557599206 Chr.3: 44244755-44244755 on Build GRCh38
rs1014306948 Chr.3: 44244971-44244971 on Build GRCh38
rs17076545 Chr.3: 44244892-44244892 on Build GRCh38
rs955082784 Chr.3: 44245028-44245028 on Build GRCh38
rs1043677119 Chr.3: 44244877-44244877 on Build GRCh38
rs759211253 Chr.3: 44244708-44244708 on Build GRCh38
rs933734674 Chr.3: 44244806-44244806 on Build GRCh38
rs574173534 Chr.3: 44244790-44244790 on Build GRCh38
rs770653177 Chr.3: 44244702-44244702 on Build GRCh38
rs768679655 Chr.3: 44245152-44245152 on Build GRCh38
rs996990136 Chr.3: 44245012-44245012 on Build GRCh38
rs1017221269 Chr.3: 44244998-44244998 on Build GRCh38
rs867618797 Chr.3: 44245040-44245040 on Build GRCh38
rs184177771 Chr.3: 44245140-44245140 on Build GRCh38
rs984408647 Chr.3: 44244767-44244767 on Build GRCh38
rs865911905 Chr.3: 44244744-44244744 on Build GRCh38
rs564499009 Chr.3: 44244995-44244995 on Build GRCh38
rs540807772 Chr.3: 44244956-44244956 on Build GRCh38
rs1023786791 Chr.3: 44244899-44244899 on Build GRCh38
rs951603265 Chr.3: 44244766-44244766 on Build GRCh38
rs775361024 Chr.3: 44244729-44244729 on Build GRCh38
rs1047027472 Chr.3: 44244974-44244974 on Build GRCh38
rs756660950 Chr.3: 44244889-44244889 on Build GRCh38
rs370707986 Chr.3: 44244794-44244794 on Build GRCh38
rs796218259 Chr.3: 44244861-44244861 on Build GRCh38

View Full Product Details