Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44338628-44339141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCAIM
Entrez Gene ID
285343
Gene Name
T-cell activation inhibitor, mitochondrial
Gene Aliases
C3orf23, TOAG-1, TOAG1
UniGene
Hs.55131
Gene Chromosome Location
Chr.3: 44338119-44409448 on Build GRCh38
Transcripts
NM_001029839.2 NM_001029840.2 NM_001282913.1 NM_001282914.1 NM_001282915.1 NM_173826.3 XM_017006251.1

SNP Detail

SNP ID Chromosome Location
rs544761032 Chr.3: 44338873-44338873 on Build GRCh38
rs138606382 Chr.3: 44339106-44339107 on Build GRCh38
rs937918228 Chr.3: 44338732-44338732 on Build GRCh38
rs1000220857 Chr.3: 44338782-44338782 on Build GRCh38
rs112471617 Chr.3: 44338661-44338661 on Build GRCh38
rs774257863 Chr.3: 44338989-44338989 on Build GRCh38
rs926269961 Chr.3: 44338731-44338731 on Build GRCh38
rs1001041315 Chr.3: 44338947-44338947 on Build GRCh38
rs909014240 Chr.3: 44338839-44338839 on Build GRCh38
rs576899499 Chr.3: 44338757-44338757 on Build GRCh38
rs182363913 Chr.3: 44338822-44338822 on Build GRCh38
rs11130025 Chr.3: 44338716-44338716 on Build GRCh38
rs569001444 Chr.3: 44338711-44338711 on Build GRCh38
rs899805330 Chr.3: 44338749-44338749 on Build GRCh38
rs979170144 Chr.3: 44338799-44338799 on Build GRCh38
rs575761919 Chr.3: 44338795-44338795 on Build GRCh38
rs1033456337 Chr.3: 44338960-44338960 on Build GRCh38
rs1042957631 Chr.3: 44338888-44338888 on Build GRCh38
rs1038433709 Chr.3: 44338859-44338859 on Build GRCh38
rs754043208 Chr.3: 44338704-44338704 on Build GRCh38
rs770462353 Chr.3: 44338972-44338972 on Build GRCh38
rs13086084 Chr.3: 44338996-44338996 on Build GRCh38
rs903991769 Chr.3: 44338892-44338892 on Build GRCh38
rs772290482 Chr.3: 44338835-44338835 on Build GRCh38
rs71325020 Chr.3: 44339032-44339032 on Build GRCh38
rs929197916 Chr.3: 44338828-44338828 on Build GRCh38
rs1029800062 Chr.3: 44338698-44338698 on Build GRCh38
rs11540403 Chr.3: 44338768-44338768 on Build GRCh38
rs569092974 Chr.3: 44339062-44339062 on Build GRCh38
rs749020517 Chr.3: 44338869-44338869 on Build GRCh38
rs946225843 Chr.3: 44338886-44338886 on Build GRCh38
rs959095986 Chr.3: 44338963-44338963 on Build GRCh38
rs772135412 Chr.3: 44339021-44339021 on Build GRCh38
rs937494730 Chr.3: 44338751-44338751 on Build GRCh38
rs371290367 Chr.3: 44338879-44338879 on Build GRCh38
rs561249285 Chr.3: 44338876-44338876 on Build GRCh38
rs937590173 Chr.3: 44338881-44338881 on Build GRCh38
rs554375930 Chr.3: 44338730-44338730 on Build GRCh38
rs992047213 Chr.3: 44338734-44338734 on Build GRCh38
rs904511544 Chr.3: 44338770-44338770 on Build GRCh38
rs929145381 Chr.3: 44338739-44338739 on Build GRCh38
rs773278731 Chr.3: 44338866-44338866 on Build GRCh38
rs1047580356 Chr.3: 44338741-44338741 on Build GRCh38
rs983727720 Chr.3: 44338829-44338829 on Build GRCh38
rs142129635 Chr.3: 44338891-44338891 on Build GRCh38
rs1029893560 Chr.3: 44338761-44338761 on Build GRCh38
rs1009725812 Chr.3: 44338765-44338765 on Build GRCh38
rs1033775900 Chr.3: 44338803-44338803 on Build GRCh38
rs968204319 Chr.3: 44338781-44338781 on Build GRCh38
rs964558661 Chr.3: 44338695-44338695 on Build GRCh38
rs1041805498 Chr.3: 44338900-44338900 on Build GRCh38
rs921383375 Chr.3: 44339031-44339031 on Build GRCh38
rs747663433 Chr.3: 44338804-44338804 on Build GRCh38
rs975001098 Chr.3: 44338697-44338697 on Build GRCh38
rs186688733 Chr.3: 44338961-44338961 on Build GRCh38
rs757396723 Chr.3: 44338706-44338706 on Build GRCh38
rs983041655 Chr.3: 44338710-44338710 on Build GRCh38
rs954585814 Chr.3: 44338823-44338823 on Build GRCh38
rs747228548 Chr.3: 44338878-44338878 on Build GRCh38
rs552053668 Chr.3: 44339025-44339025 on Build GRCh38
rs950621237 Chr.3: 44338976-44338976 on Build GRCh38
rs564090702 Chr.3: 44338941-44338942 on Build GRCh38
rs991802993 Chr.3: 44338819-44338819 on Build GRCh38
rs147142533 Chr.3: 44338653-44338653 on Build GRCh38
rs1032236735 Chr.3: 44338783-44338783 on Build GRCh38
rs941457391 Chr.3: 44338842-44338842 on Build GRCh38
rs1032702189 Chr.3: 44338689-44338689 on Build GRCh38
rs900378875 Chr.3: 44338950-44338952 on Build GRCh38
rs1009734095 Chr.3: 44338885-44338885 on Build GRCh38
rs140260915 Chr.3: 44338657-44338657 on Build GRCh38
rs35230933 Chr.3: 44339107-44339108 on Build GRCh38
rs1013289892 Chr.3: 44338971-44338971 on Build GRCh38
rs560152674 Chr.3: 44338981-44338981 on Build GRCh38
rs958432071 Chr.3: 44339052-44339052 on Build GRCh38
rs376780175 Chr.3: 44339059-44339059 on Build GRCh38
rs1057485214 Chr.3: 44338752-44338752 on Build GRCh38
rs941416889 Chr.3: 44338747-44338747 on Build GRCh38
rs926947829 Chr.3: 44339045-44339045 on Build GRCh38
rs190883922 Chr.3: 44338746-44338746 on Build GRCh38
rs1000172725 Chr.3: 44338923-44338923 on Build GRCh38
rs1057406235 Chr.3: 44338883-44338883 on Build GRCh38
rs115800254 Chr.3: 44338737-44338737 on Build GRCh38
rs959002878 Chr.3: 44338810-44338810 on Build GRCh38
rs997076179 Chr.3: 44338758-44338758 on Build GRCh38
rs887224019 Chr.3: 44338742-44338742 on Build GRCh38
rs200045773 Chr.3: 44339108-44339108 on Build GRCh38
rs1016926581 Chr.3: 44338656-44338656 on Build GRCh38
rs955517814 Chr.3: 44338705-44338705 on Build GRCh38
rs63237460 Chr.3: 44339108-44339109 on Build GRCh38
rs530607747 Chr.3: 44338762-44338762 on Build GRCh38
rs963104204 Chr.3: 44339030-44339030 on Build GRCh38
rs552041433 Chr.3: 44338658-44338658 on Build GRCh38
rs904626296 Chr.3: 44338901-44338901 on Build GRCh38
rs973051842 Chr.3: 44339038-44339038 on Build GRCh38
rs553839921 Chr.3: 44338748-44338748 on Build GRCh38
rs1030583413 Chr.3: 44338813-44338814 on Build GRCh38
rs567764197 Chr.3: 44338936-44338936 on Build GRCh38
rs963268888 Chr.3: 44339015-44339015 on Build GRCh38
rs1051039508 Chr.3: 44338875-44338875 on Build GRCh38

View Full Product Details