Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44361225-44361770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCAIM
Entrez Gene ID
285343
Gene Name
T-cell activation inhibitor, mitochondrial
Gene Aliases
C3orf23, TOAG-1, TOAG1
UniGene
Hs.55131
Gene Chromosome Location
Chr.3: 44338119-44409448 on Build GRCh38
Transcripts
NM_001282913.1 NM_001282914.1 NM_001282915.1 NM_173826.3 XM_017006251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
97903 c.205C>G p.L69V Chr.3: 44361404-44361404 on Build GRCh38
3916072 c.287C>T p.S96F Chr.3: 44361486-44361486 on Build GRCh38
4964120 c.304C>A p.P102T Chr.3: 44361503-44361503 on Build GRCh38
1044970 c.305C>A p.P102H Chr.3: 44361504-44361504 on Build GRCh38
1044969 c.182C>A p.S61Y Chr.3: 44361381-44361381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566997027 Chr.3: 44361677-44361677 on Build GRCh38
rs138023221 Chr.3: 44361483-44361483 on Build GRCh38
rs768749322 Chr.3: 44361353-44361353 on Build GRCh38
rs773087133 Chr.3: 44361320-44361320 on Build GRCh38
rs900243852 Chr.3: 44361421-44361421 on Build GRCh38
rs140545796 Chr.3: 44361462-44361462 on Build GRCh38
rs773880659 Chr.3: 44361372-44361372 on Build GRCh38
rs554076889 Chr.3: 44361280-44361280 on Build GRCh38
rs768546718 Chr.3: 44361255-44361255 on Build GRCh38
rs752828374 Chr.3: 44361443-44361452 on Build GRCh38
rs773717585 Chr.3: 44361414-44361414 on Build GRCh38
rs773948872 Chr.3: 44361518-44361518 on Build GRCh38
rs758995122 Chr.3: 44361522-44361522 on Build GRCh38
rs747324392 Chr.3: 44361349-44361349 on Build GRCh38
rs748496571 Chr.3: 44361361-44361361 on Build GRCh38
rs541906831 Chr.3: 44361428-44361428 on Build GRCh38
rs201319681 Chr.3: 44361338-44361338 on Build GRCh38
rs917897441 Chr.3: 44361282-44361282 on Build GRCh38
rs771316907 Chr.3: 44361511-44361511 on Build GRCh38
rs763817761 Chr.3: 44361457-44361457 on Build GRCh38
rs546911035 Chr.3: 44361621-44361621 on Build GRCh38
rs760252931 Chr.3: 44361530-44361530 on Build GRCh38
rs759279046 Chr.3: 44361416-44361416 on Build GRCh38
rs754579592 Chr.3: 44361321-44361321 on Build GRCh38
rs776278530 Chr.3: 44361391-44361391 on Build GRCh38
rs751569832 Chr.3: 44361476-44361476 on Build GRCh38
rs200880941 Chr.3: 44361367-44361367 on Build GRCh38
rs770594255 Chr.3: 44361393-44361396 on Build GRCh38
rs77421305 Chr.3: 44361599-44361599 on Build GRCh38
rs552392509 Chr.3: 44361687-44361687 on Build GRCh38
rs775136046 Chr.3: 44361523-44361523 on Build GRCh38
rs745826992 Chr.3: 44361705-44361705 on Build GRCh38
rs751240937 Chr.3: 44361320-44361320 on Build GRCh38
rs926722102 Chr.3: 44361731-44361731 on Build GRCh38
rs770538593 Chr.3: 44361406-44361406 on Build GRCh38
rs767355387 Chr.3: 44361322-44361322 on Build GRCh38
rs1021539405 Chr.3: 44361596-44361596 on Build GRCh38
rs748335218 Chr.3: 44361490-44361490 on Build GRCh38
rs756081222 Chr.3: 44361334-44361334 on Build GRCh38
rs779929954 Chr.3: 44361346-44361346 on Build GRCh38
rs956193476 Chr.3: 44361612-44361612 on Build GRCh38
rs149806706 Chr.3: 44361634-44361634 on Build GRCh38
rs372782572 Chr.3: 44361456-44361456 on Build GRCh38
rs562657130 Chr.3: 44361345-44361345 on Build GRCh38
rs752463758 Chr.3: 44361324-44361324 on Build GRCh38
rs749862334 Chr.3: 44361499-44361499 on Build GRCh38
rs761860289 Chr.3: 44361559-44361559 on Build GRCh38
rs1021707822 Chr.3: 44361686-44361686 on Build GRCh38
rs766360583 Chr.3: 44361422-44361439 on Build GRCh38
rs778219002 Chr.3: 44361493-44361493 on Build GRCh38
rs756221615 Chr.3: 44361456-44361463 on Build GRCh38
rs770299753 Chr.3: 44361491-44361491 on Build GRCh38
rs143382247 Chr.3: 44361449-44361449 on Build GRCh38
rs879048957 Chr.3: 44361344-44361344 on Build GRCh38
rs189405681 Chr.3: 44361474-44361474 on Build GRCh38
rs777185456 Chr.3: 44361354-44361354 on Build GRCh38
rs182038497 Chr.3: 44361644-44361644 on Build GRCh38
rs186558072 Chr.3: 44361275-44361275 on Build GRCh38
rs577135292 Chr.3: 44361314-44361314 on Build GRCh38
rs781601407 Chr.3: 44361485-44361485 on Build GRCh38
rs373365719 Chr.3: 44361541-44361541 on Build GRCh38
rs765761035 Chr.3: 44361316-44361316 on Build GRCh38
rs753563301 Chr.3: 44361542-44361542 on Build GRCh38
rs1003156954 Chr.3: 44361584-44361584 on Build GRCh38
rs386660606 Chr.3: 44361670-44361671 on Build GRCh38

View Full Product Details