Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45901901-45902395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR9
Entrez Gene ID
10803
Gene Name
C-C motif chemokine receptor 9
Gene Aliases
CC-CKR-9, CDw199, GPR-9-6, GPR28
UniGene
Hs.225946
Gene Chromosome Location
Chr.3: 45886504-45903177 on Build GRCh38
Transcripts
NM_001256369.1 NM_006641.3 NM_031200.2 XM_011533312.2 NM_001276378.1 NM_001276379.1 XM_017006644.1 XM_017006645.1

Gene Symbol
LZTFL1
Entrez Gene ID
54585
Gene Name
leucine zipper transcription factor like 1
Gene Aliases
BBS17
UniGene
Hs.30824
Gene Chromosome Location
Chr.3: 45823316-45916042 on Build GRCh38
Transcripts
NM_001256369.1 NM_006641.3 NM_031200.2 XM_011533312.2 NM_001276378.1 NM_001276379.1 XM_017006644.1 XM_017006645.1

SNP Detail

SNP ID Chromosome Location
rs74718475 Chr.3: 45902245-45902245 on Build GRCh38
rs144027662 Chr.3: 45902346-45902346 on Build GRCh38
rs200911894 Chr.3: 45902211-45902211 on Build GRCh38
rs201275143 Chr.3: 45902282-45902282 on Build GRCh38
rs201332768 Chr.3: 45902141-45902141 on Build GRCh38
rs943973879 Chr.3: 45902031-45902031 on Build GRCh38
rs200141830 Chr.3: 45902227-45902227 on Build GRCh38
rs200518653 Chr.3: 45902055-45902055 on Build GRCh38
rs1057134027 Chr.3: 45902110-45902110 on Build GRCh38
rs908588026 Chr.3: 45902336-45902336 on Build GRCh38
rs980319068 Chr.3: 45902118-45902118 on Build GRCh38
rs952339946 Chr.3: 45902079-45902079 on Build GRCh38
rs201908125 Chr.3: 45902177-45902177 on Build GRCh38
rs889902659 Chr.3: 45902194-45902194 on Build GRCh38
rs201599717 Chr.3: 45902213-45902213 on Build GRCh38
rs1057439575 Chr.3: 45902348-45902348 on Build GRCh38
rs200823591 Chr.3: 45902176-45902176 on Build GRCh38
rs199551466 Chr.3: 45902131-45902131 on Build GRCh38
rs202166025 Chr.3: 45902294-45902294 on Build GRCh38
rs148423835 Chr.3: 45902221-45902221 on Build GRCh38
rs975377287 Chr.3: 45902303-45902303 on Build GRCh38
rs200275306 Chr.3: 45901959-45901959 on Build GRCh38
rs952393284 Chr.3: 45902304-45902304 on Build GRCh38
rs201700560 Chr.3: 45901949-45901949 on Build GRCh38
rs201397521 Chr.3: 45902061-45902061 on Build GRCh38
rs1032024641 Chr.3: 45902193-45902193 on Build GRCh38
rs904723640 Chr.3: 45902171-45902171 on Build GRCh38
rs913243975 Chr.3: 45902146-45902146 on Build GRCh38
rs73830610 Chr.3: 45902327-45902327 on Build GRCh38
rs1013201763 Chr.3: 45902114-45902114 on Build GRCh38
rs201544941 Chr.3: 45902106-45902106 on Build GRCh38
rs201744927 Chr.3: 45902253-45902253 on Build GRCh38
rs1009608565 Chr.3: 45902242-45902242 on Build GRCh38
rs201818079 Chr.3: 45901968-45901968 on Build GRCh38
rs199982423 Chr.3: 45902179-45902179 on Build GRCh38
rs17765088 Chr.3: 45902103-45902103 on Build GRCh38
rs199642231 Chr.3: 45902371-45902371 on Build GRCh38
rs1005150701 Chr.3: 45902308-45902308 on Build GRCh38
rs200925805 Chr.3: 45901960-45901960 on Build GRCh38
rs975619767 Chr.3: 45902044-45902044 on Build GRCh38
rs780814289 Chr.3: 45902024-45902024 on Build GRCh38
rs896022849 Chr.3: 45902113-45902113 on Build GRCh38
rs202110703 Chr.3: 45902328-45902328 on Build GRCh38
rs577271905 Chr.3: 45902119-45902119 on Build GRCh38
rs1044541340 Chr.3: 45902162-45902162 on Build GRCh38
rs1025301402 Chr.3: 45902027-45902027 on Build GRCh38
rs552970086 Chr.3: 45902180-45902180 on Build GRCh38
rs201238517 Chr.3: 45902341-45902341 on Build GRCh38
rs200044856 Chr.3: 45902259-45902259 on Build GRCh38
rs200919915 Chr.3: 45902372-45902372 on Build GRCh38
rs200207613 Chr.3: 45902087-45902087 on Build GRCh38
rs939992030 Chr.3: 45902105-45902105 on Build GRCh38
rs532262664 Chr.3: 45902370-45902370 on Build GRCh38
rs200524594 Chr.3: 45902138-45902138 on Build GRCh38
rs965669036 Chr.3: 45902274-45902274 on Build GRCh38
rs921136224 Chr.3: 45902064-45902064 on Build GRCh38
rs755084615 Chr.3: 45901928-45901928 on Build GRCh38
rs199683998 Chr.3: 45902145-45902145 on Build GRCh38
rs200420405 Chr.3: 45902340-45902340 on Build GRCh38
rs185636686 Chr.3: 45902309-45902309 on Build GRCh38
rs866460583 Chr.3: 45902369-45902369 on Build GRCh38
rs931365666 Chr.3: 45902080-45902080 on Build GRCh38
rs199576272 Chr.3: 45902100-45902100 on Build GRCh38

View Full Product Details