Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45936343-45936846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYCO1
Entrez Gene ID
79443
Gene Name
FYVE and coiled-coil domain containing 1
Gene Aliases
CATC2, CTRCT18, RUFY3, ZFYVE7
UniGene
Hs.200227
Gene Chromosome Location
Chr.3: 45917899-45995824 on Build GRCh38
Transcripts
NM_024513.3 XM_006713333.3 XM_006713334.3 XM_011534111.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6188966 c.3985G>T p.D1329Y Chr.3: 45936503-45936503 on Build GRCh38
1734624 c.3968C>T p.T1323M Chr.3: 45936520-45936520 on Build GRCh38
1423344 c.3994G>A p.V1332M Chr.3: 45936494-45936494 on Build GRCh38
6668075 c.3993C>T p.P1331P Chr.3: 45936495-45936495 on Build GRCh38
6468926 c.4020G>A p.L1340L Chr.3: 45936468-45936468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916903213 Chr.3: 45936726-45936726 on Build GRCh38
rs1037661887 Chr.3: 45936431-45936431 on Build GRCh38
rs747333799 Chr.3: 45936468-45936468 on Build GRCh38
rs778851398 Chr.3: 45936457-45936457 on Build GRCh38
rs764629141 Chr.3: 45936417-45936417 on Build GRCh38
rs761357698 Chr.3: 45936475-45936475 on Build GRCh38
rs766380757 Chr.3: 45936487-45936487 on Build GRCh38
rs762546630 Chr.3: 45936485-45936485 on Build GRCh38
rs758204376 Chr.3: 45936424-45936424 on Build GRCh38
rs889819531 Chr.3: 45936690-45936690 on Build GRCh38
rs900649388 Chr.3: 45936806-45936806 on Build GRCh38
rs750005616 Chr.3: 45936423-45936423 on Build GRCh38
rs1873002 Chr.3: 45936407-45936407 on Build GRCh38
rs4356851 Chr.3: 45936481-45936481 on Build GRCh38
rs565705534 Chr.3: 45936607-45936607 on Build GRCh38
rs558285442 Chr.3: 45936757-45936757 on Build GRCh38
rs757559324 Chr.3: 45936455-45936455 on Build GRCh38
rs763683129 Chr.3: 45936408-45936408 on Build GRCh38
rs370189051 Chr.3: 45936437-45936437 on Build GRCh38
rs760880027 Chr.3: 45936502-45936502 on Build GRCh38
rs751515767 Chr.3: 45936489-45936489 on Build GRCh38
rs915071195 Chr.3: 45936569-45936569 on Build GRCh38
rs368641714 Chr.3: 45936532-45936532 on Build GRCh38
rs769284834 Chr.3: 45936555-45936555 on Build GRCh38
rs769208216 Chr.3: 45936537-45936537 on Build GRCh38
rs781404910 Chr.3: 45936542-45936542 on Build GRCh38
rs201278075 Chr.3: 45936529-45936529 on Build GRCh38
rs369642169 Chr.3: 45936665-45936665 on Build GRCh38
rs187028124 Chr.3: 45936411-45936411 on Build GRCh38
rs752606656 Chr.3: 45936495-45936495 on Build GRCh38
rs1056801446 Chr.3: 45936445-45936445 on Build GRCh38
rs766434247 Chr.3: 45936427-45936427 on Build GRCh38
rs377724709 Chr.3: 45936400-45936400 on Build GRCh38
rs538689578 Chr.3: 45936530-45936530 on Build GRCh38
rs954701649 Chr.3: 45936558-45936558 on Build GRCh38
rs754851907 Chr.3: 45936440-45936440 on Build GRCh38
rs113318190 Chr.3: 45936520-45936520 on Build GRCh38
rs753989630 Chr.3: 45936453-45936453 on Build GRCh38
rs772397410 Chr.3: 45936463-45936463 on Build GRCh38
rs761565992 Chr.3: 45936414-45936414 on Build GRCh38
rs189457139 Chr.3: 45936700-45936700 on Build GRCh38
rs879185522 Chr.3: 45936776-45936776 on Build GRCh38
rs750701071 Chr.3: 45936782-45936782 on Build GRCh38
rs758529962 Chr.3: 45936799-45936799 on Build GRCh38
rs541032458 Chr.3: 45936378-45936378 on Build GRCh38
rs4535265 Chr.3: 45936374-45936374 on Build GRCh38
rs757351709 Chr.3: 45936519-45936519 on Build GRCh38
rs748560471 Chr.3: 45936543-45936543 on Build GRCh38
rs945729092 Chr.3: 45936744-45936744 on Build GRCh38
rs140677166 Chr.3: 45936484-45936484 on Build GRCh38
rs749183217 Chr.3: 45936566-45936566 on Build GRCh38
rs753687109 Chr.3: 45936585-45936585 on Build GRCh38
rs575370963 Chr.3: 45936755-45936755 on Build GRCh38
rs758500527 Chr.3: 45936526-45936526 on Build GRCh38
rs1042247826 Chr.3: 45936614-45936614 on Build GRCh38
rs570228625 Chr.3: 45936465-45936465 on Build GRCh38
rs576225940 Chr.3: 45936681-45936681 on Build GRCh38
rs9865446 Chr.3: 45936651-45936651 on Build GRCh38
rs927219104 Chr.3: 45936667-45936667 on Build GRCh38
rs181217402 Chr.3: 45936723-45936723 on Build GRCh38
rs143007267 Chr.3: 45936760-45936760 on Build GRCh38
rs147523930 Chr.3: 45936802-45936802 on Build GRCh38
rs777027209 Chr.3: 45936474-45936474 on Build GRCh38
rs547953154 Chr.3: 45936525-45936525 on Build GRCh38
rs185605979 Chr.3: 45936767-45936767 on Build GRCh38
rs536794812 Chr.3: 45936480-45936480 on Build GRCh38
rs895452494 Chr.3: 45936451-45936451 on Build GRCh38
rs764276901 Chr.3: 45936564-45936565 on Build GRCh38
rs768977392 Chr.3: 45936469-45936469 on Build GRCh38
rs9864861 Chr.3: 45936513-45936513 on Build GRCh38
rs781176882 Chr.3: 45936448-45936448 on Build GRCh38
rs934107220 Chr.3: 45936680-45936680 on Build GRCh38
rs541165323 Chr.3: 45936762-45936762 on Build GRCh38
rs553896769 Chr.3: 45936565-45936565 on Build GRCh38
rs963578135 Chr.3: 45936645-45936645 on Build GRCh38
rs759599621 Chr.3: 45936494-45936494 on Build GRCh38
rs201698951 Chr.3: 45936462-45936462 on Build GRCh38
rs535850389 Chr.3: 45936612-45936612 on Build GRCh38

View Full Product Details