Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46359898-46360396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR2
Entrez Gene ID
729230
Gene Name
C-C motif chemokine receptor 2
Gene Aliases
CC-CKR-2, CCR-2, CCR2A, CCR2B, CD192, CKR2, CKR2A, CKR2B, CMKBR2, MCP-1-R
UniGene
Hs.511794
Gene Chromosome Location
Chr.3: 46353744-46360940 on Build GRCh38
Transcripts
NM_001123041.2 XM_011534069.1

SNP Detail

SNP ID Chromosome Location
rs543643209 Chr.3: 46359963-46359963 on Build GRCh38
rs944187274 Chr.3: 46360219-46360219 on Build GRCh38
rs559285648 Chr.3: 46360100-46360100 on Build GRCh38
rs549688502 Chr.3: 46360265-46360265 on Build GRCh38
rs761778888 Chr.3: 46360061-46360061 on Build GRCh38
rs960516072 Chr.3: 46360086-46360086 on Build GRCh38
rs1035803279 Chr.3: 46360067-46360067 on Build GRCh38
rs1023448289 Chr.3: 46360297-46360297 on Build GRCh38
rs372263390 Chr.3: 46360039-46360039 on Build GRCh38
rs887129801 Chr.3: 46360039-46360039 on Build GRCh38
rs887090155 Chr.3: 46360296-46360296 on Build GRCh38
rs140253702 Chr.3: 46360115-46360115 on Build GRCh38
rs574098686 Chr.3: 46360027-46360027 on Build GRCh38
rs1042100 Chr.3: 46360132-46360132 on Build GRCh38
rs1039858709 Chr.3: 46360246-46360246 on Build GRCh38
rs1048659789 Chr.3: 46360271-46360271 on Build GRCh38
rs866260614 Chr.3: 46360342-46360342 on Build GRCh38
rs41430746 Chr.3: 46360155-46360155 on Build GRCh38
rs887867491 Chr.3: 46359946-46359946 on Build GRCh38
rs188527018 Chr.3: 46360370-46360370 on Build GRCh38
rs765832227 Chr.3: 46360112-46360112 on Build GRCh38
rs912247496 Chr.3: 46360208-46360208 on Build GRCh38
rs1022954434 Chr.3: 46360021-46360021 on Build GRCh38
rs900126553 Chr.3: 46359968-46359968 on Build GRCh38
rs887889298 Chr.3: 46360294-46360295 on Build GRCh38
rs956415162 Chr.3: 46360194-46360194 on Build GRCh38
rs979324442 Chr.3: 46360160-46360160 on Build GRCh38
rs1057460916 Chr.3: 46360359-46360359 on Build GRCh38
rs371779665 Chr.3: 46360344-46360344 on Build GRCh38
rs1039963838 Chr.3: 46359967-46359967 on Build GRCh38
rs1031564546 Chr.3: 46360057-46360057 on Build GRCh38
rs995763974 Chr.3: 46360008-46360008 on Build GRCh38
rs184336934 Chr.3: 46360320-46360320 on Build GRCh38
rs553718224 Chr.3: 46360190-46360190 on Build GRCh38
rs1049213422 Chr.3: 46360293-46360293 on Build GRCh38
rs766867608 Chr.3: 46360372-46360372 on Build GRCh38
rs368842919 Chr.3: 46359928-46359928 on Build GRCh38
rs1002841311 Chr.3: 46360350-46360350 on Build GRCh38
rs914339831 Chr.3: 46360146-46360146 on Build GRCh38
rs768542505 Chr.3: 46360324-46360324 on Build GRCh38
rs988275731 Chr.3: 46360206-46360206 on Build GRCh38
rs181429510 Chr.3: 46360179-46360179 on Build GRCh38
rs763451379 Chr.3: 46360214-46360214 on Build GRCh38
rs541727099 Chr.3: 46360047-46360047 on Build GRCh38
rs577442510 Chr.3: 46360040-46360040 on Build GRCh38
rs761598058 Chr.3: 46360147-46360147 on Build GRCh38
rs1031631905 Chr.3: 46360351-46360351 on Build GRCh38
rs768656901 Chr.3: 46359975-46359975 on Build GRCh38
rs3918388 Chr.3: 46360250-46360250 on Build GRCh38
rs565506818 Chr.3: 46359988-46359988 on Build GRCh38
rs1004245965 Chr.3: 46360343-46360343 on Build GRCh38

View Full Product Details