Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47579804-47580308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSPG5
Entrez Gene ID
10675
Gene Name
chondroitin sulfate proteoglycan 5
Gene Aliases
NGC
UniGene
Hs.45127
Gene Chromosome Location
Chr.3: 47562238-47580240 on Build GRCh38
Transcripts
NM_001206942.1 NM_001206945.1

SNP Detail

SNP ID Chromosome Location
rs535108788 Chr.3: 47579917-47579917 on Build GRCh38
rs980456060 Chr.3: 47580136-47580136 on Build GRCh38
rs759261607 Chr.3: 47580101-47580101 on Build GRCh38
rs894645852 Chr.3: 47579927-47579927 on Build GRCh38
rs970885728 Chr.3: 47580132-47580132 on Build GRCh38
rs1033394898 Chr.3: 47580200-47580200 on Build GRCh38
rs571003251 Chr.3: 47579861-47579861 on Build GRCh38
rs1027396214 Chr.3: 47579925-47579925 on Build GRCh38
rs901906575 Chr.3: 47579933-47579933 on Build GRCh38
rs535431736 Chr.3: 47580199-47580199 on Build GRCh38
rs774272516 Chr.3: 47579842-47579842 on Build GRCh38
rs932728586 Chr.3: 47580022-47580022 on Build GRCh38
rs933367207 Chr.3: 47579941-47579941 on Build GRCh38
rs944653261 Chr.3: 47579828-47579828 on Build GRCh38
rs945308316 Chr.3: 47580258-47580258 on Build GRCh38
rs556952374 Chr.3: 47580226-47580226 on Build GRCh38
rs1024485970 Chr.3: 47579937-47579937 on Build GRCh38
rs553226050 Chr.3: 47579945-47579945 on Build GRCh38
rs911746582 Chr.3: 47580068-47580068 on Build GRCh38
rs889087039 Chr.3: 47579957-47579957 on Build GRCh38
rs1024872136 Chr.3: 47580242-47580242 on Build GRCh38
rs192138451 Chr.3: 47580045-47580045 on Build GRCh38
rs1006227771 Chr.3: 47579997-47579997 on Build GRCh38
rs1032073090 Chr.3: 47579964-47579964 on Build GRCh38
rs897512874 Chr.3: 47579915-47579915 on Build GRCh38
rs976713301 Chr.3: 47580277-47580277 on Build GRCh38
rs1016509451 Chr.3: 47579998-47579998 on Build GRCh38
rs114942810 Chr.3: 47580230-47580230 on Build GRCh38
rs771616822 Chr.3: 47580001-47580001 on Build GRCh38
rs993664267 Chr.3: 47580097-47580097 on Build GRCh38
rs777304601 Chr.3: 47580149-47580149 on Build GRCh38
rs913804050 Chr.3: 47580247-47580247 on Build GRCh38
rs1031960943 Chr.3: 47580279-47580279 on Build GRCh38
rs1036414566 Chr.3: 47579860-47579860 on Build GRCh38
rs1025541099 Chr.3: 47580109-47580109 on Build GRCh38

View Full Product Details