Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47817940-47818448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX30
Entrez Gene ID
22907
Gene Name
DEAH-box helicase 30
Gene Aliases
DDX30, RETCOR
UniGene
Hs.517948
Gene Chromosome Location
Chr.3: 47802909-47850196 on Build GRCh38
Transcripts
NM_014966.3 NM_138615.2 XM_006713033.1 XM_011533490.1 XM_011533491.1 XM_011533492.2 XM_011533493.1 XM_011533494.2 XM_011533495.1 XM_011533496.1 XM_011533497.2 XM_011533498.1 XM_017005914.1 XM_017005915.1 XM_017005916.1 XM_017005917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1485269 c.29-1G>T p.? Chr.3: 47818021-47818021 on Build GRCh38
4996351 c.49C>T p.R17C Chr.3: 47818042-47818042 on Build GRCh38
4514578 c.97C>T p.P33S Chr.3: 47818090-47818090 on Build GRCh38
3594804 c.64C>T p.P22S Chr.3: 47818057-47818057 on Build GRCh38
3824034 c.69A>C p.P23P Chr.3: 47818062-47818062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022767942 Chr.3: 47818353-47818353 on Build GRCh38
rs889968447 Chr.3: 47818205-47818205 on Build GRCh38
rs1020042388 Chr.3: 47818111-47818111 on Build GRCh38
rs1026976996 Chr.3: 47818291-47818291 on Build GRCh38
rs981469735 Chr.3: 47818387-47818387 on Build GRCh38
rs773341182 Chr.3: 47817963-47817963 on Build GRCh38
rs553022081 Chr.3: 47818261-47818261 on Build GRCh38
rs750582512 Chr.3: 47818134-47818134 on Build GRCh38
rs138729543 Chr.3: 47818043-47818043 on Build GRCh38
rs772923094 Chr.3: 47818063-47818063 on Build GRCh38
rs972859923 Chr.3: 47817967-47817967 on Build GRCh38
rs763171714 Chr.3: 47817977-47817977 on Build GRCh38
rs988518122 Chr.3: 47818123-47818123 on Build GRCh38
rs765434323 Chr.3: 47818112-47818112 on Build GRCh38
rs767349564 Chr.3: 47818079-47818079 on Build GRCh38
rs758505890 Chr.3: 47818136-47818136 on Build GRCh38
rs750500521 Chr.3: 47818017-47818017 on Build GRCh38
rs981762037 Chr.3: 47818283-47818283 on Build GRCh38
rs372491791 Chr.3: 47817998-47817998 on Build GRCh38
rs1006569107 Chr.3: 47818345-47818345 on Build GRCh38
rs762509799 Chr.3: 47818064-47818064 on Build GRCh38
rs370985134 Chr.3: 47817975-47817975 on Build GRCh38
rs147205504 Chr.3: 47818076-47818076 on Build GRCh38
rs972273000 Chr.3: 47818311-47818311 on Build GRCh38
rs754081951 Chr.3: 47818006-47818006 on Build GRCh38
rs950039381 Chr.3: 47818016-47818016 on Build GRCh38
rs752459812 Chr.3: 47818092-47818092 on Build GRCh38
rs150000907 Chr.3: 47818022-47818022 on Build GRCh38
rs927292637 Chr.3: 47818399-47818399 on Build GRCh38
rs780627833 Chr.3: 47818025-47818025 on Build GRCh38
rs937302906 Chr.3: 47818412-47818412 on Build GRCh38
rs200966986 Chr.3: 47818055-47818055 on Build GRCh38
rs200465272 Chr.3: 47818093-47818093 on Build GRCh38
rs375502665 Chr.3: 47818010-47818010 on Build GRCh38
rs143620188 Chr.3: 47818067-47818067 on Build GRCh38
rs370934742 Chr.3: 47818236-47818236 on Build GRCh38
rs201094375 Chr.3: 47818041-47818041 on Build GRCh38
rs550245344 Chr.3: 47817994-47817994 on Build GRCh38
rs887677293 Chr.3: 47818334-47818334 on Build GRCh38
rs1028595156 Chr.3: 47818248-47818248 on Build GRCh38
rs955806332 Chr.3: 47818402-47818402 on Build GRCh38
rs764094594 Chr.3: 47818359-47818359 on Build GRCh38
rs779006308 Chr.3: 47818014-47818014 on Build GRCh38
rs182159396 Chr.3: 47818202-47818202 on Build GRCh38
rs141621111 Chr.3: 47818061-47818061 on Build GRCh38
rs769538722 Chr.3: 47818047-47818047 on Build GRCh38
rs752134153 Chr.3: 47818162-47818162 on Build GRCh38
rs187951563 Chr.3: 47818256-47818256 on Build GRCh38
rs370921443 Chr.3: 47818066-47818066 on Build GRCh38
rs542793602 Chr.3: 47818267-47818267 on Build GRCh38
rs367902945 Chr.3: 47818166-47818166 on Build GRCh38
rs781720340 Chr.3: 47818169-47818169 on Build GRCh38
rs918714046 Chr.3: 47818347-47818347 on Build GRCh38
rs866782727 Chr.3: 47818396-47818396 on Build GRCh38
rs554029096 Chr.3: 47818206-47818206 on Build GRCh38
rs368342218 Chr.3: 47818042-47818042 on Build GRCh38
rs879589335 Chr.3: 47818411-47818411 on Build GRCh38
rs151115082 Chr.3: 47818026-47818026 on Build GRCh38
rs1041533305 Chr.3: 47818237-47818237 on Build GRCh38
rs953322981 Chr.3: 47818251-47818251 on Build GRCh38
rs571572878 Chr.3: 47818187-47818187 on Build GRCh38
rs767177116 Chr.3: 47817996-47817996 on Build GRCh38
rs751368238 Chr.3: 47817990-47817990 on Build GRCh38
rs779962682 Chr.3: 47818150-47818150 on Build GRCh38
rs753449176 Chr.3: 47818401-47818401 on Build GRCh38

View Full Product Details