Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47828782-47829282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX30
Entrez Gene ID
22907
Gene Name
DEAH-box helicase 30
Gene Aliases
DDX30, RETCOR
UniGene
Hs.517948
Gene Chromosome Location
Chr.3: 47802909-47850196 on Build GRCh38
Transcripts
NM_014966.3 NM_138615.2 XM_006713033.1 XM_011533490.1 XM_011533491.1 XM_011533492.2 XM_011533493.1 XM_011533494.2 XM_011533495.1 XM_011533496.1 XM_011533497.2 XM_011533498.1 XM_017005914.1 XM_017005915.1 XM_017005916.1 XM_017005917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5736597 c.279G>A p.W93* Chr.3: 47829047-47829047 on Build GRCh38
3660588 c.278G>A p.W93* Chr.3: 47829046-47829046 on Build GRCh38
4677367 c.332G>A p.R111Q Chr.3: 47829100-47829100 on Build GRCh38
7091093 c.322G>T p.D108Y Chr.3: 47829090-47829090 on Build GRCh38
1495555 c.340G>A p.A114T Chr.3: 47829108-47829108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749080250 Chr.3: 47829169-47829169 on Build GRCh38
rs780715407 Chr.3: 47829128-47829128 on Build GRCh38
rs979549921 Chr.3: 47828993-47828993 on Build GRCh38
rs921182114 Chr.3: 47829176-47829176 on Build GRCh38
rs749651481 Chr.3: 47828942-47828942 on Build GRCh38
rs751067047 Chr.3: 47828996-47828996 on Build GRCh38
rs540205815 Chr.3: 47828829-47828829 on Build GRCh38
rs773755567 Chr.3: 47828881-47828881 on Build GRCh38
rs766005533 Chr.3: 47829107-47829107 on Build GRCh38
rs995261046 Chr.3: 47828957-47828957 on Build GRCh38
rs559240930 Chr.3: 47828915-47828915 on Build GRCh38
rs931159995 Chr.3: 47829177-47829177 on Build GRCh38
rs372318144 Chr.3: 47828980-47828980 on Build GRCh38
rs939859236 Chr.3: 47828927-47828927 on Build GRCh38
rs750759641 Chr.3: 47829074-47829074 on Build GRCh38
rs974801771 Chr.3: 47829173-47829173 on Build GRCh38
rs769508026 Chr.3: 47829012-47829012 on Build GRCh38
rs775769512 Chr.3: 47829035-47829035 on Build GRCh38
rs150305804 Chr.3: 47829119-47829119 on Build GRCh38
rs755002768 Chr.3: 47828998-47828998 on Build GRCh38
rs772418426 Chr.3: 47829033-47829033 on Build GRCh38
rs887851175 Chr.3: 47828865-47828865 on Build GRCh38
rs375325678 Chr.3: 47829140-47829140 on Build GRCh38
rs746132966 Chr.3: 47829032-47829032 on Build GRCh38
rs550300573 Chr.3: 47828910-47828911 on Build GRCh38
rs777575503 Chr.3: 47829029-47829029 on Build GRCh38
rs754452740 Chr.3: 47829125-47829125 on Build GRCh38
rs534158789 Chr.3: 47828974-47828974 on Build GRCh38
rs954727420 Chr.3: 47829246-47829246 on Build GRCh38
rs375356846 Chr.3: 47829089-47829089 on Build GRCh38
rs765566224 Chr.3: 47829057-47829057 on Build GRCh38
rs1014942566 Chr.3: 47828938-47828938 on Build GRCh38
rs1048246295 Chr.3: 47829194-47829194 on Build GRCh38
rs1036892016 Chr.3: 47828928-47828928 on Build GRCh38
rs898216873 Chr.3: 47828952-47828952 on Build GRCh38
rs11540154 Chr.3: 47829106-47829106 on Build GRCh38
rs969359081 Chr.3: 47828911-47828925 on Build GRCh38
rs772918782 Chr.3: 47828977-47828978 on Build GRCh38
rs115079845 Chr.3: 47828969-47828969 on Build GRCh38
rs748053221 Chr.3: 47829009-47829009 on Build GRCh38
rs146413167 Chr.3: 47829218-47829218 on Build GRCh38
rs770790520 Chr.3: 47829183-47829183 on Build GRCh38
rs969568333 Chr.3: 47828991-47828991 on Build GRCh38
rs114352152 Chr.3: 47829237-47829237 on Build GRCh38
rs528516311 Chr.3: 47828828-47828828 on Build GRCh38
rs568772701 Chr.3: 47829226-47829226 on Build GRCh38
rs762329546 Chr.3: 47829056-47829056 on Build GRCh38
rs529899508 Chr.3: 47828946-47828946 on Build GRCh38
rs569880162 Chr.3: 47828981-47828981 on Build GRCh38
rs777031954 Chr.3: 47829050-47829050 on Build GRCh38
rs559122964 Chr.3: 47829203-47829203 on Build GRCh38
rs34723020 Chr.3: 47829223-47829223 on Build GRCh38
rs187470747 Chr.3: 47828999-47828999 on Build GRCh38
rs943651852 Chr.3: 47829148-47829148 on Build GRCh38
rs752599829 Chr.3: 47829131-47829131 on Build GRCh38
rs116727997 Chr.3: 47829166-47829166 on Build GRCh38
rs768712167 Chr.3: 47829044-47829044 on Build GRCh38

View Full Product Details