Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48446626-48447132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRIP
Entrez Gene ID
84126
Gene Name
ATR interacting protein
Gene Aliases
-
UniGene
Hs.694840
Gene Chromosome Location
Chr.3: 48446710-48465655 on Build GRCh38
Transcripts
NM_001271022.1 NM_001271023.1 NM_032166.3 NM_130384.2 XM_011534113.2

Gene Symbol
CCDC51
Entrez Gene ID
79714
Gene Name
coiled-coil domain containing 51
Gene Aliases
-
UniGene
Hs.187657
Gene Chromosome Location
Chr.3: 48432170-48446652 on Build GRCh38
Transcripts
NM_001271022.1 NM_001271023.1 NM_032166.3 NM_130384.2 XM_011534113.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4149944 c.73G>A p.G25S Chr.3: 48446918-48446918 on Build GRCh38
1683065 c.190G>T p.D64Y Chr.3: 48447035-48447035 on Build GRCh38
4664604 c.235C>T p.R79W Chr.3: 48447080-48447080 on Build GRCh38
6608495 c.47C>T p.P16L Chr.3: 48446892-48446892 on Build GRCh38
4890936 c.215G>C p.S72T Chr.3: 48447060-48447060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012863852 Chr.3: 48446660-48446660 on Build GRCh38
rs368960700 Chr.3: 48446956-48446956 on Build GRCh38
rs780920662 Chr.3: 48446937-48446937 on Build GRCh38
rs754712526 Chr.3: 48446860-48446860 on Build GRCh38
rs765815524 Chr.3: 48446896-48446896 on Build GRCh38
rs762360097 Chr.3: 48446895-48446895 on Build GRCh38
rs555253846 Chr.3: 48446790-48446790 on Build GRCh38
rs772976477 Chr.3: 48447041-48447041 on Build GRCh38
rs746715933 Chr.3: 48447053-48447053 on Build GRCh38
rs764497220 Chr.3: 48446889-48446889 on Build GRCh38
rs1012978179 Chr.3: 48446786-48446786 on Build GRCh38
rs766536731 Chr.3: 48446929-48446929 on Build GRCh38
rs375145275 Chr.3: 48446703-48446703 on Build GRCh38
rs930915112 Chr.3: 48446672-48446672 on Build GRCh38
rs779847347 Chr.3: 48447049-48447049 on Build GRCh38
rs777660123 Chr.3: 48446947-48446947 on Build GRCh38
rs748138008 Chr.3: 48446805-48446805 on Build GRCh38
rs746534842 Chr.3: 48446783-48446783 on Build GRCh38
rs940307660 Chr.3: 48446971-48446971 on Build GRCh38
rs978671238 Chr.3: 48446675-48446675 on Build GRCh38
rs1022995429 Chr.3: 48446665-48446665 on Build GRCh38
rs371479359 Chr.3: 48446808-48446808 on Build GRCh38
rs755229433 Chr.3: 48446934-48446934 on Build GRCh38
rs901907438 Chr.3: 48446880-48446880 on Build GRCh38
rs140238210 Chr.3: 48446777-48446777 on Build GRCh38
rs879507964 Chr.3: 48446697-48446697 on Build GRCh38
rs757688146 Chr.3: 48446877-48446877 on Build GRCh38
rs780106776 Chr.3: 48446855-48446855 on Build GRCh38
rs895943765 Chr.3: 48446653-48446653 on Build GRCh38
rs947277468 Chr.3: 48447051-48447051 on Build GRCh38
rs369530148 Chr.3: 48446992-48446992 on Build GRCh38
rs1031583048 Chr.3: 48446810-48446810 on Build GRCh38
rs749993354 Chr.3: 48446838-48446839 on Build GRCh38
rs752960407 Chr.3: 48447019-48447019 on Build GRCh38
rs72622933 Chr.3: 48446788-48446788 on Build GRCh38
rs769758692 Chr.3: 48446871-48446871 on Build GRCh38
rs148174212 Chr.3: 48447062-48447062 on Build GRCh38
rs758553037 Chr.3: 48446837-48446837 on Build GRCh38
rs116253015 Chr.3: 48446745-48446745 on Build GRCh38
rs200647747 Chr.3: 48446976-48446976 on Build GRCh38
rs745447161 Chr.3: 48446964-48446964 on Build GRCh38
rs934428841 Chr.3: 48446791-48446791 on Build GRCh38
rs770346954 Chr.3: 48446989-48446989 on Build GRCh38
rs530772573 Chr.3: 48446930-48446930 on Build GRCh38
rs1000190732 Chr.3: 48447040-48447040 on Build GRCh38
rs1052959213 Chr.3: 48446729-48446729 on Build GRCh38
rs750978381 Chr.3: 48446903-48446903 on Build GRCh38
rs765368123 Chr.3: 48446827-48446827 on Build GRCh38
rs981049663 Chr.3: 48447018-48447018 on Build GRCh38
rs1008794343 Chr.3: 48447045-48447045 on Build GRCh38
rs747320253 Chr.3: 48446859-48446859 on Build GRCh38
rs372273511 Chr.3: 48446973-48446973 on Build GRCh38
rs760919772 Chr.3: 48446817-48446817 on Build GRCh38
rs551289572 Chr.3: 48447048-48447048 on Build GRCh38
rs993623433 Chr.3: 48446915-48446915 on Build GRCh38
rs370892766 Chr.3: 48447108-48447108 on Build GRCh38
rs754385585 Chr.3: 48446820-48446820 on Build GRCh38
rs757852591 Chr.3: 48446821-48446821 on Build GRCh38
rs947274808 Chr.3: 48446814-48446814 on Build GRCh38
rs547105412 Chr.3: 48446972-48446972 on Build GRCh38
rs758297238 Chr.3: 48447066-48447066 on Build GRCh38
rs1057266367 Chr.3: 48446784-48446784 on Build GRCh38
rs1045494700 Chr.3: 48446700-48446700 on Build GRCh38
rs1032014234 Chr.3: 48446699-48446699 on Build GRCh38
rs951709373 Chr.3: 48446898-48446898 on Build GRCh38
rs535759859 Chr.3: 48446753-48446753 on Build GRCh38
rs145611247 Chr.3: 48446815-48446815 on Build GRCh38
rs745361845 Chr.3: 48447044-48447044 on Build GRCh38
rs895892689 Chr.3: 48446785-48446785 on Build GRCh38
rs778500392 Chr.3: 48446958-48446958 on Build GRCh38
rs375574173 Chr.3: 48446770-48446770 on Build GRCh38
rs549435972 Chr.3: 48446809-48446809 on Build GRCh38
rs773872853 Chr.3: 48446885-48446885 on Build GRCh38
rs764633605 Chr.3: 48446830-48446830 on Build GRCh38
rs562482620 Chr.3: 48446819-48446819 on Build GRCh38
rs766298876 Chr.3: 48446998-48446998 on Build GRCh38
rs759293280 Chr.3: 48446887-48446887 on Build GRCh38
rs928347666 Chr.3: 48446789-48446789 on Build GRCh38
rs778246445 Chr.3: 48447043-48447043 on Build GRCh38
rs201645015 Chr.3: 48446779-48446779 on Build GRCh38
rs771487037 Chr.3: 48447069-48447069 on Build GRCh38
rs755955663 Chr.3: 48446943-48446943 on Build GRCh38
rs964484171 Chr.3: 48446742-48446742 on Build GRCh38
rs529538750 Chr.3: 48447038-48447038 on Build GRCh38
rs767707232 Chr.3: 48447014-48447014 on Build GRCh38
rs372906254 Chr.3: 48447074-48447074 on Build GRCh38
rs966126838 Chr.3: 48446778-48446778 on Build GRCh38
rs545796791 Chr.3: 48446938-48446938 on Build GRCh38
rs767653932 Chr.3: 48447085-48447085 on Build GRCh38
rs759584758 Chr.3: 48447084-48447084 on Build GRCh38
rs908791210 Chr.3: 48446969-48446969 on Build GRCh38
rs557664424 Chr.3: 48446760-48446760 on Build GRCh38
rs751671447 Chr.3: 48446932-48446932 on Build GRCh38
rs749317099 Chr.3: 48446949-48446949 on Build GRCh38
rs955972773 Chr.3: 48446813-48446813 on Build GRCh38
rs955817971 Chr.3: 48446701-48446701 on Build GRCh38
rs755900581 Chr.3: 48447020-48447020 on Build GRCh38
rs943957532 Chr.3: 48447086-48447086 on Build GRCh38
rs776253376 Chr.3: 48446863-48446863 on Build GRCh38
rs1011506873 Chr.3: 48446757-48446757 on Build GRCh38
rs763526910 Chr.3: 48446993-48446993 on Build GRCh38
rs570205560 Chr.3: 48446765-48446765 on Build GRCh38
rs969141304 Chr.3: 48446674-48446674 on Build GRCh38
rs759709434 Chr.3: 48447002-48447002 on Build GRCh38
rs762935901 Chr.3: 48446906-48446906 on Build GRCh38
rs776542412 Chr.3: 48446798-48446798 on Build GRCh38
rs757223539 Chr.3: 48447039-48447039 on Build GRCh38
rs911634357 Chr.3: 48446712-48446712 on Build GRCh38
rs528593337 Chr.3: 48446970-48446970 on Build GRCh38
rs900301715 Chr.3: 48447070-48447070 on Build GRCh38
rs1026878004 Chr.3: 48446918-48446918 on Build GRCh38
rs1044447611 Chr.3: 48447007-48447007 on Build GRCh38
rs144998094 Chr.3: 48447064-48447064 on Build GRCh38
rs1035697770 Chr.3: 48446646-48446646 on Build GRCh38
rs777683743 Chr.3: 48447026-48447026 on Build GRCh38
rs748008845 Chr.3: 48446864-48446864 on Build GRCh38
rs770223988 Chr.3: 48447056-48447056 on Build GRCh38
rs539481663 Chr.3: 48446769-48446769 on Build GRCh38
rs750552725 Chr.3: 48446835-48446835 on Build GRCh38
rs777140455 Chr.3: 48446893-48446893 on Build GRCh38
rs996417186 Chr.3: 48447092-48447092 on Build GRCh38

View Full Product Details