Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49269764-49270274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf62
Entrez Gene ID
375341
Gene Name
chromosome 3 open reading frame 62
Gene Aliases
-
UniGene
Hs.403828
Gene Chromosome Location
Chr.3: 49268597-49277075 on Build GRCh38
Transcripts
NM_198562.2

SNP Detail

SNP ID Chromosome Location
rs967220936 Chr.3: 49269873-49269873 on Build GRCh38
rs909225072 Chr.3: 49269995-49269995 on Build GRCh38
rs748916659 Chr.3: 49269894-49269894 on Build GRCh38
rs573267905 Chr.3: 49269844-49269844 on Build GRCh38
rs145829132 Chr.3: 49269986-49269986 on Build GRCh38
rs768013000 Chr.3: 49270178-49270178 on Build GRCh38
rs999664294 Chr.3: 49270108-49270108 on Build GRCh38
rs773812309 Chr.3: 49270163-49270163 on Build GRCh38
rs898106956 Chr.3: 49270113-49270113 on Build GRCh38
rs902207256 Chr.3: 49270025-49270025 on Build GRCh38
rs893574785 Chr.3: 49270162-49270162 on Build GRCh38
rs949544066 Chr.3: 49270132-49270132 on Build GRCh38
rs954441092 Chr.3: 49270224-49270224 on Build GRCh38
rs1054897314 Chr.3: 49269869-49269869 on Build GRCh38
rs932117113 Chr.3: 49269950-49269950 on Build GRCh38
rs551427045 Chr.3: 49269978-49269978 on Build GRCh38
rs919739668 Chr.3: 49269849-49269849 on Build GRCh38
rs942260123 Chr.3: 49269996-49269996 on Build GRCh38
rs527518719 Chr.3: 49270198-49270198 on Build GRCh38
rs1020018370 Chr.3: 49270245-49270248 on Build GRCh38
rs117198029 Chr.3: 49269907-49269907 on Build GRCh38
rs1034058120 Chr.3: 49269788-49269788 on Build GRCh38
rs771909248 Chr.3: 49270107-49270107 on Build GRCh38
rs865907097 Chr.3: 49270081-49270081 on Build GRCh38
rs1011720430 Chr.3: 49270210-49270210 on Build GRCh38
rs987103133 Chr.3: 49270225-49270225 on Build GRCh38
rs770467932 Chr.3: 49270043-49270043 on Build GRCh38
rs1036593599 Chr.3: 49270110-49270110 on Build GRCh38
rs764396186 Chr.3: 49269958-49269961 on Build GRCh38
rs931123283 Chr.3: 49269851-49269851 on Build GRCh38
rs759984571 Chr.3: 49270173-49270173 on Build GRCh38
rs1028499775 Chr.3: 49270220-49270221 on Build GRCh38
rs543815877 Chr.3: 49269876-49269876 on Build GRCh38
rs1013505423 Chr.3: 49269960-49269960 on Build GRCh38
rs149007013 Chr.3: 49269908-49269908 on Build GRCh38
rs990073783 Chr.3: 49269828-49269828 on Build GRCh38
rs957476202 Chr.3: 49269792-49269792 on Build GRCh38

View Full Product Details