Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49271270-49271778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf62
Entrez Gene ID
375341
Gene Name
chromosome 3 open reading frame 62
Gene Aliases
-
UniGene
Hs.403828
Gene Chromosome Location
Chr.3: 49268597-49277075 on Build GRCh38
Transcripts
NM_198562.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1046127 c.554G>A p.R185Q Chr.3: 49271430-49271430 on Build GRCh38
5906711 c.631C>T p.P211S Chr.3: 49271353-49271353 on Build GRCh38
7358552 c.613G>A p.G205S Chr.3: 49271371-49271371 on Build GRCh38
1309257 c.589G>C p.E197Q Chr.3: 49271395-49271395 on Build GRCh38
1046126 c.702C>T p.G234G Chr.3: 49271282-49271282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146973096 Chr.3: 49271310-49271310 on Build GRCh38
rs769802728 Chr.3: 49271414-49271414 on Build GRCh38
rs915131751 Chr.3: 49271379-49271379 on Build GRCh38
rs919026209 Chr.3: 49271552-49271552 on Build GRCh38
rs112490821 Chr.3: 49271559-49271559 on Build GRCh38
rs147619300 Chr.3: 49271315-49271315 on Build GRCh38
rs762330869 Chr.3: 49271331-49271331 on Build GRCh38
rs750560209 Chr.3: 49271385-49271385 on Build GRCh38
rs772367719 Chr.3: 49271293-49271293 on Build GRCh38
rs1036549946 Chr.3: 49271526-49271526 on Build GRCh38
rs1048553382 Chr.3: 49271586-49271587 on Build GRCh38
rs1003536083 Chr.3: 49271693-49271693 on Build GRCh38
rs772802836 Chr.3: 49271321-49271321 on Build GRCh38
rs372595668 Chr.3: 49271390-49271390 on Build GRCh38
rs369633108 Chr.3: 49271299-49271299 on Build GRCh38
rs375740755 Chr.3: 49271452-49271452 on Build GRCh38
rs754140774 Chr.3: 49271373-49271373 on Build GRCh38
rs888712612 Chr.3: 49271300-49271300 on Build GRCh38
rs773865468 Chr.3: 49271695-49271695 on Build GRCh38
rs777513119 Chr.3: 49271487-49271487 on Build GRCh38
rs370398186 Chr.3: 49271387-49271387 on Build GRCh38
rs772999557 Chr.3: 49271423-49271423 on Build GRCh38
rs182288761 Chr.3: 49271617-49271617 on Build GRCh38
rs932989994 Chr.3: 49271442-49271442 on Build GRCh38
rs761093400 Chr.3: 49271366-49271366 on Build GRCh38
rs755984628 Chr.3: 49271461-49271461 on Build GRCh38
rs948348232 Chr.3: 49271629-49271629 on Build GRCh38
rs747905227 Chr.3: 49271413-49271413 on Build GRCh38
rs761847057 Chr.3: 49271311-49271311 on Build GRCh38
rs150294581 Chr.3: 49271382-49271382 on Build GRCh38
rs748885350 Chr.3: 49271425-49271425 on Build GRCh38
rs147201397 Chr.3: 49271612-49271612 on Build GRCh38
rs554857986 Chr.3: 49271641-49271641 on Build GRCh38
rs776780284 Chr.3: 49271308-49271308 on Build GRCh38
rs970836600 Chr.3: 49271320-49271320 on Build GRCh38
rs770585325 Chr.3: 49271439-49271439 on Build GRCh38
rs765825804 Chr.3: 49271463-49271463 on Build GRCh38
rs959608627 Chr.3: 49271708-49271708 on Build GRCh38
rs899016586 Chr.3: 49271576-49271576 on Build GRCh38
rs767104101 Chr.3: 49271437-49271437 on Build GRCh38
rs889036779 Chr.3: 49271551-49271551 on Build GRCh38
rs751632932 Chr.3: 49271491-49271491 on Build GRCh38
rs1003614483 Chr.3: 49271747-49271747 on Build GRCh38
rs138112852 Chr.3: 49271405-49271405 on Build GRCh38
rs747233422 Chr.3: 49271399-49271399 on Build GRCh38
rs770523078 Chr.3: 49271687-49271687 on Build GRCh38
rs758574075 Chr.3: 49271483-49271483 on Build GRCh38
rs766103030 Chr.3: 49271332-49271332 on Build GRCh38
rs748969791 Chr.3: 49271670-49271670 on Build GRCh38
rs1051902703 Chr.3: 49271511-49271511 on Build GRCh38
rs1037464434 Chr.3: 49271568-49271568 on Build GRCh38
rs150162031 Chr.3: 49271371-49271371 on Build GRCh38
rs531564833 Chr.3: 49271620-49271620 on Build GRCh38
rs779246246 Chr.3: 49271377-49271377 on Build GRCh38
rs750883450 Chr.3: 49271474-49271474 on Build GRCh38
rs770343603 Chr.3: 49271430-49271430 on Build GRCh38
rs1026337790 Chr.3: 49271597-49271597 on Build GRCh38
rs773783747 Chr.3: 49271431-49271431 on Build GRCh38
rs780111099 Chr.3: 49271298-49271298 on Build GRCh38
rs993519352 Chr.3: 49271585-49271585 on Build GRCh38
rs1031477749 Chr.3: 49271753-49271753 on Build GRCh38
rs1030822928 Chr.3: 49271368-49271368 on Build GRCh38
rs35091010 Chr.3: 49271527-49271528 on Build GRCh38
rs1004073189 Chr.3: 49271322-49271322 on Build GRCh38
rs755185861 Chr.3: 49271401-49271401 on Build GRCh38
rs1045066865 Chr.3: 49271661-49271661 on Build GRCh38
rs781147988 Chr.3: 49271403-49271403 on Build GRCh38
rs781255438 Chr.3: 49271342-49271342 on Build GRCh38
rs780386279 Chr.3: 49271389-49271389 on Build GRCh38
rs572252527 Chr.3: 49271607-49271607 on Build GRCh38
rs553992351 Chr.3: 49271573-49271573 on Build GRCh38
rs376629875 Chr.3: 49271375-49271375 on Build GRCh38
rs768562055 Chr.3: 49271302-49271302 on Build GRCh38

View Full Product Details