Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49276288-49276810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf62
Entrez Gene ID
375341
Gene Name
chromosome 3 open reading frame 62
Gene Aliases
-
UniGene
Hs.403828
Gene Chromosome Location
Chr.3: 49268597-49277075 on Build GRCh38
Transcripts
NM_198562.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3824149 c.347G>A p.S116N Chr.3: 49276526-49276526 on Build GRCh38
7358551 c.196A>G p.R66G Chr.3: 49276677-49276677 on Build GRCh38
731277 c.85G>T p.D29Y Chr.3: 49276788-49276788 on Build GRCh38
3783726 c.446+1G>T p.? Chr.3: 49276426-49276426 on Build GRCh38
731278 c.171G>A p.S57S Chr.3: 49276702-49276702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868287641 Chr.3: 49276456-49276457 on Build GRCh38
rs746479606 Chr.3: 49276385-49276385 on Build GRCh38
rs537539163 Chr.3: 49276443-49276443 on Build GRCh38
rs759680771 Chr.3: 49276559-49276559 on Build GRCh38
rs780482699 Chr.3: 49276745-49276745 on Build GRCh38
rs932238894 Chr.3: 49276505-49276505 on Build GRCh38
rs780868732 Chr.3: 49276394-49276394 on Build GRCh38
rs766299851 Chr.3: 49276458-49276458 on Build GRCh38
rs537882513 Chr.3: 49276704-49276706 on Build GRCh38
rs760773760 Chr.3: 49276441-49276441 on Build GRCh38
rs1034194250 Chr.3: 49276386-49276386 on Build GRCh38
rs775530374 Chr.3: 49276480-49276483 on Build GRCh38
rs754490851 Chr.3: 49276487-49276487 on Build GRCh38
rs748190744 Chr.3: 49276686-49276686 on Build GRCh38
rs760133374 Chr.3: 49276429-49276429 on Build GRCh38
rs113941560 Chr.3: 49276706-49276706 on Build GRCh38
rs1001332877 Chr.3: 49276355-49276355 on Build GRCh38
rs368027417 Chr.3: 49276326-49276326 on Build GRCh38
rs752204866 Chr.3: 49276635-49276635 on Build GRCh38
rs765399300 Chr.3: 49276596-49276596 on Build GRCh38
rs756790673 Chr.3: 49276742-49276742 on Build GRCh38
rs763891950 Chr.3: 49276663-49276664 on Build GRCh38
rs941082833 Chr.3: 49276550-49276550 on Build GRCh38
rs762705614 Chr.3: 49276603-49276603 on Build GRCh38
rs773022912 Chr.3: 49276598-49276598 on Build GRCh38
rs61749312 Chr.3: 49276695-49276695 on Build GRCh38
rs759097717 Chr.3: 49276404-49276405 on Build GRCh38
rs767159892 Chr.3: 49276477-49276478 on Build GRCh38
rs1024529841 Chr.3: 49276529-49276529 on Build GRCh38
rs771415937 Chr.3: 49276420-49276420 on Build GRCh38
rs148849582 Chr.3: 49276460-49276460 on Build GRCh38
rs752294916 Chr.3: 49276726-49276726 on Build GRCh38
rs767316320 Chr.3: 49276725-49276725 on Build GRCh38
rs760975508 Chr.3: 49276555-49276555 on Build GRCh38
rs534722215 Chr.3: 49276489-49276489 on Build GRCh38
rs767066270 Chr.3: 49276618-49276618 on Build GRCh38
rs770044127 Chr.3: 49276687-49276687 on Build GRCh38
rs1028217662 Chr.3: 49276423-49276423 on Build GRCh38
rs375872034 Chr.3: 49276653-49276653 on Build GRCh38
rs766766358 Chr.3: 49276637-49276637 on Build GRCh38
rs765116060 Chr.3: 49276761-49276761 on Build GRCh38
rs1020358122 Chr.3: 49276720-49276720 on Build GRCh38
rs753561198 Chr.3: 49276695-49276696 on Build GRCh38
rs753533071 Chr.3: 49276738-49276738 on Build GRCh38
rs750140796 Chr.3: 49276768-49276768 on Build GRCh38
rs747805158 Chr.3: 49276526-49276526 on Build GRCh38
rs771001807 Chr.3: 49276503-49276503 on Build GRCh38
rs758774110 Chr.3: 49276660-49276660 on Build GRCh38
rs751824683 Chr.3: 49276760-49276760 on Build GRCh38
rs200175271 Chr.3: 49276562-49276562 on Build GRCh38
rs780490292 Chr.3: 49276674-49276674 on Build GRCh38
rs763894323 Chr.3: 49276734-49276734 on Build GRCh38
rs779447208 Chr.3: 49276376-49276376 on Build GRCh38
rs754804103 Chr.3: 49276616-49276616 on Build GRCh38
rs1001466873 Chr.3: 49276652-49276652 on Build GRCh38
rs760534786 Chr.3: 49276636-49276637 on Build GRCh38
rs199624732 Chr.3: 49276783-49276783 on Build GRCh38
rs755862872 Chr.3: 49276641-49276641 on Build GRCh38
rs932467535 Chr.3: 49276623-49276623 on Build GRCh38
rs772657300 Chr.3: 49276431-49276431 on Build GRCh38
rs967903493 Chr.3: 49276736-49276736 on Build GRCh38
rs557579251 Chr.3: 49276684-49276684 on Build GRCh38
rs755543376 Chr.3: 49276763-49276763 on Build GRCh38
rs368106088 Chr.3: 49276426-49276426 on Build GRCh38
rs762039436 Chr.3: 49276583-49276583 on Build GRCh38
rs371684697 Chr.3: 49276599-49276599 on Build GRCh38
rs866948396 Chr.3: 49276715-49276715 on Build GRCh38
rs878948441 Chr.3: 49276722-49276722 on Build GRCh38
rs766278125 Chr.3: 49276609-49276609 on Build GRCh38
rs760235834 Chr.3: 49276730-49276730 on Build GRCh38
rs568755661 Chr.3: 49276410-49276410 on Build GRCh38
rs751502908 Chr.3: 49276615-49276615 on Build GRCh38
rs971343840 Chr.3: 49276325-49276325 on Build GRCh38
rs768224173 Chr.3: 49276388-49276388 on Build GRCh38
rs61749309 Chr.3: 49276677-49276677 on Build GRCh38
rs746194922 Chr.3: 49276546-49276546 on Build GRCh38
rs761665555 Chr.3: 49276453-49276453 on Build GRCh38
rs781641954 Chr.3: 49276779-49276779 on Build GRCh38
rs371985674 Chr.3: 49276538-49276538 on Build GRCh38
rs200997728 Chr.3: 49276781-49276781 on Build GRCh38
rs1045428487 Chr.3: 49276643-49276643 on Build GRCh38
rs17855113 Chr.3: 49276663-49276663 on Build GRCh38
rs765023440 Chr.3: 49276454-49276454 on Build GRCh38
rs776628054 Chr.3: 49276571-49276571 on Build GRCh38
rs747113998 Chr.3: 49276556-49276556 on Build GRCh38
rs149243117 Chr.3: 49276449-49276449 on Build GRCh38
rs759538759 Chr.3: 49276707-49276707 on Build GRCh38
rs758358753 Chr.3: 49276456-49276456 on Build GRCh38
rs772491183 Chr.3: 49276549-49276549 on Build GRCh38
rs780768040 Chr.3: 49276488-49276488 on Build GRCh38
rs750164043 Chr.3: 49276455-49276455 on Build GRCh38
rs766556144 Chr.3: 49276763-49276766 on Build GRCh38
rs774667709 Chr.3: 49276649-49276649 on Build GRCh38
rs921362347 Chr.3: 49276621-49276621 on Build GRCh38
rs550360442 Chr.3: 49276328-49276328 on Build GRCh38
rs13077498 Chr.3: 49276545-49276545 on Build GRCh38

View Full Product Details