Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49793631-49794121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR4
Entrez Gene ID
389118
Gene Name
cadherin related family member 4
Gene Aliases
CDH29, PRO34300
UniGene
Hs.363312
Gene Chromosome Location
Chr.3: 49790732-49802741 on Build GRCh38
Transcripts
NM_001007540.3 XM_011533700.2 XM_011533701.2 XM_017006366.1 XM_017006367.1 XM_017006368.1 XM_017006369.1 XM_017006370.1 XM_017006371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5494741 c.1482C>T p.S494S Chr.3: 49793804-49793804 on Build GRCh38
4568993 c.1435T>C p.Y479H Chr.3: 49793851-49793851 on Build GRCh38
6308107 c.1354G>A p.V452I Chr.3: 49793932-49793932 on Build GRCh38
1046274 c.1352G>T p.R451L Chr.3: 49793934-49793934 on Build GRCh38
1046273 c.1489G>C p.V497L Chr.3: 49793717-49793717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994656632 Chr.3: 49793975-49793975 on Build GRCh38
rs902326516 Chr.3: 49793670-49793670 on Build GRCh38
rs999343352 Chr.3: 49793676-49793676 on Build GRCh38
rs771129667 Chr.3: 49793996-49793996 on Build GRCh38
rs570755531 Chr.3: 49793740-49793740 on Build GRCh38
rs753496562 Chr.3: 49793864-49793864 on Build GRCh38
rs949291183 Chr.3: 49793653-49793653 on Build GRCh38
rs960686865 Chr.3: 49794094-49794094 on Build GRCh38
rs890101777 Chr.3: 49793956-49793956 on Build GRCh38
rs972934373 Chr.3: 49793820-49793820 on Build GRCh38
rs558652019 Chr.3: 49794062-49794062 on Build GRCh38
rs753808280 Chr.3: 49793919-49793919 on Build GRCh38
rs554682863 Chr.3: 49793916-49793916 on Build GRCh38
rs560516344 Chr.3: 49793830-49793830 on Build GRCh38
rs113873087 Chr.3: 49793969-49793969 on Build GRCh38
rs768204693 Chr.3: 49793708-49793708 on Build GRCh38
rs987049005 Chr.3: 49793730-49793730 on Build GRCh38
rs759585550 Chr.3: 49793732-49793732 on Build GRCh38
rs1028624783 Chr.3: 49793711-49793711 on Build GRCh38
rs1053583848 Chr.3: 49793920-49793920 on Build GRCh38
rs865836820 Chr.3: 49793856-49793856 on Build GRCh38
rs927638037 Chr.3: 49793821-49793821 on Build GRCh38
rs960446758 Chr.3: 49793846-49793846 on Build GRCh38
rs570035138 Chr.3: 49793935-49793935 on Build GRCh38
rs748437703 Chr.3: 49793883-49793883 on Build GRCh38
rs946914852 Chr.3: 49793865-49793865 on Build GRCh38
rs1040230491 Chr.3: 49793939-49793939 on Build GRCh38
rs756701544 Chr.3: 49793890-49793890 on Build GRCh38
rs755299684 Chr.3: 49793771-49793771 on Build GRCh38
rs144675334 Chr.3: 49793803-49793803 on Build GRCh38
rs924219022 Chr.3: 49793893-49793893 on Build GRCh38
rs990984843 Chr.3: 49793860-49793860 on Build GRCh38
rs755886343 Chr.3: 49793811-49793811 on Build GRCh38
rs61730923 Chr.3: 49793804-49793804 on Build GRCh38
rs949427603 Chr.3: 49793910-49793911 on Build GRCh38
rs774612363 Chr.3: 49794042-49794042 on Build GRCh38
rs747514199 Chr.3: 49793905-49793905 on Build GRCh38
rs891125978 Chr.3: 49793928-49793928 on Build GRCh38
rs916560543 Chr.3: 49793873-49793873 on Build GRCh38
rs1034656512 Chr.3: 49794074-49794074 on Build GRCh38
rs766578383 Chr.3: 49793694-49793694 on Build GRCh38
rs1027114123 Chr.3: 49794075-49794075 on Build GRCh38
rs990889443 Chr.3: 49794100-49794100 on Build GRCh38
rs1032034563 Chr.3: 49793690-49793690 on Build GRCh38
rs537481775 Chr.3: 49793936-49793936 on Build GRCh38
rs751528055 Chr.3: 49793699-49793699 on Build GRCh38
rs767187095 Chr.3: 49793736-49793736 on Build GRCh38
rs1046316339 Chr.3: 49793654-49793654 on Build GRCh38
rs1027456640 Chr.3: 49793977-49793977 on Build GRCh38
rs953010776 Chr.3: 49794092-49794093 on Build GRCh38
rs369948128 Chr.3: 49793812-49793812 on Build GRCh38
rs535266119 Chr.3: 49793915-49793915 on Build GRCh38
rs551873421 Chr.3: 49793728-49793728 on Build GRCh38
rs778456492 Chr.3: 49793902-49793902 on Build GRCh38
rs763056580 Chr.3: 49793693-49793693 on Build GRCh38
rs886332690 Chr.3: 49793988-49793988 on Build GRCh38
rs961654355 Chr.3: 49794015-49794015 on Build GRCh38
rs749368434 Chr.3: 49793993-49793993 on Build GRCh38
rs900313283 Chr.3: 49793944-49793944 on Build GRCh38

View Full Product Details