Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50236123-50236626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAI2
Entrez Gene ID
2771
Gene Name
G protein subunit alpha i2
Gene Aliases
GIP, GNAI2B, H_LUCA15.1, H_LUCA16.1
UniGene
Hs.77269
Gene Chromosome Location
Chr.3: 50226688-50259354 on Build GRCh38
Transcripts
NM_001282617.1 NM_001282618.1 NM_001282619.1 NM_001282620.1 NM_002070.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4638763 c.71G>T p.R24L Chr.3: 50236406-50236406 on Build GRCh38
6501144 c.12C>A p.T4T Chr.3: 50236347-50236347 on Build GRCh38
5802468 c.118+7delC p.? Chr.3: 50236460-50236460 on Build GRCh38
123391 c.117G>T p.L39F Chr.3: 50236452-50236452 on Build GRCh38
6415144 c.44G>A p.R15H Chr.3: 50236379-50236379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782389454 Chr.3: 50236467-50236467 on Build GRCh38
rs782141043 Chr.3: 50236347-50236347 on Build GRCh38
rs11557220 Chr.3: 50236325-50236325 on Build GRCh38
rs373887459 Chr.3: 50236468-50236468 on Build GRCh38
rs782125841 Chr.3: 50236334-50236334 on Build GRCh38
rs782454435 Chr.3: 50236451-50236451 on Build GRCh38
rs1013726883 Chr.3: 50236199-50236199 on Build GRCh38
rs897663722 Chr.3: 50236195-50236195 on Build GRCh38
rs587662487 Chr.3: 50236295-50236295 on Build GRCh38
rs587692842 Chr.3: 50236194-50236194 on Build GRCh38
rs781887853 Chr.3: 50236423-50236423 on Build GRCh38
rs587602953 Chr.3: 50236244-50236244 on Build GRCh38
rs782092831 Chr.3: 50236375-50236375 on Build GRCh38
rs782469247 Chr.3: 50236527-50236527 on Build GRCh38
rs587735553 Chr.3: 50236175-50236175 on Build GRCh38
rs782396913 Chr.3: 50236476-50236476 on Build GRCh38
rs77946516 Chr.3: 50236499-50236499 on Build GRCh38
rs782199863 Chr.3: 50236318-50236318 on Build GRCh38
rs976865637 Chr.3: 50236163-50236163 on Build GRCh38
rs781949451 Chr.3: 50236323-50236323 on Build GRCh38
rs782568968 Chr.3: 50236461-50236461 on Build GRCh38
rs782311212 Chr.3: 50236320-50236320 on Build GRCh38
rs782376225 Chr.3: 50236341-50236341 on Build GRCh38
rs1032999153 Chr.3: 50236349-50236349 on Build GRCh38
rs34944516 Chr.3: 50236313-50236313 on Build GRCh38
rs782028165 Chr.3: 50236345-50236345 on Build GRCh38
rs587757252 Chr.3: 50236143-50236143 on Build GRCh38
rs368969647 Chr.3: 50236494-50236494 on Build GRCh38
rs782410056 Chr.3: 50236337-50236337 on Build GRCh38
rs587594773 Chr.3: 50236183-50236183 on Build GRCh38
rs782353690 Chr.3: 50236490-50236490 on Build GRCh38
rs371157906 Chr.3: 50236504-50236504 on Build GRCh38
rs782736244 Chr.3: 50236382-50236382 on Build GRCh38
rs921230195 Chr.3: 50236169-50236169 on Build GRCh38
rs782039546 Chr.3: 50236489-50236489 on Build GRCh38
rs1036128609 Chr.3: 50236184-50236184 on Build GRCh38
rs782501325 Chr.3: 50236401-50236401 on Build GRCh38
rs781878883 Chr.3: 50236388-50236388 on Build GRCh38
rs782286331 Chr.3: 50236471-50236471 on Build GRCh38
rs587678505 Chr.3: 50236172-50236172 on Build GRCh38
rs377547544 Chr.3: 50236481-50236481 on Build GRCh38
rs932703371 Chr.3: 50236170-50236170 on Build GRCh38
rs782214187 Chr.3: 50236463-50236463 on Build GRCh38
rs782720927 Chr.3: 50236519-50236519 on Build GRCh38
rs587618141 Chr.3: 50236410-50236410 on Build GRCh38

View Full Product Details