Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50256643-50257152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAI2
Entrez Gene ID
2771
Gene Name
G protein subunit alpha i2
Gene Aliases
GIP, GNAI2B, H_LUCA15.1, H_LUCA16.1
UniGene
Hs.77269
Gene Chromosome Location
Chr.3: 50226688-50259354 on Build GRCh38
Transcripts
NM_001166425.1 NM_001282617.1 NM_001282618.1 NM_001282619.1 NM_001282620.1 NM_002070.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2851114 c.602A>G p.D201G Chr.3: 50256731-50256731 on Build GRCh38
6672142 c.715G>A p.E239K Chr.3: 50256844-50256844 on Build GRCh38
7159956 c.661G>C p.E221Q Chr.3: 50256838-50256838 on Build GRCh38
6672140 c.754G>A p.D252N Chr.3: 50256967-50256967 on Build GRCh38
4614923 c.681_682insT p.L228fs*5 Chr.3: 50256810-50256811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12721535 Chr.3: 50256940-50256940 on Build GRCh38
rs782365778 Chr.3: 50257074-50257074 on Build GRCh38
rs782220114 Chr.3: 50256709-50256709 on Build GRCh38
rs782512354 Chr.3: 50257002-50257002 on Build GRCh38
rs782024192 Chr.3: 50256927-50256927 on Build GRCh38
rs760792214 Chr.3: 50256844-50256844 on Build GRCh38
rs782029661 Chr.3: 50256901-50256901 on Build GRCh38
rs782779814 Chr.3: 50257129-50257129 on Build GRCh38
rs376430681 Chr.3: 50256805-50256805 on Build GRCh38
rs782598191 Chr.3: 50257077-50257077 on Build GRCh38
rs201958549 Chr.3: 50256789-50256789 on Build GRCh38
rs782787925 Chr.3: 50256875-50256875 on Build GRCh38
rs370577071 Chr.3: 50256912-50256912 on Build GRCh38
rs190048880 Chr.3: 50256869-50256869 on Build GRCh38
rs199869578 Chr.3: 50256898-50256898 on Build GRCh38
rs199594927 Chr.3: 50256907-50256907 on Build GRCh38
rs373500446 Chr.3: 50256753-50256753 on Build GRCh38
rs267599879 Chr.3: 50256939-50256939 on Build GRCh38
rs782593767 Chr.3: 50256889-50256889 on Build GRCh38
rs781818717 Chr.3: 50256966-50256966 on Build GRCh38
rs369101180 Chr.3: 50256750-50256750 on Build GRCh38
rs782811495 Chr.3: 50256929-50256929 on Build GRCh38
rs782522359 Chr.3: 50256712-50256712 on Build GRCh38
rs374608239 Chr.3: 50257118-50257118 on Build GRCh38
rs935779320 Chr.3: 50256825-50256825 on Build GRCh38
rs587657823 Chr.3: 50256816-50256816 on Build GRCh38
rs782256906 Chr.3: 50256893-50256893 on Build GRCh38
rs782669254 Chr.3: 50256677-50256677 on Build GRCh38
rs782631757 Chr.3: 50256784-50256784 on Build GRCh38
rs782454651 Chr.3: 50256957-50256957 on Build GRCh38
rs782061456 Chr.3: 50256828-50256828 on Build GRCh38
rs781895740 Chr.3: 50257044-50257044 on Build GRCh38
rs9830408 Chr.3: 50257119-50257119 on Build GRCh38
rs781783041 Chr.3: 50257099-50257099 on Build GRCh38
rs11557219 Chr.3: 50256849-50256849 on Build GRCh38
rs782051114 Chr.3: 50256888-50256888 on Build GRCh38
rs782745103 Chr.3: 50256963-50256963 on Build GRCh38
rs782277088 Chr.3: 50256905-50256905 on Build GRCh38
rs781849948 Chr.3: 50256667-50256667 on Build GRCh38
rs77643206 Chr.3: 50257071-50257071 on Build GRCh38
rs946282253 Chr.3: 50256873-50256873 on Build GRCh38
rs782641264 Chr.3: 50257014-50257014 on Build GRCh38
rs782266413 Chr.3: 50257082-50257082 on Build GRCh38
rs142945610 Chr.3: 50257063-50257063 on Build GRCh38
rs782530894 Chr.3: 50257061-50257061 on Build GRCh38
rs141703681 Chr.3: 50256804-50256804 on Build GRCh38
rs137853228 Chr.3: 50256729-50256729 on Build GRCh38
rs782016862 Chr.3: 50257011-50257011 on Build GRCh38
rs782636951 Chr.3: 50256717-50256717 on Build GRCh38
rs376218741 Chr.3: 50256697-50256697 on Build GRCh38
rs1024140028 Chr.3: 50257097-50257097 on Build GRCh38
rs141848616 Chr.3: 50256783-50256783 on Build GRCh38
rs782281236 Chr.3: 50256744-50256744 on Build GRCh38
rs139197363 Chr.3: 50257062-50257062 on Build GRCh38
rs587670723 Chr.3: 50256689-50256689 on Build GRCh38
rs782678061 Chr.3: 50256930-50256930 on Build GRCh38
rs782081212 Chr.3: 50257124-50257124 on Build GRCh38
rs782319901 Chr.3: 50257112-50257112 on Build GRCh38
rs927022731 Chr.3: 50256822-50256822 on Build GRCh38
rs200404060 Chr.3: 50256879-50256879 on Build GRCh38
rs143353731 Chr.3: 50256801-50256801 on Build GRCh38
rs782752710 Chr.3: 50256916-50256916 on Build GRCh38
rs782767890 Chr.3: 50257093-50257093 on Build GRCh38
rs782455764 Chr.3: 50256693-50256693 on Build GRCh38
rs781925628 Chr.3: 50256911-50256911 on Build GRCh38
rs782493369 Chr.3: 50256885-50256885 on Build GRCh38
rs587770948 Chr.3: 50256675-50256675 on Build GRCh38
rs138946270 Chr.3: 50256843-50256843 on Build GRCh38
rs781916903 Chr.3: 50256768-50256768 on Build GRCh38
rs782375392 Chr.3: 50257102-50257102 on Build GRCh38
rs781894924 Chr.3: 50256934-50256934 on Build GRCh38

View Full Product Details