Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50560825-50561323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf18
Entrez Gene ID
51161
Gene Name
chromosome 3 open reading frame 18
Gene Aliases
G20
UniGene
Hs.517860
Gene Chromosome Location
Chr.3: 50558025-50574835 on Build GRCh38
Transcripts
NM_001171740.2 NM_001171741.2 NM_001171743.2 NM_016210.4 XM_011533781.2 XM_011533782.2 XM_011533783.2 XM_011533784.2 XM_011533785.2 XM_011533788.1 XM_011533789.1 XM_011533790.1 XM_017006545.1 XM_017006546.1 XM_017006547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2851463 c.364G>A p.A122T Chr.3: 50560961-50560961 on Build GRCh38
7316936 c.337G>C p.E113Q Chr.3: 50560988-50560988 on Build GRCh38
2851464 c.356G>A p.R119Q Chr.3: 50560969-50560969 on Build GRCh38
2851465 c.312G>A p.T104T Chr.3: 50561013-50561013 on Build GRCh38
2851466 c.270G>T p.K90N Chr.3: 50561055-50561055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775162128 Chr.3: 50561067-50561067 on Build GRCh38
rs762701877 Chr.3: 50560883-50560883 on Build GRCh38
rs985592215 Chr.3: 50560899-50560899 on Build GRCh38
rs765902300 Chr.3: 50560884-50560884 on Build GRCh38
rs531328353 Chr.3: 50560952-50560952 on Build GRCh38
rs957091303 Chr.3: 50561219-50561219 on Build GRCh38
rs751745081 Chr.3: 50561014-50561014 on Build GRCh38
rs142961797 Chr.3: 50560961-50560961 on Build GRCh38
rs769683565 Chr.3: 50560947-50560947 on Build GRCh38
rs748084723 Chr.3: 50560943-50560943 on Build GRCh38
rs964535490 Chr.3: 50561129-50561130 on Build GRCh38
rs755129561 Chr.3: 50561016-50561016 on Build GRCh38
rs750841751 Chr.3: 50561006-50561006 on Build GRCh38
rs200548081 Chr.3: 50560970-50560970 on Build GRCh38
rs754381459 Chr.3: 50560893-50560893 on Build GRCh38
rs144985331 Chr.3: 50561021-50561021 on Build GRCh38
rs377301016 Chr.3: 50560962-50560962 on Build GRCh38
rs762441501 Chr.3: 50560974-50560974 on Build GRCh38
rs111256172 Chr.3: 50561026-50561026 on Build GRCh38
rs761655435 Chr.3: 50560877-50560877 on Build GRCh38
rs191805363 Chr.3: 50560855-50560855 on Build GRCh38
rs554064125 Chr.3: 50561298-50561298 on Build GRCh38
rs751915980 Chr.3: 50561114-50561114 on Build GRCh38
rs768709812 Chr.3: 50560939-50560939 on Build GRCh38
rs567390576 Chr.3: 50561010-50561010 on Build GRCh38
rs749170047 Chr.3: 50561033-50561033 on Build GRCh38
rs752907675 Chr.3: 50561083-50561083 on Build GRCh38
rs764762906 Chr.3: 50560890-50560890 on Build GRCh38
rs912895566 Chr.3: 50561267-50561267 on Build GRCh38
rs200958718 Chr.3: 50561050-50561050 on Build GRCh38
rs935623911 Chr.3: 50560932-50560932 on Build GRCh38
rs765840353 Chr.3: 50561075-50561075 on Build GRCh38
rs113385709 Chr.3: 50560880-50560880 on Build GRCh38
rs891490870 Chr.3: 50560953-50560953 on Build GRCh38
rs373201887 Chr.3: 50560926-50560926 on Build GRCh38
rs755219532 Chr.3: 50561116-50561116 on Build GRCh38
rs779423894 Chr.3: 50560906-50560906 on Build GRCh38
rs771810241 Chr.3: 50561065-50561065 on Build GRCh38
rs926502618 Chr.3: 50561152-50561152 on Build GRCh38
rs147519719 Chr.3: 50560957-50560957 on Build GRCh38
rs751126572 Chr.3: 50560889-50560889 on Build GRCh38
rs777536211 Chr.3: 50561031-50561031 on Build GRCh38
rs1054139480 Chr.3: 50561177-50561177 on Build GRCh38
rs145904003 Chr.3: 50561090-50561090 on Build GRCh38
rs757518600 Chr.3: 50561221-50561221 on Build GRCh38
rs979637560 Chr.3: 50561131-50561131 on Build GRCh38
rs201723362 Chr.3: 50561102-50561102 on Build GRCh38
rs374008666 Chr.3: 50561022-50561022 on Build GRCh38
rs967517901 Chr.3: 50560955-50560955 on Build GRCh38
rs988600014 Chr.3: 50561230-50561230 on Build GRCh38
rs370603003 Chr.3: 50560977-50560977 on Build GRCh38
rs903285912 Chr.3: 50561299-50561299 on Build GRCh38
rs747181419 Chr.3: 50560938-50560938 on Build GRCh38
rs140626565 Chr.3: 50560969-50560969 on Build GRCh38
rs773868596 Chr.3: 50560964-50560964 on Build GRCh38
rs143163338 Chr.3: 50561051-50561051 on Build GRCh38
rs937954332 Chr.3: 50561168-50561168 on Build GRCh38
rs765497721 Chr.3: 50561013-50561013 on Build GRCh38
rs1046337124 Chr.3: 50560851-50560851 on Build GRCh38
rs760685851 Chr.3: 50560894-50560895 on Build GRCh38
rs575603877 Chr.3: 50561277-50561277 on Build GRCh38
rs41291736 Chr.3: 50560965-50560965 on Build GRCh38
rs1012029101 Chr.3: 50561276-50561276 on Build GRCh38
rs764295409 Chr.3: 50561103-50561103 on Build GRCh38
rs760231718 Chr.3: 50561072-50561072 on Build GRCh38
rs770615706 Chr.3: 50561036-50561036 on Build GRCh38
rs766660810 Chr.3: 50561110-50561110 on Build GRCh38
rs757778840 Chr.3: 50560894-50560894 on Build GRCh38
rs139605691 Chr.3: 50560928-50560928 on Build GRCh38
rs776638049 Chr.3: 50560940-50560940 on Build GRCh38
rs544974610 Chr.3: 50561195-50561195 on Build GRCh38
rs772926953 Chr.3: 50560882-50560882 on Build GRCh38
rs763461660 Chr.3: 50561091-50561091 on Build GRCh38

View Full Product Details