Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50606768-50607276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISH
Entrez Gene ID
1154
Gene Name
cytokine inducible SH2 containing protein
Gene Aliases
BACTS2, CIS, CIS-1, G18, SOCS
UniGene
Hs.655334
Gene Chromosome Location
Chr.3: 50606454-50611831 on Build GRCh38
Transcripts
NM_013324.5 NM_145071.2 XM_011533329.2

SNP Detail

SNP ID Chromosome Location
rs904187896 Chr.3: 50607021-50607021 on Build GRCh38
rs911397871 Chr.3: 50606998-50606998 on Build GRCh38
rs536350244 Chr.3: 50606973-50606973 on Build GRCh38
rs186408718 Chr.3: 50607174-50607174 on Build GRCh38
rs540947825 Chr.3: 50607231-50607231 on Build GRCh38
rs993160823 Chr.3: 50607027-50607027 on Build GRCh38
rs575363609 Chr.3: 50606888-50606888 on Build GRCh38
rs1053665187 Chr.3: 50606840-50606840 on Build GRCh38
rs915147069 Chr.3: 50607160-50607160 on Build GRCh38
rs982236646 Chr.3: 50607080-50607080 on Build GRCh38
rs751623301 Chr.3: 50606882-50606882 on Build GRCh38
rs756308704 Chr.3: 50607099-50607099 on Build GRCh38
rs947400922 Chr.3: 50606851-50606851 on Build GRCh38
rs117227840 Chr.3: 50606912-50606912 on Build GRCh38
rs1000516669 Chr.3: 50606810-50606810 on Build GRCh38
rs969883546 Chr.3: 50607140-50607140 on Build GRCh38
rs528592775 Chr.3: 50607237-50607237 on Build GRCh38
rs889248194 Chr.3: 50606927-50606927 on Build GRCh38
rs778136736 Chr.3: 50607136-50607136 on Build GRCh38
rs34197892 Chr.3: 50606829-50606830 on Build GRCh38
rs1031620718 Chr.3: 50606821-50606821 on Build GRCh38
rs755096978 Chr.3: 50606955-50606955 on Build GRCh38
rs377268877 Chr.3: 50607127-50607127 on Build GRCh38
rs955616639 Chr.3: 50607244-50607244 on Build GRCh38
rs74957911 Chr.3: 50606790-50606790 on Build GRCh38
rs545809289 Chr.3: 50607163-50607163 on Build GRCh38
rs1038639948 Chr.3: 50606793-50606793 on Build GRCh38
rs924577819 Chr.3: 50607219-50607219 on Build GRCh38
rs918594983 Chr.3: 50607233-50607233 on Build GRCh38
rs1026376989 Chr.3: 50607047-50607047 on Build GRCh38
rs778781810 Chr.3: 50606844-50606844 on Build GRCh38
rs557855323 Chr.3: 50606860-50606860 on Build GRCh38
rs1053424662 Chr.3: 50607084-50607084 on Build GRCh38
rs943082824 Chr.3: 50607015-50607015 on Build GRCh38
rs115418080 Chr.3: 50607089-50607089 on Build GRCh38
rs566585100 Chr.3: 50606897-50606897 on Build GRCh38
rs937836391 Chr.3: 50606794-50606794 on Build GRCh38
rs904199229 Chr.3: 50606797-50606797 on Build GRCh38
rs959191880 Chr.3: 50607114-50607114 on Build GRCh38
rs1016464302 Chr.3: 50606977-50606977 on Build GRCh38
rs997985656 Chr.3: 50606890-50606890 on Build GRCh38
rs963021440 Chr.3: 50607181-50607181 on Build GRCh38
rs370153373 Chr.3: 50607116-50607116 on Build GRCh38
rs970104581 Chr.3: 50606870-50606870 on Build GRCh38
rs980192478 Chr.3: 50606879-50606879 on Build GRCh38
rs771458521 Chr.3: 50607245-50607245 on Build GRCh38
rs377563793 Chr.3: 50607035-50607035 on Build GRCh38
rs114038753 Chr.3: 50607076-50607076 on Build GRCh38
rs973388188 Chr.3: 50606911-50606911 on Build GRCh38
rs114953805 Chr.3: 50607018-50607018 on Build GRCh38
rs1042137711 Chr.3: 50606865-50606865 on Build GRCh38

View Full Product Details