Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50607950-50608457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISH
Entrez Gene ID
1154
Gene Name
cytokine inducible SH2 containing protein
Gene Aliases
BACTS2, CIS, CIS-1, G18, SOCS
UniGene
Hs.655334
Gene Chromosome Location
Chr.3: 50606454-50611831 on Build GRCh38
Transcripts
NM_013324.5 NM_145071.2 XM_011533329.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6350951 c.284G>A p.R95Q Chr.3: 50608381-50608381 on Build GRCh38
1248351 c.195G>T p.E65D Chr.3: 50608419-50608419 on Build GRCh38
149394 c.421G>A p.D141N Chr.3: 50607963-50607963 on Build GRCh38
4782282 c.472G>A p.D158N Chr.3: 50607963-50607963 on Build GRCh38
6629342 c.421C>T p.R141C Chr.3: 50608014-50608014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141046429 Chr.3: 50608382-50608382 on Build GRCh38
rs991020460 Chr.3: 50608406-50608406 on Build GRCh38
rs766498255 Chr.3: 50608113-50608113 on Build GRCh38
rs2239752 Chr.3: 50607982-50607982 on Build GRCh38
rs377739524 Chr.3: 50608348-50608348 on Build GRCh38
rs377401159 Chr.3: 50608064-50608064 on Build GRCh38
rs772515449 Chr.3: 50607988-50607988 on Build GRCh38
rs781089685 Chr.3: 50608034-50608034 on Build GRCh38
rs776935240 Chr.3: 50608012-50608012 on Build GRCh38
rs569531642 Chr.3: 50608279-50608279 on Build GRCh38
rs113456317 Chr.3: 50608217-50608217 on Build GRCh38
rs748585454 Chr.3: 50608169-50608169 on Build GRCh38
rs200906901 Chr.3: 50608150-50608150 on Build GRCh38
rs538839740 Chr.3: 50608434-50608434 on Build GRCh38
rs200588661 Chr.3: 50608106-50608106 on Build GRCh38
rs763489198 Chr.3: 50607983-50607993 on Build GRCh38
rs762164564 Chr.3: 50608095-50608095 on Build GRCh38
rs373552550 Chr.3: 50608107-50608107 on Build GRCh38
rs143786338 Chr.3: 50608258-50608258 on Build GRCh38
rs373707188 Chr.3: 50608360-50608360 on Build GRCh38
rs902196260 Chr.3: 50608247-50608247 on Build GRCh38
rs201054842 Chr.3: 50608000-50608000 on Build GRCh38
rs929428092 Chr.3: 50608136-50608136 on Build GRCh38
rs755578001 Chr.3: 50608163-50608163 on Build GRCh38
rs143405439 Chr.3: 50607981-50607981 on Build GRCh38
rs547039308 Chr.3: 50608326-50608326 on Build GRCh38
rs34581675 Chr.3: 50608146-50608146 on Build GRCh38
rs995226227 Chr.3: 50608358-50608358 on Build GRCh38
rs373005313 Chr.3: 50608340-50608340 on Build GRCh38
rs748801053 Chr.3: 50607987-50607987 on Build GRCh38
rs775575395 Chr.3: 50608356-50608356 on Build GRCh38
rs140032833 Chr.3: 50608014-50608014 on Build GRCh38
rs767350636 Chr.3: 50608121-50608121 on Build GRCh38
rs144012341 Chr.3: 50608076-50608076 on Build GRCh38
rs929825606 Chr.3: 50607984-50607984 on Build GRCh38
rs368792354 Chr.3: 50608130-50608130 on Build GRCh38
rs373879739 Chr.3: 50608233-50608233 on Build GRCh38
rs751543516 Chr.3: 50608027-50608027 on Build GRCh38
rs369545796 Chr.3: 50608019-50608019 on Build GRCh38
rs762076085 Chr.3: 50608013-50608013 on Build GRCh38
rs771241466 Chr.3: 50608422-50608422 on Build GRCh38
rs763916957 Chr.3: 50608135-50608135 on Build GRCh38
rs747577229 Chr.3: 50608067-50608067 on Build GRCh38
rs916873255 Chr.3: 50608198-50608198 on Build GRCh38
rs777306102 Chr.3: 50608212-50608212 on Build GRCh38
rs747825441 Chr.3: 50607971-50607971 on Build GRCh38
rs140940134 Chr.3: 50608120-50608120 on Build GRCh38
rs748893210 Chr.3: 50608055-50608055 on Build GRCh38
rs199740084 Chr.3: 50608154-50608154 on Build GRCh38
rs372502239 Chr.3: 50607994-50607994 on Build GRCh38
rs750557678 Chr.3: 50608017-50608017 on Build GRCh38
rs376687415 Chr.3: 50608077-50608077 on Build GRCh38
rs756587372 Chr.3: 50608389-50608389 on Build GRCh38
rs544025525 Chr.3: 50608281-50608281 on Build GRCh38
rs145632110 Chr.3: 50608065-50608065 on Build GRCh38
rs866100406 Chr.3: 50607983-50607983 on Build GRCh38
rs755895848 Chr.3: 50608051-50608051 on Build GRCh38
rs369807277 Chr.3: 50608354-50608354 on Build GRCh38
rs770083611 Chr.3: 50608103-50608103 on Build GRCh38
rs376708125 Chr.3: 50608361-50608361 on Build GRCh38
rs1006719247 Chr.3: 50608324-50608324 on Build GRCh38
rs774271488 Chr.3: 50608114-50608114 on Build GRCh38
rs377391900 Chr.3: 50608173-50608173 on Build GRCh38
rs148162012 Chr.3: 50608381-50608381 on Build GRCh38
rs753576043 Chr.3: 50608143-50608143 on Build GRCh38
rs138469436 Chr.3: 50607979-50607979 on Build GRCh38
rs749611185 Chr.3: 50608417-50608417 on Build GRCh38
rs983958208 Chr.3: 50608151-50608151 on Build GRCh38
rs1055344080 Chr.3: 50608182-50608182 on Build GRCh38
rs1043422834 Chr.3: 50608238-50608238 on Build GRCh38
rs778149920 Chr.3: 50608412-50608412 on Build GRCh38
rs752507448 Chr.3: 50608040-50608040 on Build GRCh38
rs139578499 Chr.3: 50608018-50608018 on Build GRCh38
rs201825038 Chr.3: 50607995-50607995 on Build GRCh38
rs761020637 Chr.3: 50607996-50607996 on Build GRCh38
rs746566483 Chr.3: 50608357-50608357 on Build GRCh38
rs200844383 Chr.3: 50608166-50608166 on Build GRCh38
rs199906301 Chr.3: 50608158-50608158 on Build GRCh38
rs755987753 Chr.3: 50608128-50608128 on Build GRCh38
rs771028293 Chr.3: 50608330-50608330 on Build GRCh38
rs621975 Chr.3: 50608204-50608204 on Build GRCh38
rs754822590 Chr.3: 50608032-50608032 on Build GRCh38
rs1051296851 Chr.3: 50608280-50608280 on Build GRCh38
rs622502 Chr.3: 50608362-50608362 on Build GRCh38
rs761635256 Chr.3: 50608411-50608411 on Build GRCh38
rs200402505 Chr.3: 50608054-50608054 on Build GRCh38
rs770479353 Chr.3: 50608060-50608060 on Build GRCh38
rs757848688 Chr.3: 50608370-50608370 on Build GRCh38
rs752595479 Chr.3: 50608126-50608126 on Build GRCh38

View Full Product Details