Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50841536-50842036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK3
Entrez Gene ID
1795
Gene Name
dedicator of cytokinesis 3
Gene Aliases
MOCA, PBP
UniGene
Hs.476284
Gene Chromosome Location
Chr.3: 50674969-51384198 on Build GRCh38
Transcripts
NM_004947.4 XM_005264914.3 XM_005264915.3 XM_005264916.3 XM_005264917.3 XM_005264918.3 XM_006713008.3 XM_006713009.3 XM_006713010.3 XM_017005825.1 XM_017005826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3595616 c.140C>T p.S47L Chr.3: 50841693-50841693 on Build GRCh38
1131399 c.126G>A p.W42* Chr.3: 50841679-50841679 on Build GRCh38
3696172 c.162+2T>C p.? Chr.3: 50841717-50841717 on Build GRCh38
4525587 c.133G>A p.G45R Chr.3: 50841686-50841686 on Build GRCh38
1424212 c.147G>T p.K49N Chr.3: 50841700-50841700 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922267850 Chr.3: 50841894-50841894 on Build GRCh38
rs369087161 Chr.3: 50841648-50841648 on Build GRCh38
rs770184572 Chr.3: 50841743-50841743 on Build GRCh38
rs757190134 Chr.3: 50841745-50841763 on Build GRCh38
rs758012028 Chr.3: 50841736-50841736 on Build GRCh38
rs773424903 Chr.3: 50841637-50841637 on Build GRCh38
rs1047734479 Chr.3: 50841649-50841649 on Build GRCh38
rs773340220 Chr.3: 50841738-50841739 on Build GRCh38
rs955024392 Chr.3: 50841902-50841902 on Build GRCh38
rs561621832 Chr.3: 50841628-50841628 on Build GRCh38
rs997204368 Chr.3: 50841915-50841915 on Build GRCh38
rs200327865 Chr.3: 50841728-50841728 on Build GRCh38
rs763290893 Chr.3: 50841638-50841638 on Build GRCh38
rs369797473 Chr.3: 50841733-50841733 on Build GRCh38
rs760468700 Chr.3: 50841743-50841763 on Build GRCh38
rs558397679 Chr.3: 50841743-50841744 on Build GRCh38
rs911099128 Chr.3: 50841586-50841586 on Build GRCh38
rs547319186 Chr.3: 50841748-50841748 on Build GRCh38
rs753573046 Chr.3: 50841703-50841703 on Build GRCh38
rs889046137 Chr.3: 50841965-50841965 on Build GRCh38
rs779782929 Chr.3: 50841761-50841762 on Build GRCh38
rs1011057295 Chr.3: 50841794-50841794 on Build GRCh38
rs528256652 Chr.3: 50841592-50841592 on Build GRCh38
rs763407975 Chr.3: 50841768-50841768 on Build GRCh38
rs542301410 Chr.3: 50841955-50841955 on Build GRCh38
rs778935832 Chr.3: 50841744-50841745 on Build GRCh38
rs371324492 Chr.3: 50841653-50841653 on Build GRCh38
rs374134140 Chr.3: 50841655-50841655 on Build GRCh38
rs575687760 Chr.3: 50841861-50841861 on Build GRCh38
rs752272807 Chr.3: 50841774-50841774 on Build GRCh38
rs778442464 Chr.3: 50841729-50841729 on Build GRCh38
rs773718742 Chr.3: 50841766-50841766 on Build GRCh38
rs869137602 Chr.3: 50841604-50841605 on Build GRCh38
rs776506899 Chr.3: 50841966-50841970 on Build GRCh38
rs202139176 Chr.3: 50841652-50841652 on Build GRCh38
rs978716631 Chr.3: 50841850-50841850 on Build GRCh38
rs147462808 Chr.3: 50841779-50841779 on Build GRCh38
rs758184532 Chr.3: 50841750-50841759 on Build GRCh38
rs534875147 Chr.3: 50841746-50841746 on Build GRCh38
rs748623058 Chr.3: 50841625-50841625 on Build GRCh38
rs571061455 Chr.3: 50841745-50841745 on Build GRCh38
rs190967418 Chr.3: 50841596-50841596 on Build GRCh38
rs557436765 Chr.3: 50841824-50841824 on Build GRCh38
rs375949875 Chr.3: 50841730-50841730 on Build GRCh38
rs967001028 Chr.3: 50841828-50841828 on Build GRCh38
rs779675858 Chr.3: 50841737-50841737 on Build GRCh38
rs994465491 Chr.3: 50841636-50841636 on Build GRCh38
rs533098527 Chr.3: 50841696-50841696 on Build GRCh38
rs914374813 Chr.3: 50841562-50841562 on Build GRCh38
rs551169146 Chr.3: 50841700-50841700 on Build GRCh38
rs764010258 Chr.3: 50841685-50841685 on Build GRCh38
rs1038723749 Chr.3: 50841620-50841620 on Build GRCh38
rs557935450 Chr.3: 50841962-50841962 on Build GRCh38
rs1004671473 Chr.3: 50841787-50841787 on Build GRCh38
rs536143793 Chr.3: 50841813-50841813 on Build GRCh38
rs985243160 Chr.3: 50841905-50841905 on Build GRCh38
rs539956068 Chr.3: 50841939-50841939 on Build GRCh38
rs767339541 Chr.3: 50841770-50841770 on Build GRCh38
rs1048545074 Chr.3: 50841777-50841777 on Build GRCh38
rs756023255 Chr.3: 50841664-50841664 on Build GRCh38
rs757008409 Chr.3: 50841719-50841719 on Build GRCh38
rs748709292 Chr.3: 50841738-50841738 on Build GRCh38
rs550121017 Chr.3: 50841654-50841654 on Build GRCh38
rs574989365 Chr.3: 50841781-50841781 on Build GRCh38
rs771474544 Chr.3: 50841741-50841742 on Build GRCh38
rs767633443 Chr.3: 50841751-50841751 on Build GRCh38
rs910963411 Chr.3: 50841936-50841936 on Build GRCh38

View Full Product Details