Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51090122-51090639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK3
Entrez Gene ID
1795
Gene Name
dedicator of cytokinesis 3
Gene Aliases
MOCA, PBP
UniGene
Hs.476284
Gene Chromosome Location
Chr.3: 50674969-51384198 on Build GRCh38
Transcripts
NM_004947.4 XM_005264914.3 XM_005264915.3 XM_005264916.3 XM_005264917.3 XM_005264918.3 XM_006713008.3 XM_006713009.3 XM_006713010.3 XM_017005825.1 XM_017005826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4555732 c.666G>A p.K222K Chr.3: 51090304-51090304 on Build GRCh38
2851578 c.713C>T p.S238F Chr.3: 51090351-51090351 on Build GRCh38
86437 c.604C>T p.R202C Chr.3: 51090242-51090242 on Build GRCh38
6270699 c.659G>T p.S220I Chr.3: 51090297-51090297 on Build GRCh38
7336578 c.691G>A p.E231K Chr.3: 51090329-51090329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537023419 Chr.3: 51090382-51090382 on Build GRCh38
rs757569605 Chr.3: 51090366-51090366 on Build GRCh38
rs979665227 Chr.3: 51090209-51090209 on Build GRCh38
rs757919847 Chr.3: 51090237-51090237 on Build GRCh38
rs749333957 Chr.3: 51090180-51090180 on Build GRCh38
rs570932521 Chr.3: 51090555-51090555 on Build GRCh38
rs986946312 Chr.3: 51090238-51090238 on Build GRCh38
rs1041262921 Chr.3: 51090372-51090372 on Build GRCh38
rs776051894 Chr.3: 51090194-51090197 on Build GRCh38
rs975566226 Chr.3: 51090608-51090608 on Build GRCh38
rs867973581 Chr.3: 51090267-51090267 on Build GRCh38
rs201867246 Chr.3: 51090385-51090385 on Build GRCh38
rs1003211897 Chr.3: 51090318-51090318 on Build GRCh38
rs200510014 Chr.3: 51090198-51090198 on Build GRCh38
rs760342204 Chr.3: 51090211-51090211 on Build GRCh38
rs760428493 Chr.3: 51090172-51090172 on Build GRCh38
rs868458081 Chr.3: 51090324-51090324 on Build GRCh38
rs779029507 Chr.3: 51090361-51090361 on Build GRCh38
rs893730412 Chr.3: 51090456-51090456 on Build GRCh38
rs866059893 Chr.3: 51090242-51090242 on Build GRCh38
rs749953998 Chr.3: 51090236-51090236 on Build GRCh38
rs753192341 Chr.3: 51090346-51090346 on Build GRCh38
rs759673137 Chr.3: 51090554-51090554 on Build GRCh38
rs377383748 Chr.3: 51090324-51090324 on Build GRCh38
rs942444952 Chr.3: 51090248-51090248 on Build GRCh38
rs775333465 Chr.3: 51090299-51090299 on Build GRCh38
rs776648246 Chr.3: 51090424-51090424 on Build GRCh38
rs759635635 Chr.3: 51090285-51090285 on Build GRCh38
rs149337164 Chr.3: 51090314-51090314 on Build GRCh38
rs768460523 Chr.3: 51090280-51090280 on Build GRCh38
rs1019777151 Chr.3: 51090571-51090571 on Build GRCh38
rs201510547 Chr.3: 51090183-51090183 on Build GRCh38
rs768724303 Chr.3: 51090290-51090290 on Build GRCh38
rs146177239 Chr.3: 51090417-51090417 on Build GRCh38
rs960790625 Chr.3: 51090171-51090171 on Build GRCh38
rs532254685 Chr.3: 51090573-51090573 on Build GRCh38
rs199961397 Chr.3: 51090406-51090406 on Build GRCh38
rs753659222 Chr.3: 51090321-51090321 on Build GRCh38
rs374435105 Chr.3: 51090579-51090579 on Build GRCh38
rs764990216 Chr.3: 51090334-51090334 on Build GRCh38
rs563991202 Chr.3: 51090262-51090262 on Build GRCh38
rs758012008 Chr.3: 51090337-51090337 on Build GRCh38
rs530942766 Chr.3: 51090505-51090505 on Build GRCh38
rs987492706 Chr.3: 51090566-51090566 on Build GRCh38
rs1020660676 Chr.3: 51090543-51090543 on Build GRCh38
rs577044257 Chr.3: 51090271-51090271 on Build GRCh38
rs371384870 Chr.3: 51090362-51090362 on Build GRCh38
rs750047647 Chr.3: 51090335-51090335 on Build GRCh38
rs930600948 Chr.3: 51090325-51090325 on Build GRCh38
rs142425426 Chr.3: 51090543-51090543 on Build GRCh38
rs924186593 Chr.3: 51090221-51090221 on Build GRCh38
rs541499652 Chr.3: 51090162-51090162 on Build GRCh38
rs201614726 Chr.3: 51090338-51090338 on Build GRCh38
rs771148448 Chr.3: 51090268-51090268 on Build GRCh38
rs748648148 Chr.3: 51090255-51090255 on Build GRCh38
rs772095631 Chr.3: 51090400-51090400 on Build GRCh38
rs112195040 Chr.3: 51090496-51090496 on Build GRCh38
rs886991063 Chr.3: 51090274-51090274 on Build GRCh38
rs903950278 Chr.3: 51090468-51090468 on Build GRCh38
rs770094368 Chr.3: 51090260-51090260 on Build GRCh38
rs373461836 Chr.3: 51090289-51090289 on Build GRCh38
rs138640309 Chr.3: 51090187-51090187 on Build GRCh38
rs868705158 Chr.3: 51090367-51090367 on Build GRCh38
rs1003325159 Chr.3: 51090567-51090567 on Build GRCh38
rs559628209 Chr.3: 51090355-51090355 on Build GRCh38
rs763850124 Chr.3: 51090317-51090317 on Build GRCh38
rs574740148 Chr.3: 51090154-51090154 on Build GRCh38
rs1012374402 Chr.3: 51090179-51090179 on Build GRCh38
rs919654592 Chr.3: 51090618-51090618 on Build GRCh38
rs772477837 Chr.3: 51090189-51090193 on Build GRCh38
rs982687639 Chr.3: 51090232-51090232 on Build GRCh38
rs200070432 Chr.3: 51090249-51090249 on Build GRCh38
rs966522251 Chr.3: 51090211-51090211 on Build GRCh38
rs542575344 Chr.3: 51090323-51090323 on Build GRCh38
rs778629763 Chr.3: 51090217-51090217 on Build GRCh38
rs779141667 Chr.3: 51090368-51090368 on Build GRCh38
rs1023753081 Chr.3: 51090451-51090451 on Build GRCh38
rs749581232 Chr.3: 51090266-51090266 on Build GRCh38
rs34684757 Chr.3: 51090210-51090211 on Build GRCh38

View Full Product Details