Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52402687-52403176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAP1
Entrez Gene ID
8314
Gene Name
BRCA1 associated protein 1
Gene Aliases
HUCEP-13, UCHL2, hucep-6
UniGene
Hs.106674
Gene Chromosome Location
Chr.3: 52401004-52410105 on Build GRCh38
Transcripts
NM_004656.3 XM_011534149.2 XM_011534150.2 XM_011534151.2 XM_011534152.1 XM_017007303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4533187 c.1954G>A p.E652K Chr.3: 52402808-52402808 on Build GRCh38
6848358 c.1925_1930delAGATTG p.E642_I643delEI Chr.3: 52402832-52402837 on Build GRCh38
6023803 c.1869_1872delTGGG p.S623fs*13 Chr.3: 52403156-52403159 on Build GRCh38
2853451 c.1939G>T p.E647* Chr.3: 52402823-52402823 on Build GRCh38
6848308 c.1979_1980delTC p.F660fs*1 Chr.3: 52402782-52402783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937072943 Chr.3: 52403027-52403027 on Build GRCh38
rs768169476 Chr.3: 52402906-52402906 on Build GRCh38
rs371637639 Chr.3: 52402910-52402910 on Build GRCh38
rs34526543 Chr.3: 52402967-52402968 on Build GRCh38
rs375950004 Chr.3: 52403156-52403156 on Build GRCh38
rs981612334 Chr.3: 52402957-52402957 on Build GRCh38
rs749364986 Chr.3: 52403005-52403005 on Build GRCh38
rs755112307 Chr.3: 52402878-52402878 on Build GRCh38
rs148624125 Chr.3: 52402800-52402800 on Build GRCh38
rs757077441 Chr.3: 52402716-52402716 on Build GRCh38
rs752637155 Chr.3: 52402959-52402959 on Build GRCh38
rs891021950 Chr.3: 52403074-52403074 on Build GRCh38
rs751219874 Chr.3: 52402803-52402803 on Build GRCh38
rs774743129 Chr.3: 52402741-52402741 on Build GRCh38
rs755020488 Chr.3: 52402930-52402930 on Build GRCh38
rs369615354 Chr.3: 52402865-52402865 on Build GRCh38
rs751166890 Chr.3: 52402714-52402714 on Build GRCh38
rs772485431 Chr.3: 52402900-52402900 on Build GRCh38
rs771530795 Chr.3: 52403102-52403102 on Build GRCh38
rs770353351 Chr.3: 52402967-52402967 on Build GRCh38
rs756025904 Chr.3: 52402970-52402971 on Build GRCh38
rs200578667 Chr.3: 52402902-52402902 on Build GRCh38
rs756064795 Chr.3: 52402723-52402723 on Build GRCh38
rs771378046 Chr.3: 52402765-52402765 on Build GRCh38
rs767198116 Chr.3: 52402927-52402927 on Build GRCh38
rs772742460 Chr.3: 52403105-52403105 on Build GRCh38
rs537090290 Chr.3: 52403127-52403127 on Build GRCh38
rs781565390 Chr.3: 52402962-52402962 on Build GRCh38
rs753731660 Chr.3: 52402929-52402929 on Build GRCh38
rs546069189 Chr.3: 52402757-52402757 on Build GRCh38
rs967565781 Chr.3: 52402984-52402984 on Build GRCh38
rs869312493 Chr.3: 52402993-52402993 on Build GRCh38
rs747218187 Chr.3: 52402947-52402947 on Build GRCh38
rs531046526 Chr.3: 52402928-52402928 on Build GRCh38
rs776553452 Chr.3: 52403128-52403128 on Build GRCh38
rs964783693 Chr.3: 52402842-52402842 on Build GRCh38
rs780012037 Chr.3: 52403038-52403038 on Build GRCh38
rs957749067 Chr.3: 52403069-52403069 on Build GRCh38
rs571043097 Chr.3: 52403018-52403018 on Build GRCh38
rs768732582 Chr.3: 52402740-52402740 on Build GRCh38
rs919590367 Chr.3: 52403051-52403051 on Build GRCh38
rs1030620153 Chr.3: 52403054-52403054 on Build GRCh38
rs146661777 Chr.3: 52402901-52402901 on Build GRCh38
rs376519545 Chr.3: 52402834-52402834 on Build GRCh38
rs371236827 Chr.3: 52403114-52403114 on Build GRCh38
rs989620105 Chr.3: 52403118-52403118 on Build GRCh38
rs150494756 Chr.3: 52402818-52402818 on Build GRCh38
rs201537464 Chr.3: 52402887-52402887 on Build GRCh38
rs929656088 Chr.3: 52403059-52403059 on Build GRCh38
rs753767419 Chr.3: 52402839-52402839 on Build GRCh38
rs375522429 Chr.3: 52402891-52402891 on Build GRCh38
rs372756165 Chr.3: 52402903-52402903 on Build GRCh38
rs755915077 Chr.3: 52402819-52402819 on Build GRCh38
rs878957987 Chr.3: 52402907-52402907 on Build GRCh38
rs759370336 Chr.3: 52403132-52403132 on Build GRCh38
rs551399575 Chr.3: 52402831-52402831 on Build GRCh38
rs540081893 Chr.3: 52403050-52403050 on Build GRCh38
rs551121590 Chr.3: 52402934-52402934 on Build GRCh38
rs758440651 Chr.3: 52402945-52402945 on Build GRCh38
rs766939937 Chr.3: 52402713-52402713 on Build GRCh38
rs771206537 Chr.3: 52402960-52402960 on Build GRCh38
rs775945831 Chr.3: 52402986-52402986 on Build GRCh38
rs964152039 Chr.3: 52403147-52403147 on Build GRCh38
rs773767890 Chr.3: 52402761-52402761 on Build GRCh38
rs1047061800 Chr.3: 52402889-52402889 on Build GRCh38
rs989281268 Chr.3: 52402748-52402748 on Build GRCh38
rs151308667 Chr.3: 52402816-52402816 on Build GRCh38
rs762396224 Chr.3: 52402794-52402794 on Build GRCh38
rs761399207 Chr.3: 52402806-52402806 on Build GRCh38
rs372282516 Chr.3: 52402789-52402789 on Build GRCh38
rs747679673 Chr.3: 52403089-52403089 on Build GRCh38
rs780037860 Chr.3: 52402729-52402729 on Build GRCh38
rs532207217 Chr.3: 52402731-52402731 on Build GRCh38
rs368071174 Chr.3: 52402754-52402754 on Build GRCh38
rs777883767 Chr.3: 52402946-52402946 on Build GRCh38
rs761091762 Chr.3: 52402764-52402764 on Build GRCh38
rs199598142 Chr.3: 52402720-52402720 on Build GRCh38
rs1046775979 Chr.3: 52403063-52403063 on Build GRCh38
rs745833544 Chr.3: 52402722-52402722 on Build GRCh38
rs999073962 Chr.3: 52403043-52403043 on Build GRCh38
rs1041287605 Chr.3: 52402815-52402815 on Build GRCh38
rs746220455 Chr.3: 52402966-52402966 on Build GRCh38
rs371507917 Chr.3: 52402749-52402749 on Build GRCh38

View Full Product Details