Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52696450-52696948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT8D1
Entrez Gene ID
55830
Gene Name
glycosyltransferase 8 domain containing 1
Gene Aliases
AD-017, MSTP139
UniGene
Hs.297304
Gene Chromosome Location
Chr.3: 52694484-52706083 on Build GRCh38
Transcripts
NM_001010983.2 NM_001278280.1 NM_001278281.1 NM_018446.3 NM_152932.2 XM_017006857.1 XM_017006858.1 XM_017006859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4119187 c.375T>G p.I125M Chr.3: 52696614-52696614 on Build GRCh38
4629494 c.435A>T p.E145D Chr.3: 52696554-52696554 on Build GRCh38
1047122 c.331T>C p.S111P Chr.3: 52696658-52696658 on Build GRCh38
7080025 c.432_438delGGAATCC p.E145fs*1 Chr.3: 52696551-52696557 on Build GRCh38
3365263 c.386A>C p.D129A Chr.3: 52696603-52696603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377675634 Chr.3: 52696698-52696698 on Build GRCh38
rs570052153 Chr.3: 52696815-52696815 on Build GRCh38
rs1032269734 Chr.3: 52696712-52696712 on Build GRCh38
rs377216315 Chr.3: 52696672-52696672 on Build GRCh38
rs140309601 Chr.3: 52696543-52696543 on Build GRCh38
rs753444443 Chr.3: 52696739-52696739 on Build GRCh38
rs953977883 Chr.3: 52696791-52696791 on Build GRCh38
rs748713942 Chr.3: 52696572-52696572 on Build GRCh38
rs748517802 Chr.3: 52696691-52696691 on Build GRCh38
rs747477565 Chr.3: 52696706-52696706 on Build GRCh38
rs779410148 Chr.3: 52696539-52696540 on Build GRCh38
rs374851153 Chr.3: 52696512-52696512 on Build GRCh38
rs372757912 Chr.3: 52696652-52696652 on Build GRCh38
rs756731022 Chr.3: 52696794-52696794 on Build GRCh38
rs775150970 Chr.3: 52696505-52696505 on Build GRCh38
rs1001647583 Chr.3: 52696609-52696609 on Build GRCh38
rs751495137 Chr.3: 52696526-52696526 on Build GRCh38
rs1008140952 Chr.3: 52696769-52696769 on Build GRCh38
rs769248984 Chr.3: 52696630-52696630 on Build GRCh38
rs1005512624 Chr.3: 52696670-52696670 on Build GRCh38
rs756283112 Chr.3: 52696663-52696663 on Build GRCh38
rs917329440 Chr.3: 52696901-52696901 on Build GRCh38
rs924595961 Chr.3: 52696841-52696841 on Build GRCh38
rs910810184 Chr.3: 52696480-52696480 on Build GRCh38
rs182143903 Chr.3: 52696532-52696532 on Build GRCh38
rs149028763 Chr.3: 52696659-52696659 on Build GRCh38
rs997929426 Chr.3: 52696729-52696729 on Build GRCh38
rs918607531 Chr.3: 52696575-52696575 on Build GRCh38
rs374087931 Chr.3: 52696695-52696695 on Build GRCh38
rs931531936 Chr.3: 52696577-52696577 on Build GRCh38
rs956252033 Chr.3: 52696876-52696876 on Build GRCh38
rs766440617 Chr.3: 52696668-52696668 on Build GRCh38
rs754047790 Chr.3: 52696681-52696681 on Build GRCh38
rs779213575 Chr.3: 52696687-52696687 on Build GRCh38
rs749475685 Chr.3: 52696548-52696548 on Build GRCh38
rs369170184 Chr.3: 52696648-52696648 on Build GRCh38
rs992897501 Chr.3: 52696900-52696900 on Build GRCh38
rs910580297 Chr.3: 52696861-52696861 on Build GRCh38
rs565110212 Chr.3: 52696486-52696486 on Build GRCh38
rs537128327 Chr.3: 52696837-52696837 on Build GRCh38
rs751246705 Chr.3: 52696625-52696626 on Build GRCh38
rs547679854 Chr.3: 52696592-52696592 on Build GRCh38
rs779304727 Chr.3: 52696561-52696561 on Build GRCh38
rs201046447 Chr.3: 52696651-52696651 on Build GRCh38
rs767760453 Chr.3: 52696531-52696531 on Build GRCh38
rs752320771 Chr.3: 52696587-52696587 on Build GRCh38
rs985272460 Chr.3: 52696792-52696792 on Build GRCh38
rs756193263 Chr.3: 52696538-52696538 on Build GRCh38
rs752242342 Chr.3: 52696853-52696853 on Build GRCh38
rs142470202 Chr.3: 52696886-52696886 on Build GRCh38
rs745397707 Chr.3: 52696610-52696610 on Build GRCh38
rs972859946 Chr.3: 52696838-52696838 on Build GRCh38
rs757235136 Chr.3: 52696530-52696530 on Build GRCh38
rs948541998 Chr.3: 52696908-52696908 on Build GRCh38
rs755328809 Chr.3: 52696557-52696557 on Build GRCh38
rs775038978 Chr.3: 52696638-52696638 on Build GRCh38
rs918542416 Chr.3: 52696923-52696923 on Build GRCh38
rs550240727 Chr.3: 52696740-52696740 on Build GRCh38
rs560978935 Chr.3: 52696647-52696647 on Build GRCh38
rs750325658 Chr.3: 52696661-52696661 on Build GRCh38
rs759251524 Chr.3: 52696501-52696501 on Build GRCh38
rs777141931 Chr.3: 52696707-52696707 on Build GRCh38
rs762681832 Chr.3: 52696511-52696511 on Build GRCh38
rs56156188 Chr.3: 52696683-52696683 on Build GRCh38
rs376816132 Chr.3: 52696657-52696657 on Build GRCh38
rs773644788 Chr.3: 52696601-52696601 on Build GRCh38
rs762640255 Chr.3: 52696645-52696645 on Build GRCh38

View Full Product Details