Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52700172-52700669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT8D1
Entrez Gene ID
55830
Gene Name
glycosyltransferase 8 domain containing 1
Gene Aliases
AD-017, MSTP139
UniGene
Hs.297304
Gene Chromosome Location
Chr.3: 52694484-52706083 on Build GRCh38
Transcripts
NM_001010983.2 NM_001278280.1 NM_001278281.1 NM_018446.3 NM_152932.2 XM_017006857.1 XM_017006858.1 XM_017006859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3720442 c.5C>G p.S2* Chr.3: 52700456-52700456 on Build GRCh38
3724933 c.71A>G p.H24R Chr.3: 52700306-52700306 on Build GRCh38
6671823 c.115+2T>C p.? Chr.3: 52700260-52700260 on Build GRCh38
1047123 c.79C>A p.L27I Chr.3: 52700298-52700298 on Build GRCh38
4648730 c.53T>C p.F18S Chr.3: 52700324-52700324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750771517 Chr.3: 52700595-52700596 on Build GRCh38
rs760877919 Chr.3: 52700347-52700349 on Build GRCh38
rs768670793 Chr.3: 52700256-52700256 on Build GRCh38
rs745878683 Chr.3: 52700237-52700237 on Build GRCh38
rs568037002 Chr.3: 52700624-52700624 on Build GRCh38
rs189853421 Chr.3: 52700518-52700518 on Build GRCh38
rs779985722 Chr.3: 52700411-52700411 on Build GRCh38
rs937845946 Chr.3: 52700291-52700291 on Build GRCh38
rs3733043 Chr.3: 52700472-52700472 on Build GRCh38
rs754349000 Chr.3: 52700349-52700349 on Build GRCh38
rs1031586138 Chr.3: 52700288-52700288 on Build GRCh38
rs749380855 Chr.3: 52700247-52700247 on Build GRCh38
rs780829629 Chr.3: 52700236-52700236 on Build GRCh38
rs748458376 Chr.3: 52700451-52700451 on Build GRCh38
rs772179165 Chr.3: 52700460-52700460 on Build GRCh38
rs375560836 Chr.3: 52700344-52700344 on Build GRCh38
rs758015662 Chr.3: 52700352-52700352 on Build GRCh38
rs774662293 Chr.3: 52700303-52700303 on Build GRCh38
rs569529695 Chr.3: 52700615-52700615 on Build GRCh38
rs974874229 Chr.3: 52700404-52700404 on Build GRCh38
rs760100618 Chr.3: 52700495-52700495 on Build GRCh38
rs147975364 Chr.3: 52700616-52700616 on Build GRCh38
rs1003171237 Chr.3: 52700273-52700273 on Build GRCh38
rs535256411 Chr.3: 52700514-52700514 on Build GRCh38
rs942313842 Chr.3: 52700533-52700533 on Build GRCh38
rs368586218 Chr.3: 52700246-52700246 on Build GRCh38
rs971087951 Chr.3: 52700269-52700269 on Build GRCh38
rs944393246 Chr.3: 52700640-52700640 on Build GRCh38
rs756618663 Chr.3: 52700375-52700376 on Build GRCh38
rs756555842 Chr.3: 52700220-52700220 on Build GRCh38
rs369893637 Chr.3: 52700233-52700233 on Build GRCh38
rs761907814 Chr.3: 52700329-52700329 on Build GRCh38
rs920518318 Chr.3: 52700306-52700306 on Build GRCh38
rs990694991 Chr.3: 52700304-52700304 on Build GRCh38
rs761143737 Chr.3: 52700475-52700475 on Build GRCh38
rs1006343137 Chr.3: 52700207-52700207 on Build GRCh38
rs982247225 Chr.3: 52700250-52700250 on Build GRCh38
rs78696896 Chr.3: 52700577-52700579 on Build GRCh38
rs144625501 Chr.3: 52700339-52700339 on Build GRCh38
rs959157572 Chr.3: 52700604-52700604 on Build GRCh38
rs768760770 Chr.3: 52700423-52700423 on Build GRCh38
rs756849873 Chr.3: 52700369-52700369 on Build GRCh38
rs564548558 Chr.3: 52700232-52700232 on Build GRCh38
rs771383214 Chr.3: 52700477-52700477 on Build GRCh38
rs751277703 Chr.3: 52700367-52700367 on Build GRCh38
rs764897884 Chr.3: 52700221-52700221 on Build GRCh38
rs773435276 Chr.3: 52700341-52700341 on Build GRCh38
rs371635121 Chr.3: 52700371-52700371 on Build GRCh38
rs764443274 Chr.3: 52700414-52700414 on Build GRCh38
rs763721091 Chr.3: 52700498-52700498 on Build GRCh38
rs368813501 Chr.3: 52700354-52700354 on Build GRCh38
rs376655141 Chr.3: 52700384-52700384 on Build GRCh38
rs761331698 Chr.3: 52700343-52700343 on Build GRCh38
rs777320257 Chr.3: 52700488-52700488 on Build GRCh38
rs955982722 Chr.3: 52700314-52700314 on Build GRCh38
rs895335650 Chr.3: 52700579-52700579 on Build GRCh38
rs967178759 Chr.3: 52700234-52700234 on Build GRCh38
rs773583706 Chr.3: 52700565-52700565 on Build GRCh38
rs370542404 Chr.3: 52700226-52700226 on Build GRCh38
rs763482728 Chr.3: 52700364-52700364 on Build GRCh38
rs751030830 Chr.3: 52700512-52700512 on Build GRCh38
rs375896519 Chr.3: 52700222-52700222 on Build GRCh38
rs372164481 Chr.3: 52700281-52700281 on Build GRCh38
rs202161515 Chr.3: 52700464-52700464 on Build GRCh38
rs769721940 Chr.3: 52700244-52700244 on Build GRCh38
rs930609560 Chr.3: 52700307-52700307 on Build GRCh38
rs1037802531 Chr.3: 52700535-52700535 on Build GRCh38
rs3733042 Chr.3: 52700209-52700209 on Build GRCh38
rs749181075 Chr.3: 52700420-52700420 on Build GRCh38
rs773466399 Chr.3: 52700420-52700424 on Build GRCh38
rs767006628 Chr.3: 52700347-52700347 on Build GRCh38
rs200901885 Chr.3: 52700428-52700428 on Build GRCh38
rs571905232 Chr.3: 52700614-52700614 on Build GRCh38
rs778834885 Chr.3: 52700450-52700450 on Build GRCh38

View Full Product Details