Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53285491-53286002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP1A
Entrez Gene ID
55802
Gene Name
decapping mRNA 1A
Gene Aliases
HSA275986, Nbla00360, SMAD4IP1, SMIF
UniGene
Hs.476353
Gene Chromosome Location
Chr.3: 53283423-53347627 on Build GRCh38
Transcripts
NM_001290204.1 NM_001290205.1 NM_001290206.1 NM_001290207.1 NM_018403.6

SNP Detail

SNP ID Chromosome Location
rs989018115 Chr.3: 53285848-53285848 on Build GRCh38
rs928865216 Chr.3: 53285522-53285522 on Build GRCh38
rs921987383 Chr.3: 53285943-53285943 on Build GRCh38
rs968675206 Chr.3: 53285902-53285902 on Build GRCh38
rs960088185 Chr.3: 53285566-53285566 on Build GRCh38
rs532660761 Chr.3: 53285583-53285583 on Build GRCh38
rs995290369 Chr.3: 53285782-53285782 on Build GRCh38
rs925144603 Chr.3: 53285649-53285649 on Build GRCh38
rs782378748 Chr.3: 53285558-53285558 on Build GRCh38
rs370008372 Chr.3: 53285733-53285733 on Build GRCh38
rs1049684597 Chr.3: 53285699-53285699 on Build GRCh38
rs932457595 Chr.3: 53285682-53285682 on Build GRCh38
rs73843110 Chr.3: 53285795-53285795 on Build GRCh38
rs1043111789 Chr.3: 53285632-53285632 on Build GRCh38
rs367930926 Chr.3: 53285809-53285809 on Build GRCh38
rs782030173 Chr.3: 53285894-53285894 on Build GRCh38
rs191354220 Chr.3: 53285744-53285744 on Build GRCh38
rs1026722944 Chr.3: 53285786-53285786 on Build GRCh38
rs913320312 Chr.3: 53285587-53285587 on Build GRCh38
rs73843111 Chr.3: 53285816-53285816 on Build GRCh38
rs782143973 Chr.3: 53285942-53285942 on Build GRCh38
rs548543995 Chr.3: 53285766-53285766 on Build GRCh38
rs947512682 Chr.3: 53285611-53285611 on Build GRCh38
rs960536232 Chr.3: 53285823-53285823 on Build GRCh38
rs531244759 Chr.3: 53285810-53285810 on Build GRCh38
rs1036661235 Chr.3: 53285746-53285746 on Build GRCh38

View Full Product Details