Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53287951-53288448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP1A
Entrez Gene ID
55802
Gene Name
decapping mRNA 1A
Gene Aliases
HSA275986, Nbla00360, SMAD4IP1, SMIF
UniGene
Hs.476353
Gene Chromosome Location
Chr.3: 53283423-53347627 on Build GRCh38
Transcripts
NM_001290204.1 NM_001290205.1 NM_001290206.1 NM_001290207.1 NM_018403.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7413484 c.1609A>T p.K537* Chr.3: 53288124-53288124 on Build GRCh38
1047276 c.1545G>A p.S515S Chr.3: 53288188-53288188 on Build GRCh38
5580789 c.1644G>T p.Q548H Chr.3: 53288089-53288089 on Build GRCh38
4676570 c.1458C>T p.G486G Chr.3: 53288275-53288275 on Build GRCh38
4676569 c.1493C>T p.S498F Chr.3: 53288240-53288240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781830896 Chr.3: 53288268-53288268 on Build GRCh38
rs782798835 Chr.3: 53288254-53288254 on Build GRCh38
rs782606929 Chr.3: 53288190-53288190 on Build GRCh38
rs903074995 Chr.3: 53288381-53288381 on Build GRCh38
rs782317560 Chr.3: 53288198-53288198 on Build GRCh38
rs376857911 Chr.3: 53288336-53288336 on Build GRCh38
rs60392580 Chr.3: 53288356-53288356 on Build GRCh38
rs782283971 Chr.3: 53288051-53288051 on Build GRCh38
rs909201716 Chr.3: 53288148-53288148 on Build GRCh38
rs782769306 Chr.3: 53288259-53288259 on Build GRCh38
rs782712782 Chr.3: 53288149-53288149 on Build GRCh38
rs782354513 Chr.3: 53288292-53288292 on Build GRCh38
rs782504004 Chr.3: 53288019-53288019 on Build GRCh38
rs782467938 Chr.3: 53288141-53288141 on Build GRCh38
rs781939540 Chr.3: 53288092-53288092 on Build GRCh38
rs782643597 Chr.3: 53288289-53288289 on Build GRCh38
rs782048816 Chr.3: 53288242-53288242 on Build GRCh38
rs782395545 Chr.3: 53288203-53288203 on Build GRCh38
rs782402360 Chr.3: 53288053-53288053 on Build GRCh38
rs374895560 Chr.3: 53288049-53288049 on Build GRCh38
rs371858151 Chr.3: 53288275-53288275 on Build GRCh38
rs137967603 Chr.3: 53288311-53288311 on Build GRCh38
rs782135745 Chr.3: 53288122-53288122 on Build GRCh38
rs564891451 Chr.3: 53288332-53288332 on Build GRCh38
rs782021577 Chr.3: 53288112-53288112 on Build GRCh38
rs782245439 Chr.3: 53288290-53288290 on Build GRCh38
rs782109945 Chr.3: 53288056-53288056 on Build GRCh38
rs782806485 Chr.3: 53288100-53288100 on Build GRCh38
rs781933192 Chr.3: 53288225-53288225 on Build GRCh38
rs782055343 Chr.3: 53288097-53288097 on Build GRCh38
rs781984644 Chr.3: 53288220-53288220 on Build GRCh38
rs782206853 Chr.3: 53288030-53288030 on Build GRCh38
rs782145629 Chr.3: 53288321-53288321 on Build GRCh38
rs782318047 Chr.3: 53288044-53288044 on Build GRCh38
rs782496513 Chr.3: 53288188-53288188 on Build GRCh38
rs782575567 Chr.3: 53288274-53288274 on Build GRCh38
rs782541384 Chr.3: 53288280-53288280 on Build GRCh38
rs565921330 Chr.3: 53288025-53288025 on Build GRCh38
rs372737171 Chr.3: 53288054-53288054 on Build GRCh38
rs577394517 Chr.3: 53288350-53288350 on Build GRCh38
rs782729017 Chr.3: 53288224-53288224 on Build GRCh38
rs782558157 Chr.3: 53288163-53288163 on Build GRCh38
rs966590415 Chr.3: 53288048-53288048 on Build GRCh38
rs782029481 Chr.3: 53288315-53288315 on Build GRCh38
rs557290581 Chr.3: 53288337-53288337 on Build GRCh38
rs781785116 Chr.3: 53288150-53288150 on Build GRCh38
rs957106301 Chr.3: 53287998-53287998 on Build GRCh38
rs782505743 Chr.3: 53288255-53288255 on Build GRCh38
rs368451324 Chr.3: 53288164-53288164 on Build GRCh38
rs782205179 Chr.3: 53288191-53288191 on Build GRCh38
rs986283642 Chr.3: 53288008-53288008 on Build GRCh38
rs782100430 Chr.3: 53288221-53288221 on Build GRCh38
rs782197746 Chr.3: 53288312-53288312 on Build GRCh38
rs369219389 Chr.3: 53288000-53288000 on Build GRCh38
rs782250755 Chr.3: 53288173-53288173 on Build GRCh38
rs957256318 Chr.3: 53288411-53288411 on Build GRCh38
rs537210744 Chr.3: 53288313-53288313 on Build GRCh38
rs782463571 Chr.3: 53288271-53288271 on Build GRCh38
rs782275786 Chr.3: 53288199-53288199 on Build GRCh38
rs553736453 Chr.3: 53288385-53288385 on Build GRCh38
rs62256054 Chr.3: 53288413-53288413 on Build GRCh38
rs142446763 Chr.3: 53288386-53288386 on Build GRCh38
rs782351755 Chr.3: 53288076-53288076 on Build GRCh38

View Full Product Details