Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109105570-109106071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf194
Entrez Gene ID
127003
Gene Name
chromosome 1 open reading frame 194
Gene Aliases
-
UniGene
Hs.446962
Gene Chromosome Location
Chr.1: 109105951-109113877 on Build GRCh38
Transcripts
NM_001122961.1 XM_005270448.3 XM_006710342.3 XM_006710343.3 XM_011540647.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6615214 c.442C>T p.R148* Chr.1: 109106060-109106060 on Build GRCh38
3975837 c.468C>T p.S156S Chr.1: 109106034-109106034 on Build GRCh38
6615215 c.478C>T p.R160* Chr.1: 109106060-109106060 on Build GRCh38
3671332 c.478C>A p.R160R Chr.1: 109106060-109106060 on Build GRCh38
4667360 c.479G>A p.R160Q Chr.1: 109106059-109106059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000298997 Chr.1: 109105868-109105868 on Build GRCh38
rs968369548 Chr.1: 109105606-109105606 on Build GRCh38
rs918600904 Chr.1: 109106023-109106023 on Build GRCh38
rs879058012 Chr.1: 109105905-109105905 on Build GRCh38
rs111902136 Chr.1: 109105830-109105830 on Build GRCh38
rs536121053 Chr.1: 109105980-109105980 on Build GRCh38
rs572325378 Chr.1: 109105860-109105860 on Build GRCh38
rs953903048 Chr.1: 109105739-109105739 on Build GRCh38
rs1033957744 Chr.1: 109105620-109105620 on Build GRCh38
rs909854128 Chr.1: 109105750-109105750 on Build GRCh38
rs918694072 Chr.1: 109105795-109105795 on Build GRCh38
rs941375753 Chr.1: 109105751-109105751 on Build GRCh38
rs891578665 Chr.1: 109105910-109105910 on Build GRCh38
rs766679425 Chr.1: 109106008-109106008 on Build GRCh38
rs761115597 Chr.1: 109106005-109106005 on Build GRCh38
rs572792451 Chr.1: 109106034-109106034 on Build GRCh38
rs879035229 Chr.1: 109105913-109105913 on Build GRCh38
rs539018003 Chr.1: 109105807-109105807 on Build GRCh38
rs1020384052 Chr.1: 109106038-109106038 on Build GRCh38
rs777743007 Chr.1: 109105914-109105914 on Build GRCh38
rs773718206 Chr.1: 109106002-109106002 on Build GRCh38
rs1031862369 Chr.1: 109105610-109105610 on Build GRCh38
rs553006820 Chr.1: 109105743-109105743 on Build GRCh38
rs1037098803 Chr.1: 109105790-109105790 on Build GRCh38
rs1001768590 Chr.1: 109105611-109105611 on Build GRCh38
rs999925714 Chr.1: 109105607-109105607 on Build GRCh38
rs759918072 Chr.1: 109105973-109105973 on Build GRCh38
rs536360865 Chr.1: 109105691-109105691 on Build GRCh38
rs921922562 Chr.1: 109105841-109105841 on Build GRCh38
rs907386484 Chr.1: 109105909-109105909 on Build GRCh38
rs575875898 Chr.1: 109105837-109105837 on Build GRCh38
rs577763411 Chr.1: 109105861-109105861 on Build GRCh38
rs754196688 Chr.1: 109106018-109106018 on Build GRCh38
rs977024700 Chr.1: 109105684-109105684 on Build GRCh38
rs1034598558 Chr.1: 109105925-109105925 on Build GRCh38
rs922510275 Chr.1: 109105722-109105722 on Build GRCh38
rs370124504 Chr.1: 109106047-109106047 on Build GRCh38
rs986623437 Chr.1: 109105771-109105771 on Build GRCh38
rs199625921 Chr.1: 109105800-109105800 on Build GRCh38
rs1029466594 Chr.1: 109105933-109105933 on Build GRCh38
rs930598628 Chr.1: 109105855-109105855 on Build GRCh38
rs150611714 Chr.1: 109105986-109105986 on Build GRCh38
rs1031411310 Chr.1: 109105869-109105869 on Build GRCh38
rs907420959 Chr.1: 109105600-109105600 on Build GRCh38
rs962726882 Chr.1: 109105992-109105992 on Build GRCh38
rs567093550 Chr.1: 109105671-109105671 on Build GRCh38
rs370368387 Chr.1: 109105987-109105987 on Build GRCh38
rs774344711 Chr.1: 109105985-109105985 on Build GRCh38
rs879178143 Chr.1: 109105881-109105882 on Build GRCh38
rs538513227 Chr.1: 109105843-109105843 on Build GRCh38
rs997947315 Chr.1: 109105929-109105929 on Build GRCh38
rs953870343 Chr.1: 109105938-109105938 on Build GRCh38
rs763717608 Chr.1: 109106043-109106043 on Build GRCh38
rs948280213 Chr.1: 109105858-109105858 on Build GRCh38
rs893396758 Chr.1: 109105938-109105939 on Build GRCh38
rs989849308 Chr.1: 109105654-109105654 on Build GRCh38
rs373630514 Chr.1: 109105630-109105630 on Build GRCh38
rs764925629 Chr.1: 109106028-109106028 on Build GRCh38
rs759158389 Chr.1: 109106021-109106021 on Build GRCh38
rs375722101 Chr.1: 109106044-109106044 on Build GRCh38

View Full Product Details