Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109252443-109252944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR2
Entrez Gene ID
1952
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Gene Aliases
ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3
UniGene
Hs.57652
Gene Chromosome Location
Chr.1: 109249705-109275756 on Build GRCh38
Transcripts
NM_001408.2 XM_005270580.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5457651 c.2624C>T p.T875M Chr.1: 109252703-109252703 on Build GRCh38
5907128 c.2365G>A p.D789N Chr.1: 109252444-109252444 on Build GRCh38
674477 c.2704C>T p.R902C Chr.1: 109252783-109252783 on Build GRCh38
109002 c.2641C>T p.R881* Chr.1: 109252720-109252720 on Build GRCh38
674478 c.2575G>C p.D859H Chr.1: 109252654-109252654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565216430 Chr.1: 109252653-109252653 on Build GRCh38
rs373425704 Chr.1: 109252480-109252480 on Build GRCh38
rs573729296 Chr.1: 109252513-109252513 on Build GRCh38
rs776946440 Chr.1: 109252726-109252726 on Build GRCh38
rs770965352 Chr.1: 109252709-109252709 on Build GRCh38
rs756947114 Chr.1: 109252623-109252623 on Build GRCh38
rs766127459 Chr.1: 109252705-109252705 on Build GRCh38
rs771982823 Chr.1: 109252640-109252640 on Build GRCh38
rs769134813 Chr.1: 109252536-109252536 on Build GRCh38
rs759867572 Chr.1: 109252728-109252728 on Build GRCh38
rs149656610 Chr.1: 109252904-109252904 on Build GRCh38
rs201437569 Chr.1: 109252827-109252827 on Build GRCh38
rs962268647 Chr.1: 109252704-109252704 on Build GRCh38
rs781423989 Chr.1: 109252866-109252866 on Build GRCh38
rs529873677 Chr.1: 109252735-109252735 on Build GRCh38
rs751488741 Chr.1: 109252761-109252761 on Build GRCh38
rs377135836 Chr.1: 109252661-109252661 on Build GRCh38
rs915968408 Chr.1: 109252775-109252775 on Build GRCh38
rs762773564 Chr.1: 109252897-109252897 on Build GRCh38
rs774201169 Chr.1: 109252680-109252681 on Build GRCh38
rs762667468 Chr.1: 109252686-109252686 on Build GRCh38
rs764084072 Chr.1: 109252752-109252752 on Build GRCh38
rs35689464 Chr.1: 109252513-109252514 on Build GRCh38
rs367691599 Chr.1: 109252729-109252729 on Build GRCh38
rs757931859 Chr.1: 109252603-109252603 on Build GRCh38
rs753105253 Chr.1: 109252812-109252812 on Build GRCh38
rs761847500 Chr.1: 109252553-109252553 on Build GRCh38
rs879027971 Chr.1: 109252863-109252863 on Build GRCh38
rs751067182 Chr.1: 109252668-109252668 on Build GRCh38
rs775438398 Chr.1: 109252741-109252741 on Build GRCh38
rs755819249 Chr.1: 109252473-109252473 on Build GRCh38
rs780762006 Chr.1: 109252624-109252624 on Build GRCh38
rs776854763 Chr.1: 109252907-109252907 on Build GRCh38
rs375909210 Chr.1: 109252808-109252808 on Build GRCh38
rs145105634 Chr.1: 109252525-109252525 on Build GRCh38
rs747494318 Chr.1: 109252843-109252843 on Build GRCh38
rs758729822 Chr.1: 109252836-109252836 on Build GRCh38
rs774815074 Chr.1: 109252632-109252632 on Build GRCh38
rs775920971 Chr.1: 109252896-109252896 on Build GRCh38
rs759318308 Chr.1: 109252765-109252765 on Build GRCh38
rs767034475 Chr.1: 109252781-109252781 on Build GRCh38
rs143186589 Chr.1: 109252673-109252673 on Build GRCh38
rs773358358 Chr.1: 109252644-109252644 on Build GRCh38
rs777270805 Chr.1: 109252511-109252511 on Build GRCh38
rs754886779 Chr.1: 109252579-109252579 on Build GRCh38
rs151136134 Chr.1: 109252611-109252611 on Build GRCh38
rs745566729 Chr.1: 109252685-109252685 on Build GRCh38
rs770126332 Chr.1: 109252881-109252881 on Build GRCh38
rs1026244756 Chr.1: 109252622-109252622 on Build GRCh38
rs772622829 Chr.1: 109252703-109252703 on Build GRCh38
rs749644861 Chr.1: 109252628-109252628 on Build GRCh38
rs750573929 Chr.1: 109252561-109252561 on Build GRCh38
rs953299138 Chr.1: 109252492-109252492 on Build GRCh38
rs368556615 Chr.1: 109252847-109252847 on Build GRCh38
rs371935118 Chr.1: 109252792-109252792 on Build GRCh38
rs370447679 Chr.1: 109252663-109252663 on Build GRCh38
rs770574783 Chr.1: 109252678-109252679 on Build GRCh38
rs962076401 Chr.1: 109252509-109252509 on Build GRCh38
rs928776423 Chr.1: 109252514-109252514 on Build GRCh38
rs374928321 Chr.1: 109252707-109252707 on Build GRCh38
rs1039838854 Chr.1: 109252597-109252597 on Build GRCh38
rs779256212 Chr.1: 109252695-109252695 on Build GRCh38
rs779744418 Chr.1: 109252810-109252810 on Build GRCh38
rs761361328 Chr.1: 109252779-109252779 on Build GRCh38
rs199806460 Chr.1: 109252787-109252787 on Build GRCh38
rs778322362 Chr.1: 109252842-109252842 on Build GRCh38
rs142987028 Chr.1: 109252497-109252497 on Build GRCh38
rs370616613 Chr.1: 109252651-109252651 on Build GRCh38
rs554845360 Chr.1: 109252494-109252494 on Build GRCh38
rs376119081 Chr.1: 109252501-109252501 on Build GRCh38
rs991646398 Chr.1: 109252565-109252565 on Build GRCh38
rs369355603 Chr.1: 109252467-109252467 on Build GRCh38
rs201283895 Chr.1: 109252654-109252654 on Build GRCh38
rs201340375 Chr.1: 109252570-109252570 on Build GRCh38
rs370538970 Chr.1: 109252503-109252503 on Build GRCh38
rs774901050 Chr.1: 109252548-109252548 on Build GRCh38
rs376437956 Chr.1: 109252578-109252578 on Build GRCh38
rs753655956 Chr.1: 109252495-109252495 on Build GRCh38
rs748599721 Chr.1: 109252698-109252698 on Build GRCh38
rs535934834 Chr.1: 109252572-109252572 on Build GRCh38
rs763391847 Chr.1: 109252529-109252529 on Build GRCh38
rs138887323 Chr.1: 109252708-109252708 on Build GRCh38
rs752733978 Chr.1: 109252587-109252587 on Build GRCh38
rs756780197 Chr.1: 109252670-109252670 on Build GRCh38
rs572525434 Chr.1: 109252621-109252621 on Build GRCh38
rs760728043 Chr.1: 109252650-109252650 on Build GRCh38
rs373423391 Chr.1: 109252557-109252557 on Build GRCh38
rs544567629 Chr.1: 109252630-109252630 on Build GRCh38
rs747656635 Chr.1: 109252500-109252500 on Build GRCh38
rs150411435 Chr.1: 109252662-109252662 on Build GRCh38
rs748687661 Chr.1: 109252635-109252635 on Build GRCh38
rs987828949 Chr.1: 109252747-109252747 on Build GRCh38
rs750694881 Chr.1: 109252471-109252471 on Build GRCh38
rs150322062 Chr.1: 109252720-109252720 on Build GRCh38
rs1006432764 Chr.1: 109252680-109252680 on Build GRCh38
rs138048818 Chr.1: 109252748-109252748 on Build GRCh38
rs761685003 Chr.1: 109252923-109252923 on Build GRCh38
rs746574787 Chr.1: 109252618-109252618 on Build GRCh38
rs901619773 Chr.1: 109252637-109252637 on Build GRCh38
rs141691609 Chr.1: 109252867-109252867 on Build GRCh38
rs765237291 Chr.1: 109252584-109252584 on Build GRCh38
rs760959590 Chr.1: 109252567-109252567 on Build GRCh38

View Full Product Details