Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109486927-109487438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L2
Entrez Gene ID
127002
Gene Name
ataxin 7 like 2
Gene Aliases
-
UniGene
Hs.118248
Gene Chromosome Location
Chr.1: 109483904-109492804 on Build GRCh38
Transcripts
NM_153340.4 XM_005270445.3 XM_005270446.2 XM_011540638.2 XM_011540639.2 XM_011540641.2 XM_011540644.2 XM_011540645.1 XM_011540646.2 XM_017000267.1 XM_017000268.1 XM_017000269.1 XM_017000270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7413071 c.340C>T p.P114S Chr.1: 109487048-109487048 on Build GRCh38
3360204 c.307C>A p.H103N Chr.1: 109487015-109487015 on Build GRCh38
7315609 c.461G>A p.R154Q Chr.1: 109487169-109487169 on Build GRCh38
3984063 c.305G>C p.R102T Chr.1: 109487013-109487013 on Build GRCh38
1332453 c.310delG p.L106fs*23 Chr.1: 109487018-109487018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771054504 Chr.1: 109487211-109487211 on Build GRCh38
rs753070202 Chr.1: 109487058-109487058 on Build GRCh38
rs771115277 Chr.1: 109486986-109486986 on Build GRCh38
rs749559056 Chr.1: 109487123-109487123 on Build GRCh38
rs945089830 Chr.1: 109487066-109487066 on Build GRCh38
rs980758012 Chr.1: 109487027-109487027 on Build GRCh38
rs759323358 Chr.1: 109487168-109487168 on Build GRCh38
rs988910209 Chr.1: 109487046-109487046 on Build GRCh38
rs767012090 Chr.1: 109487048-109487048 on Build GRCh38
rs530362212 Chr.1: 109487398-109487398 on Build GRCh38
rs915602677 Chr.1: 109487052-109487052 on Build GRCh38
rs753413419 Chr.1: 109487166-109487166 on Build GRCh38
rs572464653 Chr.1: 109487169-109487169 on Build GRCh38
rs566994045 Chr.1: 109487063-109487063 on Build GRCh38
rs557495851 Chr.1: 109487250-109487250 on Build GRCh38
rs778458196 Chr.1: 109486974-109486974 on Build GRCh38
rs780237234 Chr.1: 109487223-109487223 on Build GRCh38
rs755734875 Chr.1: 109487050-109487050 on Build GRCh38
rs147848834 Chr.1: 109487089-109487089 on Build GRCh38
rs544008937 Chr.1: 109487375-109487375 on Build GRCh38
rs1039347248 Chr.1: 109487174-109487174 on Build GRCh38
rs776638868 Chr.1: 109486999-109486999 on Build GRCh38
rs899418263 Chr.1: 109487227-109487227 on Build GRCh38
rs537983779 Chr.1: 109487043-109487043 on Build GRCh38
rs140914074 Chr.1: 109487024-109487024 on Build GRCh38
rs955573969 Chr.1: 109487406-109487406 on Build GRCh38
rs934469562 Chr.1: 109487120-109487120 on Build GRCh38
rs777131683 Chr.1: 109487086-109487086 on Build GRCh38
rs771917655 Chr.1: 109487139-109487139 on Build GRCh38
rs751983839 Chr.1: 109487175-109487175 on Build GRCh38
rs774200325 Chr.1: 109487133-109487133 on Build GRCh38
rs868523408 Chr.1: 109487298-109487298 on Build GRCh38
rs753745085 Chr.1: 109487177-109487179 on Build GRCh38
rs910656265 Chr.1: 109487399-109487399 on Build GRCh38
rs925903750 Chr.1: 109487096-109487096 on Build GRCh38
rs778333358 Chr.1: 109487067-109487067 on Build GRCh38
rs372080468 Chr.1: 109487249-109487249 on Build GRCh38
rs899113772 Chr.1: 109487179-109487179 on Build GRCh38
rs367843295 Chr.1: 109487230-109487230 on Build GRCh38
rs778623864 Chr.1: 109487258-109487258 on Build GRCh38
rs1024276291 Chr.1: 109486993-109486993 on Build GRCh38
rs769917745 Chr.1: 109487020-109487020 on Build GRCh38
rs950753549 Chr.1: 109486956-109486956 on Build GRCh38
rs561455373 Chr.1: 109487387-109487387 on Build GRCh38
rs371721090 Chr.1: 109487178-109487178 on Build GRCh38
rs970117271 Chr.1: 109486996-109486996 on Build GRCh38
rs1014339179 Chr.1: 109487283-109487283 on Build GRCh38
rs770929516 Chr.1: 109487397-109487397 on Build GRCh38
rs747562288 Chr.1: 109486982-109486982 on Build GRCh38
rs988596398 Chr.1: 109487377-109487377 on Build GRCh38
rs867875669 Chr.1: 109487021-109487021 on Build GRCh38
rs922477972 Chr.1: 109487379-109487379 on Build GRCh38
rs747960071 Chr.1: 109487262-109487262 on Build GRCh38
rs765957487 Chr.1: 109487160-109487160 on Build GRCh38
rs1033301264 Chr.1: 109487365-109487365 on Build GRCh38
rs752104865 Chr.1: 109487073-109487073 on Build GRCh38
rs970097099 Chr.1: 109487315-109487315 on Build GRCh38
rs370525139 Chr.1: 109487134-109487134 on Build GRCh38
rs970443913 Chr.1: 109487332-109487332 on Build GRCh38
rs757840300 Chr.1: 109487074-109487074 on Build GRCh38
rs749908193 Chr.1: 109487049-109487049 on Build GRCh38
rs369195330 Chr.1: 109487004-109487004 on Build GRCh38
rs770017357 Chr.1: 109487111-109487111 on Build GRCh38
rs373697288 Chr.1: 109487182-109487182 on Build GRCh38
rs762687905 Chr.1: 109487033-109487033 on Build GRCh38
rs765969369 Chr.1: 109487057-109487057 on Build GRCh38
rs573541171 Chr.1: 109487105-109487105 on Build GRCh38
rs772006220 Chr.1: 109487269-109487269 on Build GRCh38
rs556723283 Chr.1: 109487044-109487044 on Build GRCh38
rs201678517 Chr.1: 109487042-109487042 on Build GRCh38
rs780107005 Chr.1: 109487122-109487122 on Build GRCh38
rs201025666 Chr.1: 109487142-109487142 on Build GRCh38
rs552850912 Chr.1: 109487144-109487144 on Build GRCh38
rs992914065 Chr.1: 109486964-109486964 on Build GRCh38
rs138423700 Chr.1: 109487356-109487356 on Build GRCh38
rs564799186 Chr.1: 109487256-109487256 on Build GRCh38
rs943325913 Chr.1: 109487410-109487410 on Build GRCh38
rs913422138 Chr.1: 109487055-109487055 on Build GRCh38
rs758808306 Chr.1: 109487065-109487065 on Build GRCh38

View Full Product Details