Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109487527-109488035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L2
Entrez Gene ID
127002
Gene Name
ataxin 7 like 2
Gene Aliases
-
UniGene
Hs.118248
Gene Chromosome Location
Chr.1: 109483904-109492804 on Build GRCh38
Transcripts
NM_153340.4 XM_005270445.3 XM_005270446.2 XM_011540638.2 XM_011540639.2 XM_011540641.2 XM_011540644.2 XM_011540645.1 XM_011540646.2 XM_017000267.1 XM_017000268.1 XM_017000269.1 XM_017000270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4596966 c.608G>A p.S203N Chr.1: 109487712-109487712 on Build GRCh38
4518211 c.561_562CC>TT p.P188S Chr.1: 109487665-109487666 on Build GRCh38
279087 c.594C>T p.I198I Chr.1: 109487698-109487698 on Build GRCh38
4502047 c.603C>T p.I201I Chr.1: 109487707-109487707 on Build GRCh38
4502099 c.604C>T p.P202S Chr.1: 109487708-109487708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770910741 Chr.1: 109487744-109487744 on Build GRCh38
rs773629792 Chr.1: 109487791-109487791 on Build GRCh38
rs746919882 Chr.1: 109487734-109487734 on Build GRCh38
rs758717010 Chr.1: 109487826-109487826 on Build GRCh38
rs907820245 Chr.1: 109487848-109487848 on Build GRCh38
rs765000245 Chr.1: 109487628-109487628 on Build GRCh38
rs1047659640 Chr.1: 109487838-109487838 on Build GRCh38
rs1010217919 Chr.1: 109487959-109487959 on Build GRCh38
rs910593265 Chr.1: 109487709-109487709 on Build GRCh38
rs753874481 Chr.1: 109487793-109487793 on Build GRCh38
rs36079114 Chr.1: 109487764-109487765 on Build GRCh38
rs756709396 Chr.1: 109487650-109487650 on Build GRCh38
rs746841246 Chr.1: 109487835-109487835 on Build GRCh38
rs762052633 Chr.1: 109487634-109487634 on Build GRCh38
rs767766035 Chr.1: 109487773-109487773 on Build GRCh38
rs750879373 Chr.1: 109487646-109487646 on Build GRCh38
rs569193075 Chr.1: 109487632-109487632 on Build GRCh38
rs891352070 Chr.1: 109487977-109487977 on Build GRCh38
rs182100153 Chr.1: 109487904-109487904 on Build GRCh38
rs911395893 Chr.1: 109487757-109487757 on Build GRCh38
rs943500581 Chr.1: 109487710-109487710 on Build GRCh38
rs1032712508 Chr.1: 109487562-109487562 on Build GRCh38
rs778877453 Chr.1: 109487823-109487823 on Build GRCh38
rs77853011 Chr.1: 109487905-109487905 on Build GRCh38
rs905695614 Chr.1: 109487899-109487899 on Build GRCh38
rs761030840 Chr.1: 109487792-109487792 on Build GRCh38
rs957609470 Chr.1: 109487663-109487663 on Build GRCh38
rs755021247 Chr.1: 109487800-109487800 on Build GRCh38
rs772997212 Chr.1: 109487563-109487563 on Build GRCh38
rs770335760 Chr.1: 109487599-109487599 on Build GRCh38
rs760717788 Chr.1: 109487593-109487593 on Build GRCh38
rs1054983099 Chr.1: 109487912-109487912 on Build GRCh38
rs1054510048 Chr.1: 109487927-109487927 on Build GRCh38
rs759203140 Chr.1: 109487622-109487622 on Build GRCh38
rs745399084 Chr.1: 109487760-109487760 on Build GRCh38
rs34784291 Chr.1: 109487754-109487755 on Build GRCh38
rs776229888 Chr.1: 109487750-109487750 on Build GRCh38
rs929533508 Chr.1: 109487787-109487787 on Build GRCh38
rs372058914 Chr.1: 109487804-109487804 on Build GRCh38
rs1032975327 Chr.1: 109487641-109487641 on Build GRCh38
rs149490071 Chr.1: 109487682-109487682 on Build GRCh38
rs536643831 Chr.1: 109487569-109487569 on Build GRCh38
rs554704029 Chr.1: 109487894-109487894 on Build GRCh38
rs114944620 Chr.1: 109487819-109487819 on Build GRCh38
rs777370940 Chr.1: 109487834-109487834 on Build GRCh38
rs1044576504 Chr.1: 109487886-109487886 on Build GRCh38
rs188638622 Chr.1: 109487551-109487551 on Build GRCh38
rs369959556 Chr.1: 109487698-109487698 on Build GRCh38
rs146979128 Chr.1: 109487635-109487635 on Build GRCh38
rs192545791 Chr.1: 109487665-109487665 on Build GRCh38
rs781240476 Chr.1: 109487847-109487847 on Build GRCh38
rs949961364 Chr.1: 109487871-109487871 on Build GRCh38
rs867885012 Chr.1: 109487829-109487829 on Build GRCh38
rs878928190 Chr.1: 109487592-109487592 on Build GRCh38
rs779137740 Chr.1: 109487684-109487684 on Build GRCh38
rs891491056 Chr.1: 109487934-109487934 on Build GRCh38
rs78926405 Chr.1: 109487689-109487689 on Build GRCh38
rs552524351 Chr.1: 109487610-109487610 on Build GRCh38
rs762724236 Chr.1: 109487772-109487772 on Build GRCh38
rs769465395 Chr.1: 109487769-109487769 on Build GRCh38
rs555529624 Chr.1: 109487938-109487938 on Build GRCh38
rs373813012 Chr.1: 109487719-109487719 on Build GRCh38
rs1020482779 Chr.1: 109487969-109487969 on Build GRCh38
rs775260176 Chr.1: 109487770-109487770 on Build GRCh38
rs753929611 Chr.1: 109487668-109487668 on Build GRCh38
rs1000233611 Chr.1: 109487560-109487560 on Build GRCh38

View Full Product Details