Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109489849-109490354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L2
Entrez Gene ID
127002
Gene Name
ataxin 7 like 2
Gene Aliases
-
UniGene
Hs.118248
Gene Chromosome Location
Chr.1: 109483904-109492804 on Build GRCh38
Transcripts
NM_153340.4 XM_005270445.3 XM_005270446.2 XM_011540638.2 XM_011540639.2 XM_011540641.2 XM_011540644.2 XM_011540645.1 XM_011540646.2 XM_017000267.1 XM_017000268.1 XM_017000269.1 XM_017000270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
272379 c.1291C>T p.R431W Chr.1: 109490325-109490325 on Build GRCh38
3862121 c.1215C>T p.T405T Chr.1: 109490107-109490107 on Build GRCh38
5461902 c.1314A>G p.A438A Chr.1: 109490348-109490348 on Build GRCh38
237032 c.1038-3C>T p.? Chr.1: 109489927-109489927 on Build GRCh38
7313508 c.1307G>T p.C436F Chr.1: 109490341-109490341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138073319 Chr.1: 109489902-109489902 on Build GRCh38
rs532308242 Chr.1: 109489904-109489904 on Build GRCh38
rs758981436 Chr.1: 109489963-109489963 on Build GRCh38
rs377631182 Chr.1: 109490149-109490149 on Build GRCh38
rs760445527 Chr.1: 109490065-109490065 on Build GRCh38
rs200119345 Chr.1: 109490061-109490061 on Build GRCh38
rs747218321 Chr.1: 109489938-109489940 on Build GRCh38
rs777944125 Chr.1: 109489969-109489969 on Build GRCh38
rs757266918 Chr.1: 109490083-109490083 on Build GRCh38
rs750537784 Chr.1: 109490163-109490163 on Build GRCh38
rs763526138 Chr.1: 109489931-109489931 on Build GRCh38
rs780205589 Chr.1: 109489906-109489906 on Build GRCh38
rs990966113 Chr.1: 109490021-109490021 on Build GRCh38
rs369007438 Chr.1: 109490069-109490069 on Build GRCh38
rs372308681 Chr.1: 109490072-109490072 on Build GRCh38
rs772784423 Chr.1: 109490136-109490136 on Build GRCh38
rs747970785 Chr.1: 109490238-109490238 on Build GRCh38
rs747085636 Chr.1: 109489986-109489986 on Build GRCh38
rs781395725 Chr.1: 109490087-109490087 on Build GRCh38
rs776404767 Chr.1: 109490253-109490253 on Build GRCh38
rs202047303 Chr.1: 109489885-109489885 on Build GRCh38
rs961396530 Chr.1: 109489946-109489946 on Build GRCh38
rs762175688 Chr.1: 109490291-109490291 on Build GRCh38
rs547436670 Chr.1: 109490289-109490289 on Build GRCh38
rs746008233 Chr.1: 109490091-109490091 on Build GRCh38
rs753253386 Chr.1: 109489948-109489948 on Build GRCh38
rs959354565 Chr.1: 109490127-109490127 on Build GRCh38
rs982770757 Chr.1: 109489973-109489973 on Build GRCh38
rs779773400 Chr.1: 109490099-109490099 on Build GRCh38
rs773810500 Chr.1: 109490034-109490034 on Build GRCh38
rs915429975 Chr.1: 109490043-109490043 on Build GRCh38
rs772267997 Chr.1: 109489879-109489879 on Build GRCh38
rs202089921 Chr.1: 109489980-109489980 on Build GRCh38
rs373264806 Chr.1: 109490158-109490158 on Build GRCh38
rs774134380 Chr.1: 109490112-109490112 on Build GRCh38
rs746154752 Chr.1: 109490002-109490002 on Build GRCh38
rs781245120 Chr.1: 109489996-109489996 on Build GRCh38
rs747191156 Chr.1: 109489882-109489882 on Build GRCh38
rs767105035 Chr.1: 109490060-109490060 on Build GRCh38
rs746552397 Chr.1: 109490248-109490248 on Build GRCh38
rs771147831 Chr.1: 109489987-109489987 on Build GRCh38
rs772061389 Chr.1: 109490240-109490240 on Build GRCh38
rs934366750 Chr.1: 109490079-109490079 on Build GRCh38
rs748174280 Chr.1: 109490115-109490115 on Build GRCh38
rs1035435595 Chr.1: 109490221-109490221 on Build GRCh38
rs772552140 Chr.1: 109490245-109490245 on Build GRCh38
rs764294279 Chr.1: 109490267-109490267 on Build GRCh38
rs924737445 Chr.1: 109490207-109490207 on Build GRCh38
rs1003514979 Chr.1: 109490214-109490214 on Build GRCh38
rs780217919 Chr.1: 109490169-109490169 on Build GRCh38
rs185034916 Chr.1: 109490325-109490325 on Build GRCh38
rs766154966 Chr.1: 109490147-109490147 on Build GRCh38
rs776442128 Chr.1: 109489892-109489892 on Build GRCh38
rs928273132 Chr.1: 109490010-109490010 on Build GRCh38
rs762941184 Chr.1: 109489930-109489930 on Build GRCh38
rs182037261 Chr.1: 109490167-109490167 on Build GRCh38
rs771480212 Chr.1: 109490117-109490117 on Build GRCh38
rs900998726 Chr.1: 109489886-109489886 on Build GRCh38
rs769571017 Chr.1: 109490005-109490005 on Build GRCh38
rs760349926 Chr.1: 109490141-109490141 on Build GRCh38
rs778835491 Chr.1: 109490237-109490237 on Build GRCh38
rs570473924 Chr.1: 109490300-109490300 on Build GRCh38
rs773494067 Chr.1: 109489880-109489880 on Build GRCh38
rs1041670119 Chr.1: 109490047-109490047 on Build GRCh38
rs982442617 Chr.1: 109489871-109489871 on Build GRCh38
rs900074949 Chr.1: 109490278-109490278 on Build GRCh38
rs759325363 Chr.1: 109490254-109490254 on Build GRCh38
rs866777358 Chr.1: 109490082-109490082 on Build GRCh38
rs374247564 Chr.1: 109490148-109490148 on Build GRCh38
rs562739459 Chr.1: 109490018-109490018 on Build GRCh38
rs761478393 Chr.1: 109489934-109489934 on Build GRCh38
rs754754909 Chr.1: 109490225-109490225 on Build GRCh38
rs997388909 Chr.1: 109490063-109490063 on Build GRCh38
rs778459954 Chr.1: 109490081-109490081 on Build GRCh38
rs768722536 Chr.1: 109490145-109490145 on Build GRCh38
rs200569345 Chr.1: 109489905-109489905 on Build GRCh38
rs766504933 Chr.1: 109490326-109490326 on Build GRCh38
rs768601316 Chr.1: 109490019-109490019 on Build GRCh38
rs41279734 Chr.1: 109490109-109490109 on Build GRCh38
rs775390150 Chr.1: 109489921-109489921 on Build GRCh38
rs888292017 Chr.1: 109490114-109490114 on Build GRCh38
rs754056969 Chr.1: 109490333-109490333 on Build GRCh38
rs138449778 Chr.1: 109489932-109489932 on Build GRCh38
rs770610369 Chr.1: 109490249-109490249 on Build GRCh38
rs145225962 Chr.1: 109489935-109489935 on Build GRCh38
rs749560811 Chr.1: 109490176-109490176 on Build GRCh38
rs867647050 Chr.1: 109490004-109490005 on Build GRCh38
rs761196507 Chr.1: 109490042-109490042 on Build GRCh38
rs779480797 Chr.1: 109489945-109489945 on Build GRCh38
rs200126569 Chr.1: 109490159-109490159 on Build GRCh38
rs961362809 Chr.1: 109490184-109490184 on Build GRCh38
rs765633159 Chr.1: 109490071-109490071 on Build GRCh38
rs768099100 Chr.1: 109490308-109490308 on Build GRCh38
rs369361561 Chr.1: 109490152-109490152 on Build GRCh38
rs377314546 Chr.1: 109490097-109490097 on Build GRCh38
rs901055605 Chr.1: 109490096-109490096 on Build GRCh38
rs866346797 Chr.1: 109490031-109490031 on Build GRCh38
rs751808036 Chr.1: 109489940-109489940 on Build GRCh38
rs749111871 Chr.1: 109490108-109490108 on Build GRCh38
rs201175394 Chr.1: 109489954-109489954 on Build GRCh38

View Full Product Details