Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109497575-109498083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561D1
Entrez Gene ID
284613
Gene Name
cytochrome b561 family member D1
Gene Aliases
-
UniGene
Hs.514682
Gene Chromosome Location
Chr.1: 109492830-109500441 on Build GRCh38
Transcripts
NM_001134400.1 NM_001134402.1 NM_001134403.1 NM_001134404.1 NM_182580.2 XM_005270775.2 XM_005270776.3 XM_005270777.2 XM_011541286.2 XM_011541287.2 XM_017001078.1 XM_017001079.1

SNP Detail

SNP ID Chromosome Location
rs753116905 Chr.1: 109497983-109497983 on Build GRCh38
rs923835320 Chr.1: 109497668-109497668 on Build GRCh38
rs911280315 Chr.1: 109497989-109497989 on Build GRCh38
rs918564430 Chr.1: 109498025-109498025 on Build GRCh38
rs775176746 Chr.1: 109497834-109497839 on Build GRCh38
rs992224957 Chr.1: 109497795-109497795 on Build GRCh38
rs556578053 Chr.1: 109497962-109497962 on Build GRCh38
rs935264019 Chr.1: 109497669-109497669 on Build GRCh38
rs953182543 Chr.1: 109497980-109497980 on Build GRCh38
rs979158816 Chr.1: 109497664-109497664 on Build GRCh38
rs1000924584 Chr.1: 109497953-109497953 on Build GRCh38
rs925518506 Chr.1: 109497905-109497905 on Build GRCh38
rs112813866 Chr.1: 109497627-109497627 on Build GRCh38
rs200895907 Chr.1: 109497838-109497838 on Build GRCh38
rs755561289 Chr.1: 109497955-109497955 on Build GRCh38
rs1038876588 Chr.1: 109498003-109498003 on Build GRCh38
rs889542559 Chr.1: 109497972-109497972 on Build GRCh38
rs1035253229 Chr.1: 109497868-109497868 on Build GRCh38
rs186135695 Chr.1: 109497836-109497836 on Build GRCh38
rs948110502 Chr.1: 109497879-109497879 on Build GRCh38
rs897400986 Chr.1: 109497856-109497856 on Build GRCh38
rs549946005 Chr.1: 109497875-109497875 on Build GRCh38
rs751647221 Chr.1: 109497936-109497936 on Build GRCh38
rs759074533 Chr.1: 109497733-109497733 on Build GRCh38
rs764935611 Chr.1: 109497642-109497642 on Build GRCh38
rs986022870 Chr.1: 109497988-109497988 on Build GRCh38
rs1038465247 Chr.1: 109497984-109497984 on Build GRCh38
rs994077207 Chr.1: 109497999-109497999 on Build GRCh38
rs546236120 Chr.1: 109497831-109497836 on Build GRCh38
rs960767137 Chr.1: 109497791-109497791 on Build GRCh38
rs1038397960 Chr.1: 109497851-109497851 on Build GRCh38
rs1023798384 Chr.1: 109497935-109497936 on Build GRCh38
rs929982191 Chr.1: 109498050-109498050 on Build GRCh38
rs2297797 Chr.1: 109498058-109498058 on Build GRCh38
rs756655457 Chr.1: 109497830-109497831 on Build GRCh38
rs994352043 Chr.1: 109497863-109497863 on Build GRCh38
rs765657525 Chr.1: 109498036-109498036 on Build GRCh38
rs139074238 Chr.1: 109497615-109497615 on Build GRCh38
rs386634480 Chr.1: 109497832-109497838 on Build GRCh38
rs190265131 Chr.1: 109497837-109497837 on Build GRCh38
rs756571932 Chr.1: 109497993-109497993 on Build GRCh38
rs979847226 Chr.1: 109497881-109497881 on Build GRCh38
rs1012424865 Chr.1: 109497925-109497925 on Build GRCh38
rs761706943 Chr.1: 109497641-109497641 on Build GRCh38
rs1006520443 Chr.1: 109497978-109497978 on Build GRCh38
rs931804099 Chr.1: 109497756-109497756 on Build GRCh38
rs6666119 Chr.1: 109497688-109497688 on Build GRCh38
rs933726593 Chr.1: 109497959-109497959 on Build GRCh38
rs535742462 Chr.1: 109498026-109498026 on Build GRCh38
rs751202420 Chr.1: 109497811-109497811 on Build GRCh38
rs1007212376 Chr.1: 109497644-109497644 on Build GRCh38
rs532889442 Chr.1: 109497790-109497790 on Build GRCh38
rs935599945 Chr.1: 109497935-109497935 on Build GRCh38
rs149887195 Chr.1: 109497772-109497772 on Build GRCh38
rs3215981 Chr.1: 109497834-109497838 on Build GRCh38
rs150750883 Chr.1: 109497606-109497606 on Build GRCh38
rs2297796 Chr.1: 109497833-109497833 on Build GRCh38
rs902040266 Chr.1: 109497946-109497946 on Build GRCh38
rs11398789 Chr.1: 109497838-109497839 on Build GRCh38
rs2297795 Chr.1: 109497832-109497832 on Build GRCh38
rs7412657 Chr.1: 109497726-109497726 on Build GRCh38
rs962952227 Chr.1: 109497724-109497724 on Build GRCh38
rs750389442 Chr.1: 109497831-109497832 on Build GRCh38
rs147209297 Chr.1: 109497832-109497836 on Build GRCh38
rs866484618 Chr.1: 109497838-109497838 on Build GRCh38

View Full Product Details