Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109499217-109499711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561D1
Entrez Gene ID
284613
Gene Name
cytochrome b561 family member D1
Gene Aliases
-
UniGene
Hs.514682
Gene Chromosome Location
Chr.1: 109492830-109500441 on Build GRCh38
Transcripts
NM_001134400.1 NM_001134402.1 NM_001134403.1 NM_001134404.1 NM_182580.2 XM_005270775.2 XM_005270776.3 XM_005270777.2 XM_011541286.2 XM_011541287.2 XM_017001078.1 XM_017001079.1

SNP Detail

SNP ID Chromosome Location
rs911756316 Chr.1: 109499600-109499600 on Build GRCh38
rs905709849 Chr.1: 109499438-109499438 on Build GRCh38
rs958044388 Chr.1: 109499261-109499261 on Build GRCh38
rs551064469 Chr.1: 109499690-109499690 on Build GRCh38
rs913732449 Chr.1: 109499404-109499404 on Build GRCh38
rs558805407 Chr.1: 109499374-109499375 on Build GRCh38
rs886695513 Chr.1: 109499511-109499511 on Build GRCh38
rs920583125 Chr.1: 109499633-109499633 on Build GRCh38
rs980818381 Chr.1: 109499398-109499398 on Build GRCh38
rs913778554 Chr.1: 109499593-109499593 on Build GRCh38
rs928980446 Chr.1: 109499387-109499387 on Build GRCh38
rs1007931767 Chr.1: 109499262-109499262 on Build GRCh38
rs577221281 Chr.1: 109499336-109499336 on Build GRCh38
rs926338938 Chr.1: 109499399-109499399 on Build GRCh38
rs1026421565 Chr.1: 109499332-109499332 on Build GRCh38
rs555298675 Chr.1: 109499249-109499249 on Build GRCh38
rs974893573 Chr.1: 109499632-109499632 on Build GRCh38
rs773160501 Chr.1: 109499589-109499589 on Build GRCh38
rs2091329 Chr.1: 109499457-109499457 on Build GRCh38
rs1032430302 Chr.1: 109499450-109499450 on Build GRCh38
rs950924934 Chr.1: 109499334-109499334 on Build GRCh38
rs541007400 Chr.1: 109499287-109499287 on Build GRCh38
rs559737185 Chr.1: 109499489-109499489 on Build GRCh38
rs1002676971 Chr.1: 109499449-109499449 on Build GRCh38
rs552109366 Chr.1: 109499574-109499574 on Build GRCh38
rs911953651 Chr.1: 109499463-109499463 on Build GRCh38
rs943428847 Chr.1: 109499605-109499605 on Build GRCh38
rs958178142 Chr.1: 109499402-109499402 on Build GRCh38
rs989439289 Chr.1: 109499403-109499403 on Build GRCh38
rs954853535 Chr.1: 109499286-109499286 on Build GRCh38
rs1001984381 Chr.1: 109499578-109499578 on Build GRCh38
rs1033762734 Chr.1: 109499579-109499579 on Build GRCh38
rs899187439 Chr.1: 109499487-109499487 on Build GRCh38
rs537460107 Chr.1: 109499660-109499660 on Build GRCh38
rs373208509 Chr.1: 109499513-109499513 on Build GRCh38
rs928972225 Chr.1: 109499612-109499612 on Build GRCh38
rs1032729973 Chr.1: 109499254-109499254 on Build GRCh38
rs957764210 Chr.1: 109499588-109499588 on Build GRCh38
rs966633495 Chr.1: 109499598-109499598 on Build GRCh38
rs746658161 Chr.1: 109499348-109499348 on Build GRCh38
rs543072788 Chr.1: 109499397-109499397 on Build GRCh38
rs181202160 Chr.1: 109499643-109499643 on Build GRCh38
rs7520465 Chr.1: 109499466-109499466 on Build GRCh38
rs917150400 Chr.1: 109499683-109499683 on Build GRCh38
rs563067151 Chr.1: 109499405-109499405 on Build GRCh38
rs1014352835 Chr.1: 109499517-109499517 on Build GRCh38
rs749134518 Chr.1: 109499551-109499551 on Build GRCh38
rs1047626727 Chr.1: 109499390-109499390 on Build GRCh38
rs530027640 Chr.1: 109499510-109499510 on Build GRCh38
rs184338046 Chr.1: 109499276-109499276 on Build GRCh38
rs569040124 Chr.1: 109499469-109499469 on Build GRCh38
rs1024642306 Chr.1: 109499337-109499337 on Build GRCh38
rs1039431563 Chr.1: 109499468-109499468 on Build GRCh38
rs1014023880 Chr.1: 109499401-109499401 on Build GRCh38
rs189595922 Chr.1: 109499320-109499320 on Build GRCh38
rs988353058 Chr.1: 109499266-109499266 on Build GRCh38
rs987272182 Chr.1: 109499599-109499599 on Build GRCh38

View Full Product Details