Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109592655-109593183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAI3
Entrez Gene ID
2773
Gene Name
G protein subunit alpha i3
Gene Aliases
87U6, ARCND1
UniGene
Hs.73799
Gene Chromosome Location
Chr.1: 109548564-109595843 on Build GRCh38
Transcripts
NM_006496.3

SNP Detail

SNP ID Chromosome Location
rs538350464 Chr.1: 109592909-109592909 on Build GRCh38
rs7371 Chr.1: 109592686-109592686 on Build GRCh38
rs964988308 Chr.1: 109592688-109592688 on Build GRCh38
rs765674270 Chr.1: 109592980-109592980 on Build GRCh38
rs1027563397 Chr.1: 109592693-109592693 on Build GRCh38
rs368914083 Chr.1: 109592706-109592706 on Build GRCh38
rs560234721 Chr.1: 109593073-109593073 on Build GRCh38
rs115879755 Chr.1: 109593120-109593120 on Build GRCh38
rs548519023 Chr.1: 109592872-109592872 on Build GRCh38
rs753660445 Chr.1: 109592819-109592819 on Build GRCh38
rs550461663 Chr.1: 109592945-109592945 on Build GRCh38
rs1028525982 Chr.1: 109592746-109592746 on Build GRCh38
rs548695540 Chr.1: 109592809-109592810 on Build GRCh38
rs191918287 Chr.1: 109592889-109592889 on Build GRCh38
rs996071637 Chr.1: 109592692-109592692 on Build GRCh38
rs530293115 Chr.1: 109592681-109592681 on Build GRCh38
rs750477671 Chr.1: 109593079-109593079 on Build GRCh38
rs927647369 Chr.1: 109592830-109592832 on Build GRCh38
rs552680463 Chr.1: 109592888-109592888 on Build GRCh38
rs780564640 Chr.1: 109593157-109593157 on Build GRCh38
rs1053554140 Chr.1: 109593082-109593082 on Build GRCh38
rs41280244 Chr.1: 109593127-109593127 on Build GRCh38
rs567456905 Chr.1: 109592965-109592965 on Build GRCh38
rs939288132 Chr.1: 109592867-109592867 on Build GRCh38
rs189839861 Chr.1: 109592762-109592762 on Build GRCh38
rs901161882 Chr.1: 109593111-109593111 on Build GRCh38
rs983892172 Chr.1: 109592763-109592763 on Build GRCh38
rs946525985 Chr.1: 109592949-109592949 on Build GRCh38
rs1028556782 Chr.1: 109593158-109593158 on Build GRCh38
rs981868123 Chr.1: 109592987-109592987 on Build GRCh38
rs998940459 Chr.1: 109593067-109593067 on Build GRCh38
rs904580735 Chr.1: 109593028-109593028 on Build GRCh38
rs148596284 Chr.1: 109592820-109592820 on Build GRCh38
rs75351728 Chr.1: 109592899-109592899 on Build GRCh38

View Full Product Details