Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109921759-109922269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF1
Entrez Gene ID
1435
Gene Name
colony stimulating factor 1
Gene Aliases
CSF-1, MCSF
UniGene
Hs.173894
Gene Chromosome Location
Chr.1: 109910506-109930994 on Build GRCh38
Transcripts
NM_000757.5 NM_172210.2 NM_172211.3 NM_172212.2 XM_017000369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5353462 c.403G>A p.V135I Chr.1: 109921853-109921853 on Build GRCh38
5752898 c.497T>G p.I166S Chr.1: 109921947-109921947 on Build GRCh38
4217619 c.486G>T p.K162N Chr.1: 109921936-109921936 on Build GRCh38
6036820 c.481G>T p.D161Y Chr.1: 109921931-109921931 on Build GRCh38
4217617 c.420G>A p.E140E Chr.1: 109921870-109921870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201377788 Chr.1: 109921931-109921931 on Build GRCh38
rs1054665639 Chr.1: 109922234-109922234 on Build GRCh38
rs377085953 Chr.1: 109921853-109921853 on Build GRCh38
rs775583014 Chr.1: 109921920-109921920 on Build GRCh38
rs576527357 Chr.1: 109921785-109921785 on Build GRCh38
rs906002771 Chr.1: 109921827-109921827 on Build GRCh38
rs745478521 Chr.1: 109921859-109921859 on Build GRCh38
rs760678409 Chr.1: 109921809-109921809 on Build GRCh38
rs895842550 Chr.1: 109921944-109921944 on Build GRCh38
rs12721517 Chr.1: 109922079-109922079 on Build GRCh38
rs147128516 Chr.1: 109921990-109921990 on Build GRCh38
rs765826628 Chr.1: 109922045-109922045 on Build GRCh38
rs769778625 Chr.1: 109922010-109922010 on Build GRCh38
rs760171661 Chr.1: 109922043-109922043 on Build GRCh38
rs780314082 Chr.1: 109922215-109922215 on Build GRCh38
rs765555629 Chr.1: 109922135-109922135 on Build GRCh38
rs199611804 Chr.1: 109921814-109921814 on Build GRCh38
rs911330149 Chr.1: 109922074-109922074 on Build GRCh38
rs753443931 Chr.1: 109921982-109921982 on Build GRCh38
rs779131069 Chr.1: 109921879-109921879 on Build GRCh38
rs778192732 Chr.1: 109921895-109921895 on Build GRCh38
rs747810307 Chr.1: 109922033-109922033 on Build GRCh38
rs775733987 Chr.1: 109922011-109922011 on Build GRCh38
rs759668151 Chr.1: 109921915-109921915 on Build GRCh38
rs755839156 Chr.1: 109921877-109921877 on Build GRCh38
rs144386894 Chr.1: 109922235-109922235 on Build GRCh38
rs770123506 Chr.1: 109921916-109921916 on Build GRCh38
rs919467136 Chr.1: 109922224-109922224 on Build GRCh38
rs763299983 Chr.1: 109922024-109922024 on Build GRCh38
rs748579696 Chr.1: 109921798-109921798 on Build GRCh38
rs768541401 Chr.1: 109922025-109922025 on Build GRCh38
rs556686021 Chr.1: 109921781-109921781 on Build GRCh38
rs530586578 Chr.1: 109922099-109922099 on Build GRCh38
rs747532766 Chr.1: 109921902-109921902 on Build GRCh38
rs200319483 Chr.1: 109922031-109922031 on Build GRCh38
rs1000616230 Chr.1: 109922052-109922052 on Build GRCh38
rs376516821 Chr.1: 109921807-109921807 on Build GRCh38
rs752598125 Chr.1: 109921825-109921825 on Build GRCh38
rs748611545 Chr.1: 109921882-109921882 on Build GRCh38
rs778137740 Chr.1: 109921985-109921985 on Build GRCh38
rs370213494 Chr.1: 109921857-109921857 on Build GRCh38
rs750624916 Chr.1: 109922200-109922200 on Build GRCh38
rs560880023 Chr.1: 109922116-109922116 on Build GRCh38
rs111936717 Chr.1: 109921883-109921883 on Build GRCh38
rs753445382 Chr.1: 109922046-109922046 on Build GRCh38
rs771489960 Chr.1: 109922002-109922002 on Build GRCh38
rs202069343 Chr.1: 109921812-109921812 on Build GRCh38
rs878976230 Chr.1: 109922159-109922159 on Build GRCh38
rs985516018 Chr.1: 109922063-109922063 on Build GRCh38
rs776825597 Chr.1: 109921911-109921911 on Build GRCh38
rs917553048 Chr.1: 109922154-109922154 on Build GRCh38
rs774336199 Chr.1: 109922030-109922030 on Build GRCh38
rs377701597 Chr.1: 109921858-109921858 on Build GRCh38
rs762780495 Chr.1: 109921924-109921924 on Build GRCh38
rs1056039415 Chr.1: 109922126-109922126 on Build GRCh38
rs947741661 Chr.1: 109922189-109922189 on Build GRCh38
rs776665315 Chr.1: 109921813-109921813 on Build GRCh38
rs878901328 Chr.1: 109922193-109922194 on Build GRCh38
rs751503599 Chr.1: 109921932-109921932 on Build GRCh38
rs372834853 Chr.1: 109921829-109921829 on Build GRCh38
rs758709724 Chr.1: 109921984-109921984 on Build GRCh38
rs767556584 Chr.1: 109922033-109922033 on Build GRCh38
rs772623563 Chr.1: 109921801-109921801 on Build GRCh38
rs973942154 Chr.1: 109922103-109922103 on Build GRCh38
rs200625266 Chr.1: 109921971-109921971 on Build GRCh38
rs74387109 Chr.1: 109921973-109921973 on Build GRCh38
rs761881593 Chr.1: 109921936-109921936 on Build GRCh38
rs1009477181 Chr.1: 109922107-109922107 on Build GRCh38
rs373767428 Chr.1: 109921840-109921840 on Build GRCh38
rs929950164 Chr.1: 109922113-109922113 on Build GRCh38
rs1000761284 Chr.1: 109922220-109922220 on Build GRCh38
rs546230702 Chr.1: 109922218-109922218 on Build GRCh38
rs181284402 Chr.1: 109922042-109922042 on Build GRCh38
rs770954483 Chr.1: 109921903-109921903 on Build GRCh38
rs543326949 Chr.1: 109922201-109922201 on Build GRCh38
rs766913690 Chr.1: 109921948-109921948 on Build GRCh38
rs544123610 Chr.1: 109922108-109922108 on Build GRCh38
rs149650587 Chr.1: 109922161-109922161 on Build GRCh38
rs778285294 Chr.1: 109922158-109922158 on Build GRCh38
rs1057152342 Chr.1: 109921852-109921852 on Build GRCh38
rs986211522 Chr.1: 109922199-109922199 on Build GRCh38
rs761509089 Chr.1: 109922115-109922115 on Build GRCh38
rs764058053 Chr.1: 109921830-109921830 on Build GRCh38
rs575859755 Chr.1: 109921960-109921960 on Build GRCh38
rs781130023 Chr.1: 109922008-109922008 on Build GRCh38

View Full Product Details