Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110012577-110013120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCYL1
Entrez Gene ID
10768
Gene Name
adenosylhomocysteinase like 1
Gene Aliases
DCAL, IRBIT, PPP1R78, PRO0233, XPVKONA
UniGene
Hs.743973
Gene Chromosome Location
Chr.1: 109984741-110023742 on Build GRCh38
Transcripts
NM_001242673.1 NM_001242674.1 NM_001242675.1 NM_001242676.1 NM_006621.5 XM_011540535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4020451 c.538T>C p.Y180H Chr.1: 110012957-110012957 on Build GRCh38
6019520 c.574G>A p.E192K Chr.1: 110012993-110012993 on Build GRCh38
5497557 c.495C>A p.L165L Chr.1: 110012914-110012914 on Build GRCh38
5453811 c.580+9G>A p.? Chr.1: 110013008-110013008 on Build GRCh38
7281033 c.502C>A p.L168M Chr.1: 110012921-110012921 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045788753 Chr.1: 110012745-110012745 on Build GRCh38
rs753114845 Chr.1: 110012755-110012755 on Build GRCh38
rs570092670 Chr.1: 110012682-110012682 on Build GRCh38
rs761835990 Chr.1: 110013010-110013010 on Build GRCh38
rs898726205 Chr.1: 110013053-110013053 on Build GRCh38
rs369037779 Chr.1: 110012886-110012886 on Build GRCh38
rs986830503 Chr.1: 110012849-110012849 on Build GRCh38
rs186704904 Chr.1: 110012747-110012747 on Build GRCh38
rs916715501 Chr.1: 110012752-110012776 on Build GRCh38
rs5777000 Chr.1: 110012612-110012613 on Build GRCh38
rs911326675 Chr.1: 110012612-110012612 on Build GRCh38
rs964096935 Chr.1: 110013047-110013047 on Build GRCh38
rs541477273 Chr.1: 110013085-110013085 on Build GRCh38
rs893348476 Chr.1: 110012791-110012791 on Build GRCh38
rs1020557773 Chr.1: 110012824-110012824 on Build GRCh38
rs758031053 Chr.1: 110012621-110012622 on Build GRCh38
rs1418320 Chr.1: 110012968-110012968 on Build GRCh38
rs767703467 Chr.1: 110012914-110012914 on Build GRCh38
rs1010395828 Chr.1: 110012803-110012803 on Build GRCh38
rs746463767 Chr.1: 110013017-110013017 on Build GRCh38
rs754695961 Chr.1: 110012956-110012956 on Build GRCh38
rs776116690 Chr.1: 110013033-110013033 on Build GRCh38
rs778815036 Chr.1: 110012962-110012962 on Build GRCh38
rs555697950 Chr.1: 110012740-110012740 on Build GRCh38
rs1047674184 Chr.1: 110012694-110012694 on Build GRCh38
rs766459959 Chr.1: 110012930-110012930 on Build GRCh38
rs748064006 Chr.1: 110012995-110012995 on Build GRCh38
rs376686971 Chr.1: 110013008-110013008 on Build GRCh38
rs576618569 Chr.1: 110012872-110012872 on Build GRCh38
rs556879731 Chr.1: 110012621-110012621 on Build GRCh38
rs867757997 Chr.1: 110012966-110012966 on Build GRCh38
rs920387442 Chr.1: 110012628-110012628 on Build GRCh38
rs951405000 Chr.1: 110012613-110012613 on Build GRCh38
rs774699717 Chr.1: 110012890-110012890 on Build GRCh38
rs202012516 Chr.1: 110012873-110012873 on Build GRCh38
rs562423332 Chr.1: 110012937-110012937 on Build GRCh38
rs930473086 Chr.1: 110012635-110012635 on Build GRCh38
rs138957901 Chr.1: 110012687-110012687 on Build GRCh38
rs756279164 Chr.1: 110012919-110012919 on Build GRCh38
rs1013434430 Chr.1: 110012613-110012614 on Build GRCh38
rs553732082 Chr.1: 110012836-110012836 on Build GRCh38
rs754117077 Chr.1: 110012936-110012936 on Build GRCh38
rs770504525 Chr.1: 110012852-110012852 on Build GRCh38
rs753928039 Chr.1: 110013009-110013010 on Build GRCh38
rs557218578 Chr.1: 110012833-110012833 on Build GRCh38
rs374682148 Chr.1: 110012781-110012781 on Build GRCh38
rs925210070 Chr.1: 110012715-110012715 on Build GRCh38
rs770459940 Chr.1: 110013023-110013023 on Build GRCh38
rs764285757 Chr.1: 110012912-110012913 on Build GRCh38
rs541470186 Chr.1: 110012790-110012790 on Build GRCh38
rs763187850 Chr.1: 110012877-110012877 on Build GRCh38
rs1001251389 Chr.1: 110012751-110012751 on Build GRCh38
rs768044571 Chr.1: 110012613-110012613 on Build GRCh38
rs995916456 Chr.1: 110013081-110013081 on Build GRCh38
rs1047064837 Chr.1: 110013091-110013091 on Build GRCh38
rs531475428 Chr.1: 110013007-110013007 on Build GRCh38
rs373297401 Chr.1: 110012892-110012892 on Build GRCh38
rs987800830 Chr.1: 110012748-110012748 on Build GRCh38
rs886345051 Chr.1: 110012708-110012708 on Build GRCh38
rs370928626 Chr.1: 110012622-110012622 on Build GRCh38
rs762470533 Chr.1: 110012908-110012908 on Build GRCh38

View Full Product Details