Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110043532-110044024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRIP1
Entrez Gene ID
85369
Gene Name
striatin interacting protein 1
Gene Aliases
FAM40A, FAR11A
UniGene
Hs.584996
Gene Chromosome Location
Chr.1: 110031577-110054641 on Build GRCh38
Transcripts
NM_001270768.1 NM_033088.3 NR_073071.1 XM_006710995.2 XM_017002629.1 XM_017002630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
376507 c.1174G>C p.E392Q Chr.1: 110043744-110043744 on Build GRCh38
4928164 c.1186T>C p.F396L Chr.1: 110043756-110043756 on Build GRCh38
5930526 c.1217C>T p.P406L Chr.1: 110043787-110043787 on Build GRCh38
3740528 c.1279A>C p.K427Q Chr.1: 110043849-110043849 on Build GRCh38
3730809 c.1168A>C p.S390R Chr.1: 110043738-110043738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774228410 Chr.1: 110043725-110043725 on Build GRCh38
rs78090069 Chr.1: 110043958-110043958 on Build GRCh38
rs761438696 Chr.1: 110043809-110043809 on Build GRCh38
rs199640956 Chr.1: 110043899-110043899 on Build GRCh38
rs539514986 Chr.1: 110044001-110044001 on Build GRCh38
rs375880292 Chr.1: 110043608-110043608 on Build GRCh38
rs374504597 Chr.1: 110043812-110043812 on Build GRCh38
rs934647001 Chr.1: 110043555-110043555 on Build GRCh38
rs182910788 Chr.1: 110043595-110043595 on Build GRCh38
rs758469732 Chr.1: 110043743-110043743 on Build GRCh38
rs142966740 Chr.1: 110043674-110043674 on Build GRCh38
rs376135440 Chr.1: 110043736-110043736 on Build GRCh38
rs775282645 Chr.1: 110043790-110043790 on Build GRCh38
rs201299982 Chr.1: 110043807-110043807 on Build GRCh38
rs754111230 Chr.1: 110043735-110043735 on Build GRCh38
rs753010532 Chr.1: 110043833-110043833 on Build GRCh38
rs765701364 Chr.1: 110043831-110043831 on Build GRCh38
rs763761224 Chr.1: 110043679-110043679 on Build GRCh38
rs768670475 Chr.1: 110043883-110043883 on Build GRCh38
rs1023430609 Chr.1: 110043986-110043986 on Build GRCh38
rs749167658 Chr.1: 110043791-110043791 on Build GRCh38
rs531611091 Chr.1: 110043970-110043970 on Build GRCh38
rs370219565 Chr.1: 110043623-110043623 on Build GRCh38
rs758844866 Chr.1: 110043839-110043839 on Build GRCh38
rs779505794 Chr.1: 110043596-110043596 on Build GRCh38
rs746162659 Chr.1: 110043868-110043868 on Build GRCh38
rs772186947 Chr.1: 110043603-110043603 on Build GRCh38
rs781251064 Chr.1: 110043864-110043864 on Build GRCh38
rs371636182 Chr.1: 110043782-110043782 on Build GRCh38
rs759401927 Chr.1: 110043647-110043647 on Build GRCh38
rs774469137 Chr.1: 110043886-110043886 on Build GRCh38
rs929199689 Chr.1: 110043996-110043996 on Build GRCh38
rs111920837 Chr.1: 110043723-110043723 on Build GRCh38
rs932720815 Chr.1: 110043564-110043564 on Build GRCh38
rs192921031 Chr.1: 110043943-110043943 on Build GRCh38
rs751602765 Chr.1: 110043844-110043844 on Build GRCh38
rs759804557 Chr.1: 110043829-110043829 on Build GRCh38
rs777764496 Chr.1: 110043752-110043752 on Build GRCh38
rs112456881 Chr.1: 110043821-110043821 on Build GRCh38
rs767775883 Chr.1: 110043717-110043719 on Build GRCh38
rs745622488 Chr.1: 110043762-110043762 on Build GRCh38
rs551981992 Chr.1: 110043558-110043558 on Build GRCh38
rs200373879 Chr.1: 110043666-110043666 on Build GRCh38
rs761495434 Chr.1: 110043727-110043727 on Build GRCh38
rs371099476 Chr.1: 110043796-110043796 on Build GRCh38
rs751115577 Chr.1: 110043702-110043702 on Build GRCh38
rs1037588559 Chr.1: 110043598-110043598 on Build GRCh38
rs775829115 Chr.1: 110044002-110044002 on Build GRCh38
rs781512083 Chr.1: 110043759-110043759 on Build GRCh38
rs771481016 Chr.1: 110043557-110043557 on Build GRCh38
rs888921676 Chr.1: 110043568-110043568 on Build GRCh38
rs773945942 Chr.1: 110043799-110043799 on Build GRCh38
rs759885612 Chr.1: 110043608-110043608 on Build GRCh38
rs747105130 Chr.1: 110043754-110043754 on Build GRCh38
rs754239456 Chr.1: 110043594-110043594 on Build GRCh38
rs922592099 Chr.1: 110043865-110043865 on Build GRCh38
rs772736493 Chr.1: 110043900-110043900 on Build GRCh38
rs146631809 Chr.1: 110043739-110043739 on Build GRCh38
rs1050250044 Chr.1: 110043566-110043566 on Build GRCh38
rs374128518 Chr.1: 110043716-110043716 on Build GRCh38
rs767550981 Chr.1: 110043947-110043947 on Build GRCh38
rs867431299 Chr.1: 110043760-110043760 on Build GRCh38
rs897764641 Chr.1: 110043969-110043969 on Build GRCh38
rs762351041 Chr.1: 110043678-110043678 on Build GRCh38
rs200485650 Chr.1: 110043768-110043768 on Build GRCh38
rs757426185 Chr.1: 110043755-110043755 on Build GRCh38
rs747637938 Chr.1: 110043897-110043897 on Build GRCh38
rs747299417 Chr.1: 110043610-110043610 on Build GRCh38
rs568299058 Chr.1: 110043874-110043874 on Build GRCh38
rs757371214 Chr.1: 110043857-110043857 on Build GRCh38
rs1049915270 Chr.1: 110043788-110043788 on Build GRCh38
rs756292131 Chr.1: 110043870-110043870 on Build GRCh38
rs932753397 Chr.1: 110043913-110043913 on Build GRCh38
rs768723577 Chr.1: 110043794-110043794 on Build GRCh38
rs753170429 Chr.1: 110043724-110043726 on Build GRCh38
rs772965127 Chr.1: 110043824-110043824 on Build GRCh38
rs771146778 Chr.1: 110043618-110043618 on Build GRCh38
rs910392704 Chr.1: 110043942-110043942 on Build GRCh38
rs756843599 Chr.1: 110043797-110043798 on Build GRCh38
rs753307430 Chr.1: 110043742-110043742 on Build GRCh38
rs199976658 Chr.1: 110043658-110043658 on Build GRCh38
rs985487818 Chr.1: 110043937-110043937 on Build GRCh38

View Full Product Details