Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57295418-57295927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH12
Entrez Gene ID
201625
Gene Name
dynein axonemal heavy chain 12
Gene Aliases
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
UniGene
Hs.201378
Gene Chromosome Location
Chr.3: 57293699-57545203 on Build GRCh38
Transcripts
NM_001291661.1 XM_011533466.1 XM_011533467.1 XM_011533468.1 XM_011533469.1 XM_017005861.1 XM_017005862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1424846 c.9021-2A>G p.? Chr.3: 57295594-57295594 on Build GRCh38
1424845 c.9074T>C p.I3025T Chr.3: 57295539-57295539 on Build GRCh38
3380665 c.9063G>A p.L3021L Chr.3: 57295550-57295550 on Build GRCh38
6511161 c.9063G>T p.L3021L Chr.3: 57295550-57295550 on Build GRCh38
5344530 c.9021-7G>A p.? Chr.3: 57295599-57295599 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1023241882 Chr.3: 57295762-57295762 on Build GRCh38
rs917049757 Chr.3: 57295647-57295647 on Build GRCh38
rs890828881 Chr.3: 57295688-57295688 on Build GRCh38
rs928543715 Chr.3: 57295795-57295795 on Build GRCh38
rs913876403 Chr.3: 57295833-57295833 on Build GRCh38
rs187725790 Chr.3: 57295656-57295656 on Build GRCh38
rs547555114 Chr.3: 57295547-57295547 on Build GRCh38
rs758023672 Chr.3: 57295679-57295679 on Build GRCh38
rs139180663 Chr.3: 57295445-57295445 on Build GRCh38
rs978406677 Chr.3: 57295576-57295576 on Build GRCh38
rs926976933 Chr.3: 57295825-57295825 on Build GRCh38
rs753465062 Chr.3: 57295560-57295561 on Build GRCh38
rs542465098 Chr.3: 57295457-57295457 on Build GRCh38
rs968444323 Chr.3: 57295804-57295804 on Build GRCh38
rs751529133 Chr.3: 57295484-57295484 on Build GRCh38
rs192498797 Chr.3: 57295829-57295829 on Build GRCh38
rs571248483 Chr.3: 57295515-57295515 on Build GRCh38
rs569466486 Chr.3: 57295691-57295691 on Build GRCh38
rs958874930 Chr.3: 57295834-57295834 on Build GRCh38
rs752296494 Chr.3: 57295627-57295627 on Build GRCh38
rs553402276 Chr.3: 57295723-57295723 on Build GRCh38
rs762366655 Chr.3: 57295842-57295842 on Build GRCh38
rs11542068 Chr.3: 57295588-57295588 on Build GRCh38
rs921473741 Chr.3: 57295895-57295895 on Build GRCh38
rs1010010628 Chr.3: 57295766-57295766 on Build GRCh38
rs185114251 Chr.3: 57295856-57295856 on Build GRCh38
rs1047112248 Chr.3: 57295694-57295694 on Build GRCh38
rs988523361 Chr.3: 57295501-57295501 on Build GRCh38
rs988159851 Chr.3: 57295818-57295818 on Build GRCh38
rs1030249642 Chr.3: 57295742-57295742 on Build GRCh38
rs1057395860 Chr.3: 57295625-57295625 on Build GRCh38
rs774785613 Chr.3: 57295460-57295460 on Build GRCh38
rs757136596 Chr.3: 57295495-57295495 on Build GRCh38
rs1015189082 Chr.3: 57295699-57295699 on Build GRCh38
rs536059912 Chr.3: 57295638-57295638 on Build GRCh38
rs899985028 Chr.3: 57295610-57295610 on Build GRCh38
rs397875697 Chr.3: 57295454-57295454 on Build GRCh38
rs1039698965 Chr.3: 57295704-57295704 on Build GRCh38
rs374138336 Chr.3: 57295599-57295599 on Build GRCh38
rs962446570 Chr.3: 57295750-57295750 on Build GRCh38
rs538213001 Chr.3: 57295812-57295812 on Build GRCh38
rs531299058 Chr.3: 57295503-57295503 on Build GRCh38
rs749406765 Chr.3: 57295602-57295602 on Build GRCh38
rs892557210 Chr.3: 57295517-57295517 on Build GRCh38
rs370865086 Chr.3: 57295544-57295544 on Build GRCh38
rs992725899 Chr.3: 57295847-57295847 on Build GRCh38
rs558288852 Chr.3: 57295768-57295769 on Build GRCh38
rs562449893 Chr.3: 57295482-57295482 on Build GRCh38
rs937267348 Chr.3: 57295602-57295603 on Build GRCh38
rs930070474 Chr.3: 57295667-57295667 on Build GRCh38
rs900120717 Chr.3: 57295717-57295720 on Build GRCh38
rs545620223 Chr.3: 57295752-57295752 on Build GRCh38
rs182586789 Chr.3: 57295513-57295513 on Build GRCh38
rs997533073 Chr.3: 57295653-57295653 on Build GRCh38
rs981799226 Chr.3: 57295788-57295788 on Build GRCh38
rs955903519 Chr.3: 57295810-57295810 on Build GRCh38
rs969914486 Chr.3: 57295787-57295787 on Build GRCh38

View Full Product Details