Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57296701-57297245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH12
Entrez Gene ID
201625
Gene Name
dynein axonemal heavy chain 12
Gene Aliases
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
UniGene
Hs.201378
Gene Chromosome Location
Chr.3: 57293699-57545203 on Build GRCh38
Transcripts
NM_001291661.1 XM_011533466.1 XM_011533467.1 XM_011533468.1 XM_011533469.1 XM_017005861.1 XM_017005862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5399866 c.8920G>A p.E2974K Chr.3: 57296855-57296855 on Build GRCh38
4949769 c.8791G>T p.D2931Y Chr.3: 57296984-57296984 on Build GRCh38
4450812 c.8791-1G>A p.? Chr.3: 57296985-57296985 on Build GRCh38
1047632 c.8838C>A p.F2946L Chr.3: 57296937-57296937 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs986849200 Chr.3: 57297146-57297146 on Build GRCh38
rs571907702 Chr.3: 57297045-57297045 on Build GRCh38
rs1004102466 Chr.3: 57297034-57297034 on Build GRCh38
rs376893671 Chr.3: 57296959-57296959 on Build GRCh38
rs894264865 Chr.3: 57296821-57296821 on Build GRCh38
rs760469862 Chr.3: 57296808-57296808 on Build GRCh38
rs542342832 Chr.3: 57297170-57297170 on Build GRCh38
rs972892502 Chr.3: 57296946-57296946 on Build GRCh38
rs752875015 Chr.3: 57296882-57296882 on Build GRCh38
rs560938300 Chr.3: 57297192-57297192 on Build GRCh38
rs1028574514 Chr.3: 57297111-57297111 on Build GRCh38
rs766033073 Chr.3: 57296809-57296809 on Build GRCh38
rs886744588 Chr.3: 57297029-57297029 on Build GRCh38
rs747451985 Chr.3: 57296958-57296958 on Build GRCh38
rs554216557 Chr.3: 57297180-57297180 on Build GRCh38
rs368920234 Chr.3: 57296817-57296817 on Build GRCh38
rs565189811 Chr.3: 57296944-57296944 on Build GRCh38
rs920059279 Chr.3: 57296952-57296952 on Build GRCh38
rs778492680 Chr.3: 57296911-57296911 on Build GRCh38
rs758539842 Chr.3: 57296886-57296886 on Build GRCh38
rs1019663783 Chr.3: 57296819-57296819 on Build GRCh38
rs979413270 Chr.3: 57297193-57297193 on Build GRCh38
rs1042922877 Chr.3: 57297131-57297131 on Build GRCh38
rs781160024 Chr.3: 57297220-57297220 on Build GRCh38
rs148298005 Chr.3: 57296814-57296814 on Build GRCh38
rs1849523 Chr.3: 57296969-57296969 on Build GRCh38
rs502442 Chr.3: 57297199-57297199 on Build GRCh38
rs904334716 Chr.3: 57297147-57297147 on Build GRCh38
rs544970814 Chr.3: 57296847-57296847 on Build GRCh38
rs955488442 Chr.3: 57297119-57297119 on Build GRCh38
rs779165568 Chr.3: 57296826-57296826 on Build GRCh38
rs1016827703 Chr.3: 57297080-57297080 on Build GRCh38
rs961403525 Chr.3: 57296892-57296892 on Build GRCh38
rs925590460 Chr.3: 57297198-57297198 on Build GRCh38
rs1029315594 Chr.3: 57297191-57297191 on Build GRCh38
rs781361874 Chr.3: 57297000-57297000 on Build GRCh38
rs372635632 Chr.3: 57296810-57296810 on Build GRCh38
rs141429638 Chr.3: 57297096-57297096 on Build GRCh38
rs201274393 Chr.3: 57296999-57296999 on Build GRCh38
rs975308376 Chr.3: 57297097-57297097 on Build GRCh38
rs945756695 Chr.3: 57297122-57297122 on Build GRCh38
rs1056786965 Chr.3: 57297101-57297101 on Build GRCh38
rs891556069 Chr.3: 57297115-57297115 on Build GRCh38
rs150647472 Chr.3: 57296805-57296806 on Build GRCh38
rs116214085 Chr.3: 57297211-57297211 on Build GRCh38
rs1028346720 Chr.3: 57296793-57296793 on Build GRCh38

View Full Product Details