Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57301545-57302044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH12
Entrez Gene ID
201625
Gene Name
dynein axonemal heavy chain 12
Gene Aliases
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
UniGene
Hs.201378
Gene Chromosome Location
Chr.3: 57293699-57545203 on Build GRCh38
Transcripts
NM_001291661.1 XM_011533466.1 XM_011533467.1 XM_011533468.1 XM_011533469.1 XM_017005860.1 XM_017005861.1 XM_017005862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7200085 c.8629G>T p.A2877S Chr.3: 57301896-57301896 on Build GRCh38
3824449 c.8693G>A p.G2898E Chr.3: 57301832-57301832 on Build GRCh38
4944086 c.8644G>T p.V2882F Chr.3: 57301881-57301881 on Build GRCh38
3824448 c.8778G>T p.L2926F Chr.3: 57301747-57301747 on Build GRCh38
3714601 c.8772C>A p.A2924A Chr.3: 57301753-57301753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774029444 Chr.3: 57301699-57301699 on Build GRCh38
rs374472466 Chr.3: 57301932-57301932 on Build GRCh38
rs562734656 Chr.3: 57301805-57301805 on Build GRCh38
rs765197537 Chr.3: 57301711-57301711 on Build GRCh38
rs768228450 Chr.3: 57301696-57301696 on Build GRCh38
rs757947796 Chr.3: 57301719-57301720 on Build GRCh38
rs757664944 Chr.3: 57301922-57301922 on Build GRCh38
rs757030626 Chr.3: 57301682-57301691 on Build GRCh38
rs692903 Chr.3: 57301989-57301989 on Build GRCh38
rs753616247 Chr.3: 57301827-57301827 on Build GRCh38
rs3223090 Chr.3: 57301679-57301680 on Build GRCh38
rs113952788 Chr.3: 57301589-57301765 on Build GRCh38
rs113518450 Chr.3: 57301910-57301910 on Build GRCh38
rs1035932981 Chr.3: 57301717-57301727 on Build GRCh38
rs982717898 Chr.3: 57301792-57301792 on Build GRCh38
rs374403630 Chr.3: 57301770-57301770 on Build GRCh38
rs771563572 Chr.3: 57301715-57301715 on Build GRCh38
rs749000069 Chr.3: 57301688-57301688 on Build GRCh38
rs994398233 Chr.3: 57301990-57301990 on Build GRCh38
rs1012425303 Chr.3: 57301591-57301591 on Build GRCh38
rs955496197 Chr.3: 57301764-57301764 on Build GRCh38
rs878863136 Chr.3: 57301678-57301685 on Build GRCh38
rs67229457 Chr.3: 57301678-57301687 on Build GRCh38
rs878876985 Chr.3: 57301678-57301683 on Build GRCh38
rs760905563 Chr.3: 57301684-57301691 on Build GRCh38
rs960274244 Chr.3: 57301725-57301725 on Build GRCh38
rs200860193 Chr.3: 57301999-57302001 on Build GRCh38
rs768193668 Chr.3: 57301849-57301849 on Build GRCh38
rs533042711 Chr.3: 57301785-57301785 on Build GRCh38
rs36020852 Chr.3: 57301864-57301864 on Build GRCh38
rs1052952678 Chr.3: 57301592-57301592 on Build GRCh38
rs4060726 Chr.3: 57301848-57301848 on Build GRCh38
rs905945115 Chr.3: 57301926-57301926 on Build GRCh38
rs1001470958 Chr.3: 57301669-57301672 on Build GRCh38
rs974396803 Chr.3: 57301810-57301810 on Build GRCh38
rs4681970 Chr.3: 57301719-57301719 on Build GRCh38
rs746981485 Chr.3: 57301726-57301729 on Build GRCh38
rs537970750 Chr.3: 57301964-57301964 on Build GRCh38
rs963740015 Chr.3: 57301727-57301727 on Build GRCh38
rs200349291 Chr.3: 57301678-57301691 on Build GRCh38
rs62252001 Chr.3: 57301663-57301663 on Build GRCh38
rs866343301 Chr.3: 57301844-57301844 on Build GRCh38
rs116071948 Chr.3: 57301658-57301658 on Build GRCh38
rs956390342 Chr.3: 57301669-57301669 on Build GRCh38
rs947477052 Chr.3: 57301911-57301911 on Build GRCh38
rs1037668531 Chr.3: 57301815-57301815 on Build GRCh38
rs756484190 Chr.3: 57301958-57301958 on Build GRCh38
rs66545318 Chr.3: 57301715-57301728 on Build GRCh38
rs57748737 Chr.3: 57301719-57301728 on Build GRCh38
rs745418145 Chr.3: 57301715-57301727 on Build GRCh38
rs922427166 Chr.3: 57301751-57301751 on Build GRCh38
rs761367228 Chr.3: 57301708-57301708 on Build GRCh38
rs747562232 Chr.3: 57301615-57301615 on Build GRCh38
rs1004234895 Chr.3: 57301903-57301903 on Build GRCh38
rs906694979 Chr.3: 57301892-57301892 on Build GRCh38
rs984530249 Chr.3: 57301754-57301754 on Build GRCh38
rs1010954391 Chr.3: 57301707-57301707 on Build GRCh38
rs760559757 Chr.3: 57301741-57301741 on Build GRCh38
rs1043170149 Chr.3: 57301918-57301918 on Build GRCh38
rs190402768 Chr.3: 57301834-57301834 on Build GRCh38
rs71919228 Chr.3: 57301678-57301689 on Build GRCh38
rs184571375 Chr.3: 57301668-57301668 on Build GRCh38
rs940711457 Chr.3: 57301806-57301806 on Build GRCh38
rs199748125 Chr.3: 57301771-57301771 on Build GRCh38
rs1057057805 Chr.3: 57301969-57301969 on Build GRCh38
rs199566484 Chr.3: 57301881-57301881 on Build GRCh38
rs746264010 Chr.3: 57301923-57301923 on Build GRCh38

View Full Product Details