Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57309175-57309717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH12
Entrez Gene ID
201625
Gene Name
dynein axonemal heavy chain 12
Gene Aliases
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
UniGene
Hs.201378
Gene Chromosome Location
Chr.3: 57293699-57545203 on Build GRCh38
Transcripts
NM_001291661.1 XM_011533466.1 XM_011533467.1 XM_011533468.1 XM_011533469.1 XM_017005860.1 XM_017005861.1 XM_017005862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6584362 c.8482-7C>G p.? Chr.3: 57309261-57309261 on Build GRCh38
5553207 c.8455G>T p.E2819* Chr.3: 57309692-57309692 on Build GRCh38
4755953 c.8485C>T p.P2829S Chr.3: 57309251-57309251 on Build GRCh38
4635039 c.8526T>G p.P2842P Chr.3: 57309210-57309210 on Build GRCh38
1047636 c.8496C>T p.F2832F Chr.3: 57309240-57309240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002500698 Chr.3: 57309393-57309393 on Build GRCh38
rs144781300 Chr.3: 57309288-57309288 on Build GRCh38
rs761904449 Chr.3: 57309590-57309590 on Build GRCh38
rs1001066226 Chr.3: 57309420-57309420 on Build GRCh38
rs1049773619 Chr.3: 57309227-57309227 on Build GRCh38
rs980321098 Chr.3: 57309255-57309255 on Build GRCh38
rs1054352355 Chr.3: 57309340-57309340 on Build GRCh38
rs1026755921 Chr.3: 57309537-57309537 on Build GRCh38
rs532300604 Chr.3: 57309240-57309240 on Build GRCh38
rs953877892 Chr.3: 57309514-57309516 on Build GRCh38
rs938586420 Chr.3: 57309265-57309265 on Build GRCh38
rs961276567 Chr.3: 57309431-57309431 on Build GRCh38
rs944010916 Chr.3: 57309264-57309264 on Build GRCh38
rs867029222 Chr.3: 57309455-57309455 on Build GRCh38
rs919637567 Chr.3: 57309201-57309201 on Build GRCh38
rs372818251 Chr.3: 57309546-57309546 on Build GRCh38
rs542569056 Chr.3: 57309486-57309486 on Build GRCh38
rs144270479 Chr.3: 57309516-57309516 on Build GRCh38
rs1001077417 Chr.3: 57309302-57309302 on Build GRCh38
rs1050338079 Chr.3: 57309434-57309434 on Build GRCh38
rs1042363639 Chr.3: 57309672-57309672 on Build GRCh38
rs932713396 Chr.3: 57309220-57309220 on Build GRCh38
rs533543458 Chr.3: 57309611-57309611 on Build GRCh38
rs912610463 Chr.3: 57309536-57309536 on Build GRCh38
rs763216841 Chr.3: 57309635-57309635 on Build GRCh38
rs764299331 Chr.3: 57309684-57309684 on Build GRCh38
rs767635317 Chr.3: 57309628-57309628 on Build GRCh38
rs376326862 Chr.3: 57309605-57309605 on Build GRCh38
rs912579378 Chr.3: 57309234-57309234 on Build GRCh38
rs1042622910 Chr.3: 57309283-57309283 on Build GRCh38
rs755881046 Chr.3: 57309551-57309554 on Build GRCh38
rs754979494 Chr.3: 57309531-57309531 on Build GRCh38
rs878962476 Chr.3: 57309230-57309230 on Build GRCh38
rs890076752 Chr.3: 57309468-57309468 on Build GRCh38
rs917911561 Chr.3: 57309355-57309355 on Build GRCh38
rs973961958 Chr.3: 57309500-57309500 on Build GRCh38
rs927254553 Chr.3: 57309263-57309263 on Build GRCh38
rs113232555 Chr.3: 57309509-57309509 on Build GRCh38
rs1022685944 Chr.3: 57309370-57309370 on Build GRCh38
rs552325385 Chr.3: 57309271-57309271 on Build GRCh38
rs776618725 Chr.3: 57309221-57309221 on Build GRCh38
rs760958048 Chr.3: 57309675-57309675 on Build GRCh38
rs1008482130 Chr.3: 57309507-57309507 on Build GRCh38
rs994210908 Chr.3: 57309530-57309530 on Build GRCh38
rs759316623 Chr.3: 57309225-57309225 on Build GRCh38
rs971060285 Chr.3: 57309384-57309384 on Build GRCh38
rs767037368 Chr.3: 57309239-57309239 on Build GRCh38
rs779744005 Chr.3: 57309396-57309396 on Build GRCh38
rs537975979 Chr.3: 57309369-57309369 on Build GRCh38
rs924226825 Chr.3: 57309676-57309676 on Build GRCh38
rs773245864 Chr.3: 57309632-57309635 on Build GRCh38
rs892910481 Chr.3: 57309351-57309351 on Build GRCh38
rs1042483969 Chr.3: 57309389-57309389 on Build GRCh38
rs189224063 Chr.3: 57309522-57309522 on Build GRCh38

View Full Product Details