Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57314396-57314943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH12
Entrez Gene ID
201625
Gene Name
dynein axonemal heavy chain 12
Gene Aliases
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
UniGene
Hs.201378
Gene Chromosome Location
Chr.3: 57293699-57545203 on Build GRCh38
Transcripts
NM_001291661.1 XM_011533466.1 XM_011533467.1 XM_011533468.1 XM_011533469.1 XM_017005860.1 XM_017005861.1 XM_017005862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2782019 c.8035C>T p.R2679C Chr.3: 57314517-57314517 on Build GRCh38
7278583 c.8042G>A p.S2681N Chr.3: 57314510-57314510 on Build GRCh38
6270706 c.7922C>A p.A2641D Chr.3: 57314630-57314630 on Build GRCh38
4755954 c.8036G>A p.R2679H Chr.3: 57314516-57314516 on Build GRCh38
3380666 c.7996G>T p.G2666C Chr.3: 57314556-57314556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182875567 Chr.3: 57314841-57314841 on Build GRCh38
rs749169478 Chr.3: 57314680-57314680 on Build GRCh38
rs1041842974 Chr.3: 57314771-57314771 on Build GRCh38
rs541980384 Chr.3: 57314723-57314723 on Build GRCh38
rs749999091 Chr.3: 57314641-57314641 on Build GRCh38
rs945833644 Chr.3: 57314759-57314760 on Build GRCh38
rs913678154 Chr.3: 57314736-57314736 on Build GRCh38
rs760809223 Chr.3: 57314566-57314566 on Build GRCh38
rs907615912 Chr.3: 57314651-57314651 on Build GRCh38
rs1051606001 Chr.3: 57314716-57314716 on Build GRCh38
rs968749921 Chr.3: 57314458-57314458 on Build GRCh38
rs932440780 Chr.3: 57314875-57314875 on Build GRCh38
rs1024495832 Chr.3: 57314541-57314541 on Build GRCh38
rs544105396 Chr.3: 57314903-57314903 on Build GRCh38
rs9852641 Chr.3: 57314880-57314880 on Build GRCh38
rs912446036 Chr.3: 57314877-57314877 on Build GRCh38
rs993019067 Chr.3: 57314517-57314517 on Build GRCh38
rs750504638 Chr.3: 57314527-57314527 on Build GRCh38
rs1034632266 Chr.3: 57314469-57314469 on Build GRCh38
rs373678191 Chr.3: 57314746-57314746 on Build GRCh38
rs979300420 Chr.3: 57314759-57314759 on Build GRCh38
rs894457119 Chr.3: 57314835-57314835 on Build GRCh38
rs190681757 Chr.3: 57314713-57314713 on Build GRCh38
rs921103523 Chr.3: 57314784-57314784 on Build GRCh38
rs771307646 Chr.3: 57314657-57314658 on Build GRCh38
rs571087663 Chr.3: 57314876-57314876 on Build GRCh38
rs577361356 Chr.3: 57314461-57314461 on Build GRCh38
rs951601073 Chr.3: 57314557-57314557 on Build GRCh38
rs754137662 Chr.3: 57314895-57314895 on Build GRCh38
rs890304424 Chr.3: 57314717-57314717 on Build GRCh38
rs941740603 Chr.3: 57314652-57314654 on Build GRCh38
rs146130771 Chr.3: 57314443-57314443 on Build GRCh38
rs868283922 Chr.3: 57314590-57314590 on Build GRCh38
rs766934256 Chr.3: 57314497-57314497 on Build GRCh38
rs776300939 Chr.3: 57314448-57314451 on Build GRCh38
rs921876734 Chr.3: 57314702-57314702 on Build GRCh38
rs150487595 Chr.3: 57314658-57314658 on Build GRCh38
rs904596511 Chr.3: 57314788-57314788 on Build GRCh38
rs765530452 Chr.3: 57314542-57314542 on Build GRCh38
rs780676295 Chr.3: 57314606-57314606 on Build GRCh38
rs754339301 Chr.3: 57314504-57314504 on Build GRCh38
rs867977856 Chr.3: 57314468-57314468 on Build GRCh38
rs960420445 Chr.3: 57314677-57314677 on Build GRCh38
rs751190724 Chr.3: 57314573-57314573 on Build GRCh38
rs867873899 Chr.3: 57314516-57314516 on Build GRCh38
rs553915190 Chr.3: 57314589-57314589 on Build GRCh38
rs539543089 Chr.3: 57314528-57314528 on Build GRCh38
rs1000224130 Chr.3: 57314795-57314795 on Build GRCh38
rs757359364 Chr.3: 57314655-57314657 on Build GRCh38
rs140052672 Chr.3: 57314670-57314670 on Build GRCh38
rs759741120 Chr.3: 57314650-57314650 on Build GRCh38
rs188330730 Chr.3: 57314450-57314450 on Build GRCh38
rs1010335277 Chr.3: 57314433-57314433 on Build GRCh38
rs112181197 Chr.3: 57314430-57314430 on Build GRCh38
rs1002418034 Chr.3: 57314669-57314669 on Build GRCh38
rs1034330152 Chr.3: 57314803-57314803 on Build GRCh38

View Full Product Details