Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57583523-57584037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF4
Entrez Gene ID
378
Gene Name
ADP ribosylation factor 4
Gene Aliases
ARF2
UniGene
Hs.652183
Gene Chromosome Location
Chr.3: 57571363-57597488 on Build GRCh38
Transcripts
NM_001660.3 NM_001322176.1

Gene Symbol
PDE12
Entrez Gene ID
201626
Gene Name
phosphodiesterase 12
Gene Aliases
2'-PDE, 2-PDE
UniGene
Hs.572993
Gene Chromosome Location
Chr.3: 57556247-57656480 on Build GRCh38
Transcripts
NM_001660.3 NM_001322176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
234405 c.189C>T p.F63F Chr.3: 57583967-57583967 on Build GRCh38
6424566 c.197G>T p.W66L Chr.3: 57583959-57583959 on Build GRCh38
1183327 c.232T>G p.W78G Chr.3: 57583924-57583924 on Build GRCh38
6331940 c.199G>A p.D67N Chr.3: 57583957-57583957 on Build GRCh38
98335 c.181A>G p.I61V Chr.3: 57583975-57583975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000816920 Chr.3: 57583799-57583799 on Build GRCh38
rs926819954 Chr.3: 57583718-57583718 on Build GRCh38
rs888282956 Chr.3: 57583828-57583828 on Build GRCh38
rs1018225468 Chr.3: 57583857-57583857 on Build GRCh38
rs111830807 Chr.3: 57583790-57583790 on Build GRCh38
rs1049294682 Chr.3: 57583699-57583699 on Build GRCh38
rs536425087 Chr.3: 57583776-57583776 on Build GRCh38
rs771477934 Chr.3: 57583550-57583550 on Build GRCh38
rs938139193 Chr.3: 57583724-57583724 on Build GRCh38
rs538643023 Chr.3: 57584003-57584003 on Build GRCh38
rs571408445 Chr.3: 57583716-57583716 on Build GRCh38
rs905186867 Chr.3: 57583778-57583778 on Build GRCh38
rs562816707 Chr.3: 57583583-57583583 on Build GRCh38
rs748867312 Chr.3: 57583667-57583667 on Build GRCh38
rs779360434 Chr.3: 57583976-57583976 on Build GRCh38
rs993557084 Chr.3: 57583811-57583811 on Build GRCh38
rs567382886 Chr.3: 57583753-57583753 on Build GRCh38
rs62260504 Chr.3: 57583605-57583605 on Build GRCh38
rs534078069 Chr.3: 57583727-57583727 on Build GRCh38
rs372202011 Chr.3: 57583885-57583885 on Build GRCh38
rs748613969 Chr.3: 57583981-57583981 on Build GRCh38
rs755555652 Chr.3: 57583925-57583925 on Build GRCh38
rs867807759 Chr.3: 57583673-57583673 on Build GRCh38
rs911943887 Chr.3: 57583602-57583602 on Build GRCh38
rs761459315 Chr.3: 57583870-57583870 on Build GRCh38
rs141747522 Chr.3: 57583693-57583693 on Build GRCh38
rs150527751 Chr.3: 57583695-57583695 on Build GRCh38
rs1019432793 Chr.3: 57583837-57583837 on Build GRCh38
rs77771120 Chr.3: 57583728-57583728 on Build GRCh38
rs199895622 Chr.3: 57583853-57583853 on Build GRCh38
rs1028622007 Chr.3: 57583804-57583804 on Build GRCh38
rs953923584 Chr.3: 57583816-57583816 on Build GRCh38
rs375206081 Chr.3: 57583694-57583694 on Build GRCh38
rs1056935005 Chr.3: 57583798-57583798 on Build GRCh38
rs61758690 Chr.3: 57583949-57583949 on Build GRCh38
rs867291321 Chr.3: 57583993-57583993 on Build GRCh38
rs966033368 Chr.3: 57583629-57583629 on Build GRCh38
rs772444015 Chr.3: 57583985-57583985 on Build GRCh38
rs753846757 Chr.3: 57583887-57583887 on Build GRCh38
rs972069250 Chr.3: 57583669-57583669 on Build GRCh38
rs750687844 Chr.3: 57583867-57583867 on Build GRCh38
rs11550597 Chr.3: 57583953-57583953 on Build GRCh38
rs963755675 Chr.3: 57583628-57583628 on Build GRCh38
rs745521800 Chr.3: 57584011-57584011 on Build GRCh38
rs1008450232 Chr.3: 57583820-57583820 on Build GRCh38
rs1009306363 Chr.3: 57583743-57583743 on Build GRCh38

View Full Product Details