Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58192434-58192963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1L3
Entrez Gene ID
1776
Gene Name
deoxyribonuclease I like 3
Gene Aliases
DHP2, DNAS1L3, LSD, SLEB16
UniGene
Hs.476453
Gene Chromosome Location
Chr.3: 58192626-58211003 on Build GRCh38
Transcripts
NM_001256560.1 NM_004944.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5399907 c.878C>T p.S293F Chr.3: 58192727-58192727 on Build GRCh38
3596852 c.903G>A p.K301K Chr.3: 58192702-58192702 on Build GRCh38
5511300 c.816C>T p.S272S Chr.3: 58192789-58192789 on Build GRCh38
1309414 c.870C>T p.S290S Chr.3: 58192735-58192735 on Build GRCh38
4678796 c.848C>A p.S283Y Chr.3: 58192757-58192757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374318062 Chr.3: 58192694-58192694 on Build GRCh38
rs948586401 Chr.3: 58192482-58192482 on Build GRCh38
rs1052916190 Chr.3: 58192887-58192887 on Build GRCh38
rs116685574 Chr.3: 58192524-58192524 on Build GRCh38
rs758356463 Chr.3: 58192667-58192667 on Build GRCh38
rs376419946 Chr.3: 58192699-58192699 on Build GRCh38
rs753708524 Chr.3: 58192829-58192829 on Build GRCh38
rs780196315 Chr.3: 58192695-58192695 on Build GRCh38
rs775093470 Chr.3: 58192779-58192779 on Build GRCh38
rs907155946 Chr.3: 58192513-58192513 on Build GRCh38
rs775684232 Chr.3: 58192739-58192740 on Build GRCh38
rs201385315 Chr.3: 58192626-58192626 on Build GRCh38
rs752086633 Chr.3: 58192653-58192653 on Build GRCh38
rs971263886 Chr.3: 58192460-58192460 on Build GRCh38
rs755019518 Chr.3: 58192840-58192840 on Build GRCh38
rs765888878 Chr.3: 58192685-58192685 on Build GRCh38
rs764180604 Chr.3: 58192790-58192790 on Build GRCh38
rs769227334 Chr.3: 58192725-58192725 on Build GRCh38
rs939776935 Chr.3: 58192503-58192503 on Build GRCh38
rs772885009 Chr.3: 58192622-58192622 on Build GRCh38
rs900168550 Chr.3: 58192600-58192600 on Build GRCh38
rs754904161 Chr.3: 58192900-58192900 on Build GRCh38
rs970045878 Chr.3: 58192745-58192745 on Build GRCh38
rs143481658 Chr.3: 58192943-58192943 on Build GRCh38
rs774817141 Chr.3: 58192729-58192729 on Build GRCh38
rs985566015 Chr.3: 58192910-58192910 on Build GRCh38
rs756365385 Chr.3: 58192819-58192819 on Build GRCh38
rs199826386 Chr.3: 58192834-58192834 on Build GRCh38
rs113005222 Chr.3: 58192802-58192802 on Build GRCh38
rs891809699 Chr.3: 58192683-58192683 on Build GRCh38
rs781607897 Chr.3: 58192686-58192686 on Build GRCh38
rs574759432 Chr.3: 58192486-58192486 on Build GRCh38
rs77435715 Chr.3: 58192521-58192521 on Build GRCh38
rs753396774 Chr.3: 58192862-58192862 on Build GRCh38
rs765144115 Chr.3: 58192901-58192901 on Build GRCh38
rs934590246 Chr.3: 58192869-58192869 on Build GRCh38
rs1042560741 Chr.3: 58192577-58192577 on Build GRCh38
rs891419323 Chr.3: 58192525-58192525 on Build GRCh38
rs369219413 Chr.3: 58192673-58192673 on Build GRCh38
rs915918565 Chr.3: 58192472-58192472 on Build GRCh38
rs780088738 Chr.3: 58192822-58192822 on Build GRCh38
rs760130295 Chr.3: 58192635-58192635 on Build GRCh38
rs746100795 Chr.3: 58192687-58192687 on Build GRCh38
rs997177562 Chr.3: 58192621-58192621 on Build GRCh38
rs762203578 Chr.3: 58192736-58192736 on Build GRCh38
rs1001015664 Chr.3: 58192588-58192588 on Build GRCh38
rs757098263 Chr.3: 58192678-58192678 on Build GRCh38
rs1032851070 Chr.3: 58192620-58192620 on Build GRCh38
rs751426201 Chr.3: 58192676-58192676 on Build GRCh38
rs571543105 Chr.3: 58192663-58192663 on Build GRCh38
rs750128475 Chr.3: 58192806-58192806 on Build GRCh38
rs1011282674 Chr.3: 58192541-58192541 on Build GRCh38
rs1017188764 Chr.3: 58192586-58192586 on Build GRCh38
rs762324739 Chr.3: 58192788-58192788 on Build GRCh38
rs544694171 Chr.3: 58192512-58192512 on Build GRCh38
rs772419715 Chr.3: 58192749-58192749 on Build GRCh38
rs779462251 Chr.3: 58192848-58192848 on Build GRCh38
rs748636732 Chr.3: 58192853-58192853 on Build GRCh38
rs759238109 Chr.3: 58192753-58192753 on Build GRCh38
rs1045645262 Chr.3: 58192492-58192492 on Build GRCh38
rs532308549 Chr.3: 58192659-58192659 on Build GRCh38
rs151161986 Chr.3: 58192751-58192751 on Build GRCh38
rs764725052 Chr.3: 58192765-58192765 on Build GRCh38
rs766060143 Chr.3: 58192801-58192801 on Build GRCh38
rs962750133 Chr.3: 58192693-58192693 on Build GRCh38
rs770717530 Chr.3: 58192651-58192651 on Build GRCh38
rs764788522 Chr.3: 58192638-58192638 on Build GRCh38
rs934496562 Chr.3: 58192515-58192515 on Build GRCh38
rs772767812 Chr.3: 58192462-58192462 on Build GRCh38

View Full Product Details