Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58570225-58570763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM107A
Entrez Gene ID
11170
Gene Name
family with sequence similarity 107 member A
Gene Aliases
DRR1, TU3A
UniGene
Hs.506357
Gene Chromosome Location
Chr.3: 58564112-58627610 on Build GRCh38
Transcripts
NM_001076778.2 NM_001282713.1 NM_001282714.1 NM_007177.3

SNP Detail

SNP ID Chromosome Location
rs1026607077 Chr.3: 58570266-58570266 on Build GRCh38
rs992792594 Chr.3: 58570657-58570657 on Build GRCh38
rs965119643 Chr.3: 58570346-58570346 on Build GRCh38
rs879112411 Chr.3: 58570698-58570698 on Build GRCh38
rs866944075 Chr.3: 58570631-58570631 on Build GRCh38
rs917254792 Chr.3: 58570671-58570671 on Build GRCh38
rs201156666 Chr.3: 58570637-58570637 on Build GRCh38
rs895778900 Chr.3: 58570301-58570301 on Build GRCh38
rs751165624 Chr.3: 58570586-58570605 on Build GRCh38
rs867778033 Chr.3: 58570617-58570617 on Build GRCh38
rs541083863 Chr.3: 58570574-58570574 on Build GRCh38
rs767742265 Chr.3: 58570501-58570501 on Build GRCh38
rs992845362 Chr.3: 58570465-58570465 on Build GRCh38
rs868491117 Chr.3: 58570619-58570619 on Build GRCh38
rs865973715 Chr.3: 58570613-58570613 on Build GRCh38
rs930861359 Chr.3: 58570524-58570524 on Build GRCh38
rs796635801 Chr.3: 58570408-58570409 on Build GRCh38
rs868458907 Chr.3: 58570428-58570428 on Build GRCh38
rs879758441 Chr.3: 58570660-58570661 on Build GRCh38
rs956370727 Chr.3: 58570484-58570484 on Build GRCh38
rs905147905 Chr.3: 58570250-58570250 on Build GRCh38
rs920757744 Chr.3: 58570487-58570487 on Build GRCh38
rs924587059 Chr.3: 58570429-58570429 on Build GRCh38
rs994681373 Chr.3: 58570349-58570349 on Build GRCh38
rs1003812672 Chr.3: 58570547-58570547 on Build GRCh38
rs979086298 Chr.3: 58570373-58570373 on Build GRCh38
rs915015245 Chr.3: 58570530-58570530 on Build GRCh38
rs377532163 Chr.3: 58570599-58570599 on Build GRCh38
rs1003862022 Chr.3: 58570280-58570280 on Build GRCh38
rs996555761 Chr.3: 58570615-58570615 on Build GRCh38
rs926411676 Chr.3: 58570659-58570659 on Build GRCh38
rs1044541769 Chr.3: 58570486-58570486 on Build GRCh38
rs115261732 Chr.3: 58570678-58570678 on Build GRCh38
rs917308399 Chr.3: 58570469-58570469 on Build GRCh38
rs1019302971 Chr.3: 58570289-58570289 on Build GRCh38
rs995497864 Chr.3: 58570263-58570263 on Build GRCh38
rs976991296 Chr.3: 58570463-58570463 on Build GRCh38
rs557599407 Chr.3: 58570584-58570599 on Build GRCh38
rs541487282 Chr.3: 58570480-58570480 on Build GRCh38
rs1028582690 Chr.3: 58570362-58570362 on Build GRCh38
rs765623852 Chr.3: 58570586-58570603 on Build GRCh38
rs147952091 Chr.3: 58570273-58570273 on Build GRCh38
rs750942727 Chr.3: 58570544-58570544 on Build GRCh38
rs552028723 Chr.3: 58570468-58570468 on Build GRCh38
rs975188120 Chr.3: 58570356-58570356 on Build GRCh38
rs780425641 Chr.3: 58570702-58570702 on Build GRCh38
rs980632484 Chr.3: 58570630-58570630 on Build GRCh38
rs980586008 Chr.3: 58570701-58570701 on Build GRCh38
rs141766186 Chr.3: 58570579-58570579 on Build GRCh38
rs1048432070 Chr.3: 58570528-58570528 on Build GRCh38
rs934637107 Chr.3: 58570446-58570446 on Build GRCh38
rs71091391 Chr.3: 58570636-58570637 on Build GRCh38
rs764387580 Chr.3: 58570361-58570361 on Build GRCh38
rs192325619 Chr.3: 58570367-58570367 on Build GRCh38
rs867392645 Chr.3: 58570625-58570625 on Build GRCh38
rs1040700634 Chr.3: 58570609-58570609 on Build GRCh38
rs542897672 Chr.3: 58570631-58570636 on Build GRCh38
rs777285782 Chr.3: 58570588-58570603 on Build GRCh38
rs955951637 Chr.3: 58570623-58570623 on Build GRCh38
rs371909507 Chr.3: 58570584-58570603 on Build GRCh38
rs5849262 Chr.3: 58570649-58570650 on Build GRCh38
rs867197875 Chr.3: 58570621-58570621 on Build GRCh38
rs762010607 Chr.3: 58570482-58570482 on Build GRCh38
rs1005698900 Chr.3: 58570391-58570391 on Build GRCh38
rs1018396882 Chr.3: 58570401-58570401 on Build GRCh38
rs1031989843 Chr.3: 58570622-58570622 on Build GRCh38
rs886767072 Chr.3: 58570269-58570269 on Build GRCh38
rs545936003 Chr.3: 58570265-58570265 on Build GRCh38
rs895818726 Chr.3: 58570679-58570679 on Build GRCh38
rs868324349 Chr.3: 58570627-58570627 on Build GRCh38

View Full Product Details