Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58645425-58645928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3D
Entrez Gene ID
131177
Gene Name
family with sequence similarity 3 member D
Gene Aliases
EF7, OIT1
UniGene
Hs.61265
Gene Chromosome Location
Chr.3: 58633943-58666843 on Build GRCh38
Transcripts
NM_138805.2 XM_005264862.4 XM_005264863.4 XM_005264864.4 XM_006712965.3 XM_006712967.3 XM_011533348.2 XM_011533349.2 XM_011533350.2 XM_011533351.2 XM_011533352.2 XM_011533353.2 XM_011533354.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1309418 c.146-1G>C p.? Chr.3: 58645627-58645627 on Build GRCh38
1251869 c.232G>A p.V78M Chr.3: 58645540-58645540 on Build GRCh38
4119693 c.261C>T p.R87R Chr.3: 58645511-58645511 on Build GRCh38
5601464 c.258C>T p.D86D Chr.3: 58645514-58645514 on Build GRCh38
1424986 c.243T>C p.T81T Chr.3: 58645529-58645529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753372371 Chr.3: 58645556-58645556 on Build GRCh38
rs200028657 Chr.3: 58645693-58645704 on Build GRCh38
rs143089006 Chr.3: 58645715-58645715 on Build GRCh38
rs75529659 Chr.3: 58645512-58645512 on Build GRCh38
rs944851047 Chr.3: 58645894-58645894 on Build GRCh38
rs778478340 Chr.3: 58645513-58645513 on Build GRCh38
rs763950619 Chr.3: 58645636-58645636 on Build GRCh38
rs755247033 Chr.3: 58645501-58645501 on Build GRCh38
rs1044898652 Chr.3: 58645829-58645829 on Build GRCh38
rs147876713 Chr.3: 58645607-58645607 on Build GRCh38
rs761663670 Chr.3: 58645544-58645544 on Build GRCh38
rs764917440 Chr.3: 58645676-58645676 on Build GRCh38
rs370131941 Chr.3: 58645616-58645616 on Build GRCh38
rs773856551 Chr.3: 58645479-58645481 on Build GRCh38
rs111621495 Chr.3: 58645457-58645458 on Build GRCh38
rs535746548 Chr.3: 58645668-58645668 on Build GRCh38
rs375479665 Chr.3: 58645759-58645759 on Build GRCh38
rs142021178 Chr.3: 58645549-58645549 on Build GRCh38
rs779347432 Chr.3: 58645502-58645502 on Build GRCh38
rs771080138 Chr.3: 58645528-58645528 on Build GRCh38
rs747547903 Chr.3: 58645585-58645585 on Build GRCh38
rs770054191 Chr.3: 58645603-58645603 on Build GRCh38
rs761515493 Chr.3: 58645822-58645822 on Build GRCh38
rs756988687 Chr.3: 58645678-58645688 on Build GRCh38
rs111951623 Chr.3: 58645765-58645765 on Build GRCh38
rs555583943 Chr.3: 58645696-58645696 on Build GRCh38
rs994146687 Chr.3: 58645739-58645739 on Build GRCh38
rs865911193 Chr.3: 58645716-58645716 on Build GRCh38
rs939316409 Chr.3: 58645865-58645865 on Build GRCh38
rs762838006 Chr.3: 58645755-58645755 on Build GRCh38
rs757238293 Chr.3: 58645587-58645587 on Build GRCh38
rs773094560 Chr.3: 58645618-58645618 on Build GRCh38
rs759273054 Chr.3: 58645480-58645481 on Build GRCh38
rs776211117 Chr.3: 58645557-58645557 on Build GRCh38
rs749932215 Chr.3: 58645473-58645473 on Build GRCh38
rs759070186 Chr.3: 58645572-58645572 on Build GRCh38
rs752155268 Chr.3: 58645536-58645536 on Build GRCh38
rs75207696 Chr.3: 58645451-58645451 on Build GRCh38
rs752433761 Chr.3: 58645678-58645678 on Build GRCh38
rs773184577 Chr.3: 58645553-58645553 on Build GRCh38
rs867687354 Chr.3: 58645723-58645723 on Build GRCh38
rs140725039 Chr.3: 58645584-58645584 on Build GRCh38
rs930745064 Chr.3: 58645532-58645532 on Build GRCh38
rs759025588 Chr.3: 58645675-58645675 on Build GRCh38
rs144069279 Chr.3: 58645485-58645485 on Build GRCh38
rs767777519 Chr.3: 58645838-58645838 on Build GRCh38
rs377609615 Chr.3: 58645555-58645555 on Build GRCh38
rs769339261 Chr.3: 58645592-58645592 on Build GRCh38
rs573206608 Chr.3: 58645655-58645655 on Build GRCh38
rs138971806 Chr.3: 58645811-58645811 on Build GRCh38
rs372504404 Chr.3: 58645748-58645748 on Build GRCh38
rs144804892 Chr.3: 58645541-58645541 on Build GRCh38
rs368973392 Chr.3: 58645540-58645540 on Build GRCh38
rs758879825 Chr.3: 58645578-58645578 on Build GRCh38
rs976280522 Chr.3: 58645904-58645904 on Build GRCh38
rs373299385 Chr.3: 58645495-58645495 on Build GRCh38
rs3217534 Chr.3: 58645482-58645483 on Build GRCh38
rs112971990 Chr.3: 58645466-58645470 on Build GRCh38
rs984247265 Chr.3: 58645523-58645523 on Build GRCh38
rs17059569 Chr.3: 58645594-58645594 on Build GRCh38
rs900792181 Chr.3: 58645582-58645582 on Build GRCh38
rs146148015 Chr.3: 58645740-58645740 on Build GRCh38
rs952748179 Chr.3: 58645747-58645747 on Build GRCh38
rs755679139 Chr.3: 58645487-58645487 on Build GRCh38
rs1044848925 Chr.3: 58645687-58645687 on Build GRCh38
rs540275482 Chr.3: 58645760-58645760 on Build GRCh38
rs377517424 Chr.3: 58645526-58645526 on Build GRCh38
rs374555282 Chr.3: 58645598-58645598 on Build GRCh38
rs926455658 Chr.3: 58645840-58645840 on Build GRCh38
rs766950758 Chr.3: 58645472-58645472 on Build GRCh38
rs374165284 Chr.3: 58645453-58645454 on Build GRCh38
rs375272324 Chr.3: 58645489-58645489 on Build GRCh38
rs148186316 Chr.3: 58645462-58645463 on Build GRCh38
rs753568345 Chr.3: 58645499-58645499 on Build GRCh38
rs904209347 Chr.3: 58645597-58645597 on Build GRCh38
rs915006181 Chr.3: 58645798-58645798 on Build GRCh38
rs71892228 Chr.3: 58645476-58645477 on Build GRCh38
rs753878891 Chr.3: 58645676-58645677 on Build GRCh38
rs116302699 Chr.3: 58645457-58645457 on Build GRCh38
rs1031393697 Chr.3: 58645602-58645602 on Build GRCh38
rs373820724 Chr.3: 58645674-58645674 on Build GRCh38
rs56017056 Chr.3: 58645484-58645485 on Build GRCh38
rs983907583 Chr.3: 58645784-58645784 on Build GRCh38
rs764863969 Chr.3: 58645571-58645571 on Build GRCh38
rs761757289 Chr.3: 58645466-58645466 on Build GRCh38
rs895796025 Chr.3: 58645719-58645719 on Build GRCh38
rs553311501 Chr.3: 58645610-58645610 on Build GRCh38
rs139892208 Chr.3: 58645486-58645486 on Build GRCh38
rs754664017 Chr.3: 58645563-58645563 on Build GRCh38
rs768279530 Chr.3: 58645542-58645542 on Build GRCh38
rs755636267 Chr.3: 58645624-58645626 on Build GRCh38
rs199788537 Chr.3: 58645543-58645543 on Build GRCh38
rs187056895 Chr.3: 58645593-58645593 on Build GRCh38

View Full Product Details