Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58666494-58666995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3D
Entrez Gene ID
131177
Gene Name
family with sequence similarity 3 member D
Gene Aliases
EF7, OIT1
UniGene
Hs.61265
Gene Chromosome Location
Chr.3: 58633943-58666843 on Build GRCh38
Transcripts
NM_138805.2 XM_005264863.4 XM_006712967.3 XM_011533348.2 XM_011533350.2 XM_011533351.2 XM_011533352.2

SNP Detail

SNP ID Chromosome Location
rs1036493271 Chr.3: 58666783-58666783 on Build GRCh38
rs974753642 Chr.3: 58666975-58666975 on Build GRCh38
rs73836302 Chr.3: 58666835-58666835 on Build GRCh38
rs981750636 Chr.3: 58666746-58666746 on Build GRCh38
rs960833791 Chr.3: 58666735-58666735 on Build GRCh38
rs948314143 Chr.3: 58666776-58666776 on Build GRCh38
rs374830669 Chr.3: 58666682-58666682 on Build GRCh38
rs570571938 Chr.3: 58666790-58666790 on Build GRCh38
rs530692105 Chr.3: 58666774-58666774 on Build GRCh38
rs550351812 Chr.3: 58666534-58666534 on Build GRCh38
rs924146016 Chr.3: 58666531-58666531 on Build GRCh38
rs978606924 Chr.3: 58666730-58666730 on Build GRCh38
rs916780937 Chr.3: 58666900-58666900 on Build GRCh38
rs989713632 Chr.3: 58666687-58666687 on Build GRCh38
rs995028856 Chr.3: 58666586-58666586 on Build GRCh38
rs3749290 Chr.3: 58666565-58666565 on Build GRCh38
rs1026144144 Chr.3: 58666611-58666611 on Build GRCh38
rs939589130 Chr.3: 58666533-58666533 on Build GRCh38
rs780108415 Chr.3: 58666766-58666766 on Build GRCh38
rs1023724252 Chr.3: 58666960-58666960 on Build GRCh38
rs867808299 Chr.3: 58666644-58666644 on Build GRCh38
rs768546519 Chr.3: 58666782-58666782 on Build GRCh38
rs886200903 Chr.3: 58666809-58666809 on Build GRCh38
rs1050375582 Chr.3: 58666821-58666821 on Build GRCh38
rs546274647 Chr.3: 58666873-58666873 on Build GRCh38
rs769608296 Chr.3: 58666707-58666707 on Build GRCh38
rs1031180914 Chr.3: 58666639-58666639 on Build GRCh38
rs72877788 Chr.3: 58666728-58666728 on Build GRCh38
rs1008804303 Chr.3: 58666641-58666641 on Build GRCh38
rs999546910 Chr.3: 58666870-58666870 on Build GRCh38
rs924121140 Chr.3: 58666731-58666731 on Build GRCh38
rs960780400 Chr.3: 58666878-58666878 on Build GRCh38
rs780617910 Chr.3: 58666577-58666577 on Build GRCh38
rs920293789 Chr.3: 58666781-58666781 on Build GRCh38
rs978556427 Chr.3: 58666638-58666638 on Build GRCh38
rs777536028 Chr.3: 58666817-58666817 on Build GRCh38
rs989599369 Chr.3: 58666908-58666908 on Build GRCh38
rs895417416 Chr.3: 58666555-58666555 on Build GRCh38
rs1022754392 Chr.3: 58666727-58666727 on Build GRCh38
rs566091957 Chr.3: 58666907-58666907 on Build GRCh38
rs994771402 Chr.3: 58666526-58666526 on Build GRCh38
rs1008778109 Chr.3: 58666857-58666857 on Build GRCh38
rs1008709463 Chr.3: 58666814-58666814 on Build GRCh38
rs996538409 Chr.3: 58666840-58666840 on Build GRCh38
rs542078773 Chr.3: 58666643-58666643 on Build GRCh38
rs745399631 Chr.3: 58666617-58666617 on Build GRCh38
rs992252847 Chr.3: 58666879-58666879 on Build GRCh38
rs140473065 Chr.3: 58666777-58666777 on Build GRCh38
rs992305181 Chr.3: 58666747-58666747 on Build GRCh38
rs772426799 Chr.3: 58666869-58666869 on Build GRCh38
rs889817214 Chr.3: 58666538-58666538 on Build GRCh38
rs1047962092 Chr.3: 58666798-58666798 on Build GRCh38
rs1022278658 Chr.3: 58666841-58666841 on Build GRCh38
rs964638981 Chr.3: 58666701-58666701 on Build GRCh38
rs534971853 Chr.3: 58666914-58666914 on Build GRCh38
rs1044457886 Chr.3: 58666568-58666568 on Build GRCh38

View Full Product Details