Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 62465276-62465792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADPS
Entrez Gene ID
8618
Gene Name
calcium dependent secretion activator
Gene Aliases
CADPS1, CAPS, CAPS1, UNC-31
UniGene
Hs.654933
Gene Chromosome Location
Chr.3: 62398346-62875389 on Build GRCh38
Transcripts
NM_003716.3 NM_183393.2 NM_183394.2 XM_006713378.2 XM_011534177.1 XM_011534178.1 XM_011534180.1 XM_011534182.1 XM_011534183.1 XM_011534189.1 XM_011534191.2 XM_011534192.1 XM_011534193.1 XM_011534194.2 XM_011534195.1 XM_011534196.1 XM_011534197.2 XM_011534198.2 XM_011534199.2 XM_011534200.2 XM_011534203.1 XM_017007358.1 XM_017007359.1 XM_017007360.1 XM_017007361.1 XM_017007362.1 XM_017007363.1 XM_017007364.1 XM_017007365.1 XM_017007366.1 XM_017007367.1 XM_017007368.1 XM_017007369.1 XM_017007370.1 XM_017007371.1 XM_017007372.1 XM_017007373.1 XM_017007374.1 XM_017007375.1 XM_017007376.1 XM_017007377.1 XM_017007378.1 XM_017007379.1 XM_017007380.1 XM_017007381.1 XM_017007382.1 XM_017007383.1 XM_017007384.1 XM_017007385.1 XM_017007386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4119736 c.3519C>T p.T1173T Chr.3: 62465367-62465367 on Build GRCh38
4668838 c.3566_3567insT p.L1189fs*12 Chr.3: 62465436-62465437 on Build GRCh38
2783092 c.3555C>T p.F1185F Chr.3: 62465448-62465448 on Build GRCh38
5680771 c.3475A>T p.R1159* Chr.3: 62465411-62465411 on Build GRCh38
4878988 c.3556G>A p.V1186I Chr.3: 62465447-62465447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772693397 Chr.3: 62465441-62465441 on Build GRCh38
rs776898923 Chr.3: 62465322-62465322 on Build GRCh38
rs907629178 Chr.3: 62465717-62465717 on Build GRCh38
rs757466325 Chr.3: 62465364-62465364 on Build GRCh38
rs370366004 Chr.3: 62465315-62465315 on Build GRCh38
rs577766565 Chr.3: 62465519-62465519 on Build GRCh38
rs984533193 Chr.3: 62465670-62465670 on Build GRCh38
rs62243467 Chr.3: 62465367-62465367 on Build GRCh38
rs992148685 Chr.3: 62465542-62465542 on Build GRCh38
rs904027656 Chr.3: 62465498-62465498 on Build GRCh38
rs376866233 Chr.3: 62465403-62465403 on Build GRCh38
rs940378954 Chr.3: 62465372-62465372 on Build GRCh38
rs929453886 Chr.3: 62465559-62465559 on Build GRCh38
rs980047260 Chr.3: 62465564-62465564 on Build GRCh38
rs759466232 Chr.3: 62465762-62465762 on Build GRCh38
rs557374462 Chr.3: 62465382-62465382 on Build GRCh38
rs762889047 Chr.3: 62465492-62465492 on Build GRCh38
rs771644515 Chr.3: 62465439-62465439 on Build GRCh38
rs539896281 Chr.3: 62465637-62465637 on Build GRCh38
rs767703694 Chr.3: 62465424-62465424 on Build GRCh38
rs573123399 Chr.3: 62465630-62465630 on Build GRCh38
rs1039029755 Chr.3: 62465636-62465636 on Build GRCh38
rs1056608257 Chr.3: 62465549-62465549 on Build GRCh38
rs968811961 Chr.3: 62465522-62465522 on Build GRCh38
rs750984032 Chr.3: 62465471-62465471 on Build GRCh38
rs558377212 Chr.3: 62465685-62465685 on Build GRCh38
rs997440269 Chr.3: 62465669-62465669 on Build GRCh38
rs576910766 Chr.3: 62465725-62465725 on Build GRCh38
rs760114957 Chr.3: 62465330-62465330 on Build GRCh38
rs758736576 Chr.3: 62465470-62465471 on Build GRCh38
rs887857576 Chr.3: 62465584-62465584 on Build GRCh38
rs776530791 Chr.3: 62465533-62465533 on Build GRCh38
rs181384985 Chr.3: 62465544-62465544 on Build GRCh38
rs148264169 Chr.3: 62465476-62465476 on Build GRCh38
rs758553825 Chr.3: 62465349-62465349 on Build GRCh38
rs776168275 Chr.3: 62465479-62465479 on Build GRCh38
rs764177763 Chr.3: 62465355-62465355 on Build GRCh38
rs971298314 Chr.3: 62465554-62465554 on Build GRCh38
rs747591987 Chr.3: 62465430-62465430 on Build GRCh38
rs902090459 Chr.3: 62465402-62465402 on Build GRCh38
rs1006670925 Chr.3: 62465609-62465609 on Build GRCh38
rs927111998 Chr.3: 62465590-62465590 on Build GRCh38
rs955821270 Chr.3: 62465486-62465486 on Build GRCh38
rs979778520 Chr.3: 62465537-62465537 on Build GRCh38
rs1021821539 Chr.3: 62465740-62465740 on Build GRCh38
rs1048303129 Chr.3: 62465572-62465572 on Build GRCh38
rs754901644 Chr.3: 62465377-62465377 on Build GRCh38
rs940472431 Chr.3: 62465727-62465727 on Build GRCh38
rs770354818 Chr.3: 62465477-62465477 on Build GRCh38
rs766790318 Chr.3: 62465316-62465316 on Build GRCh38
rs938268278 Chr.3: 62465545-62465545 on Build GRCh38
rs1030223315 Chr.3: 62465687-62465687 on Build GRCh38
rs968490203 Chr.3: 62465763-62465763 on Build GRCh38
rs371358789 Chr.3: 62465356-62465356 on Build GRCh38
rs765902736 Chr.3: 62465343-62465343 on Build GRCh38
rs752766770 Chr.3: 62465348-62465348 on Build GRCh38
rs536583073 Chr.3: 62465532-62465532 on Build GRCh38
rs770554516 Chr.3: 62465448-62465448 on Build GRCh38
rs574220713 Chr.3: 62465447-62465447 on Build GRCh38
rs779870268 Chr.3: 62465366-62465366 on Build GRCh38
rs777122141 Chr.3: 62465450-62465450 on Build GRCh38
rs1000919246 Chr.3: 62465499-62465499 on Build GRCh38
rs951972666 Chr.3: 62465591-62465591 on Build GRCh38
rs761920713 Chr.3: 62465381-62465381 on Build GRCh38
rs1040574477 Chr.3: 62465400-62465400 on Build GRCh38

View Full Product Details